Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33342900-33343377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNAR1
Entrez Gene ID
3454
Gene Name
interferon alpha and beta receptor subunit 1
Gene Aliases
AVP, IFN-alpha-REC, IFNAR, IFNBR, IFRC
UniGene
Hs.529400
Gene Chromosome Location
Chr.21: 33324443-33360361 on Build GRCh38
Transcripts
NM_000629.2 XM_005260964.2 XM_011529552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
444386 c.410C>T p.A137V Chr.21: 33343301-33343301 on Build GRCh38
4923657 c.463A>T p.M155L Chr.21: 33343354-33343354 on Build GRCh38
6190167 c.479G>T p.G160V Chr.21: 33343370-33343370 on Build GRCh38
6569592 c.412G>A p.E138K Chr.21: 33343303-33343303 on Build GRCh38
1234981 c.395A>G p.E132G Chr.21: 33343286-33343286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531380680 Chr.21: 33342957-33342957 on Build GRCh38
rs1010647768 Chr.21: 33343291-33343291 on Build GRCh38
rs202161321 Chr.21: 33343257-33343257 on Build GRCh38
rs753923384 Chr.21: 33343354-33343354 on Build GRCh38
rs1019671723 Chr.21: 33343020-33343020 on Build GRCh38
rs985402903 Chr.21: 33343017-33343017 on Build GRCh38
rs889670559 Chr.21: 33343213-33343213 on Build GRCh38
rs1042645483 Chr.21: 33343156-33343156 on Build GRCh38
rs529994436 Chr.21: 33343094-33343094 on Build GRCh38
rs62226369 Chr.21: 33343061-33343061 on Build GRCh38
rs772951896 Chr.21: 33343282-33343282 on Build GRCh38
rs550618264 Chr.21: 33343007-33343007 on Build GRCh38
rs569860128 Chr.21: 33343220-33343220 on Build GRCh38
rs111291706 Chr.21: 33343272-33343272 on Build GRCh38
rs953189203 Chr.21: 33343335-33343335 on Build GRCh38
rs1032237664 Chr.21: 33343212-33343212 on Build GRCh38
rs764529643 Chr.21: 33343324-33343324 on Build GRCh38
rs150805896 Chr.21: 33343322-33343322 on Build GRCh38
rs1012411247 Chr.21: 33343148-33343148 on Build GRCh38
rs965885404 Chr.21: 33343266-33343266 on Build GRCh38
rs761347916 Chr.21: 33343273-33343273 on Build GRCh38
rs373257370 Chr.21: 33343307-33343307 on Build GRCh38
rs1032185313 Chr.21: 33342956-33342956 on Build GRCh38
rs549022719 Chr.21: 33343280-33343280 on Build GRCh38
rs918761175 Chr.21: 33343044-33343044 on Build GRCh38
rs368102237 Chr.21: 33343039-33343039 on Build GRCh38
rs905447164 Chr.21: 33343230-33343230 on Build GRCh38
rs375511312 Chr.21: 33343222-33343222 on Build GRCh38
rs895303184 Chr.21: 33343092-33343092 on Build GRCh38
rs188755098 Chr.21: 33343053-33343053 on Build GRCh38
rs532616623 Chr.21: 33343297-33343297 on Build GRCh38
rs548550739 Chr.21: 33343180-33343180 on Build GRCh38
rs1000200998 Chr.21: 33343210-33343210 on Build GRCh38
rs1000781831 Chr.21: 33342933-33342933 on Build GRCh38
rs896247267 Chr.21: 33343124-33343124 on Build GRCh38
rs530076063 Chr.21: 33343214-33343214 on Build GRCh38
rs746934323 Chr.21: 33343256-33343257 on Build GRCh38
rs759868066 Chr.21: 33343288-33343288 on Build GRCh38
rs538192301 Chr.21: 33343250-33343250 on Build GRCh38
rs937680743 Chr.21: 33343058-33343058 on Build GRCh38
rs972834492 Chr.21: 33343295-33343295 on Build GRCh38
rs906752416 Chr.21: 33343158-33343158 on Build GRCh38
rs779044975 Chr.21: 33343246-33343246 on Build GRCh38
rs984382786 Chr.21: 33343047-33343047 on Build GRCh38
rs753212923 Chr.21: 33343308-33343308 on Build GRCh38
rs193278382 Chr.21: 33343255-33343255 on Build GRCh38
rs564949379 Chr.21: 33343034-33343034 on Build GRCh38
rs373897082 Chr.21: 33343038-33343038 on Build GRCh38
rs183815122 Chr.21: 33343021-33343021 on Build GRCh38
rs954047410 Chr.21: 33342970-33342970 on Build GRCh38
rs949159065 Chr.21: 33343142-33343142 on Build GRCh38
rs746931314 Chr.21: 33343264-33343264 on Build GRCh38
rs757487290 Chr.21: 33343228-33343228 on Build GRCh38
rs931593956 Chr.21: 33343045-33343045 on Build GRCh38
rs967066271 Chr.21: 33342926-33342926 on Build GRCh38
rs1001067428 Chr.21: 33343208-33343208 on Build GRCh38
rs768114021 Chr.21: 33343268-33343268 on Build GRCh38
rs1028780709 Chr.21: 33343293-33343293 on Build GRCh38

View Full Product Details