Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18244876-18245460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLII
Entrez Gene ID
2314
Gene Name
FLII, actin remodeling protein
Gene Aliases
FLI, FLIL, Fli1
UniGene
Hs.513984
Gene Chromosome Location
Chr.17: 18244815-18258916 on Build GRCh38
Transcripts
NM_001256264.1 NM_001256265.1 NM_002018.3 XM_005256555.3 XM_005256556.3 XM_005256558.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
976261 c.3672C>T p.C1224C Chr.17: 18245357-18245357 on Build GRCh38
6404970 c.3625G>T p.G1209W Chr.17: 18245404-18245404 on Build GRCh38
704823 c.3783G>C p.W1261C Chr.17: 18245165-18245165 on Build GRCh38
976260 c.3713G>A p.R1238Q Chr.17: 18245235-18245235 on Build GRCh38
2740879 c.3695G>A p.R1232Q Chr.17: 18245253-18245253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377010006 Chr.17: 18245289-18245289 on Build GRCh38
rs774605767 Chr.17: 18245249-18245249 on Build GRCh38
rs770448874 Chr.17: 18245207-18245207 on Build GRCh38
rs765582634 Chr.17: 18245030-18245030 on Build GRCh38
rs567357608 Chr.17: 18245031-18245031 on Build GRCh38
rs764239209 Chr.17: 18245336-18245336 on Build GRCh38
rs566174133 Chr.17: 18245349-18245349 on Build GRCh38
rs780385408 Chr.17: 18245037-18245037 on Build GRCh38
rs780939355 Chr.17: 18245364-18245364 on Build GRCh38
rs746327361 Chr.17: 18245319-18245319 on Build GRCh38
rs201341928 Chr.17: 18245170-18245170 on Build GRCh38
rs73980862 Chr.17: 18244910-18244910 on Build GRCh38
rs370069054 Chr.17: 18245300-18245300 on Build GRCh38
rs769162401 Chr.17: 18245056-18245056 on Build GRCh38
rs368495750 Chr.17: 18245229-18245229 on Build GRCh38
rs968863288 Chr.17: 18245204-18245204 on Build GRCh38
rs377076526 Chr.17: 18245182-18245182 on Build GRCh38
rs779710734 Chr.17: 18245239-18245239 on Build GRCh38
rs746716074 Chr.17: 18245291-18245291 on Build GRCh38
rs144058499 Chr.17: 18245227-18245227 on Build GRCh38
rs758698251 Chr.17: 18245035-18245035 on Build GRCh38
rs201144517 Chr.17: 18245337-18245337 on Build GRCh38
rs556343793 Chr.17: 18245344-18245344 on Build GRCh38
rs756042691 Chr.17: 18245088-18245088 on Build GRCh38
rs201912251 Chr.17: 18245235-18245235 on Build GRCh38
rs753001586 Chr.17: 18245125-18245126 on Build GRCh38
rs1029504988 Chr.17: 18245216-18245216 on Build GRCh38
rs750759339 Chr.17: 18245034-18245034 on Build GRCh38
rs758495014 Chr.17: 18245350-18245351 on Build GRCh38
rs761833143 Chr.17: 18245343-18245343 on Build GRCh38
rs1038171912 Chr.17: 18245110-18245110 on Build GRCh38
rs567421136 Chr.17: 18245296-18245296 on Build GRCh38
rs780405351 Chr.17: 18245394-18245394 on Build GRCh38
rs753560467 Chr.17: 18245390-18245390 on Build GRCh38
rs746447108 Chr.17: 18245386-18245386 on Build GRCh38
rs772992287 Chr.17: 18245079-18245079 on Build GRCh38
rs964274602 Chr.17: 18244926-18244926 on Build GRCh38
rs774869065 Chr.17: 18245200-18245200 on Build GRCh38
rs1029061728 Chr.17: 18245383-18245383 on Build GRCh38
rs765772556 Chr.17: 18245123-18245123 on Build GRCh38
rs746621494 Chr.17: 18245240-18245240 on Build GRCh38
rs761422711 Chr.17: 18245048-18245049 on Build GRCh38
rs956176354 Chr.17: 18244948-18244948 on Build GRCh38
rs905533773 Chr.17: 18244939-18244939 on Build GRCh38
rs774864298 Chr.17: 18245137-18245138 on Build GRCh38
rs771654215 Chr.17: 18245237-18245237 on Build GRCh38
rs559178771 Chr.17: 18245339-18245339 on Build GRCh38
rs778820625 Chr.17: 18245298-18245299 on Build GRCh38
rs750208218 Chr.17: 18245276-18245276 on Build GRCh38
rs563225941 Chr.17: 18245230-18245230 on Build GRCh38
rs773105809 Chr.17: 18245419-18245419 on Build GRCh38
rs774966965 Chr.17: 18245316-18245316 on Build GRCh38
rs750416517 Chr.17: 18245346-18245346 on Build GRCh38
rs147081178 Chr.17: 18245231-18245231 on Build GRCh38
rs939849008 Chr.17: 18244908-18244908 on Build GRCh38
rs777635942 Chr.17: 18245185-18245185 on Build GRCh38
rs963413917 Chr.17: 18245380-18245380 on Build GRCh38
rs765181810 Chr.17: 18245274-18245274 on Build GRCh38
rs974021418 Chr.17: 18245246-18245246 on Build GRCh38
rs532381790 Chr.17: 18244914-18244914 on Build GRCh38
rs766818378 Chr.17: 18245153-18245153 on Build GRCh38
rs374452292 Chr.17: 18245226-18245226 on Build GRCh38
rs898273921 Chr.17: 18244924-18244924 on Build GRCh38
rs991598259 Chr.17: 18245355-18245355 on Build GRCh38
rs200226967 Chr.17: 18245116-18245116 on Build GRCh38
rs759891827 Chr.17: 18245252-18245252 on Build GRCh38
rs745569501 Chr.17: 18245193-18245193 on Build GRCh38
rs148683920 Chr.17: 18245322-18245322 on Build GRCh38
rs1003087994 Chr.17: 18244942-18244943 on Build GRCh38
rs750231703 Chr.17: 18244937-18244937 on Build GRCh38
rs375634815 Chr.17: 18245097-18245097 on Build GRCh38
rs764218959 Chr.17: 18245257-18245257 on Build GRCh38
rs755687641 Chr.17: 18245376-18245376 on Build GRCh38
rs11653817 Chr.17: 18245156-18245156 on Build GRCh38
rs569997820 Chr.17: 18245331-18245331 on Build GRCh38
rs767386496 Chr.17: 18245437-18245437 on Build GRCh38
rs780853576 Chr.17: 18245297-18245297 on Build GRCh38
rs199914435 Chr.17: 18245241-18245241 on Build GRCh38
rs185087719 Chr.17: 18245191-18245191 on Build GRCh38
rs752751809 Chr.17: 18245181-18245181 on Build GRCh38
rs149231616 Chr.17: 18245194-18245194 on Build GRCh38
rs374445084 Chr.17: 18244902-18244902 on Build GRCh38
rs374226495 Chr.17: 18245347-18245347 on Build GRCh38
rs769713788 Chr.17: 18245063-18245063 on Build GRCh38
rs781326145 Chr.17: 18245049-18245049 on Build GRCh38
rs757382701 Chr.17: 18245280-18245280 on Build GRCh38
rs1039741700 Chr.17: 18245305-18245305 on Build GRCh38
rs565835176 Chr.17: 18244971-18244971 on Build GRCh38
rs973969069 Chr.17: 18245393-18245393 on Build GRCh38
rs373435657 Chr.17: 18245288-18245288 on Build GRCh38
rs752460534 Chr.17: 18245439-18245439 on Build GRCh38
rs773906779 Chr.17: 18245209-18245209 on Build GRCh38
rs769984657 Chr.17: 18245415-18245415 on Build GRCh38
rs62072502 Chr.17: 18245420-18245420 on Build GRCh38
rs767647295 Chr.17: 18244903-18244903 on Build GRCh38
rs369821133 Chr.17: 18245247-18245247 on Build GRCh38
rs8821 Chr.17: 18245220-18245220 on Build GRCh38
rs193075988 Chr.17: 18244988-18244988 on Build GRCh38
rs753843419 Chr.17: 18245232-18245232 on Build GRCh38
rs547009769 Chr.17: 18245192-18245192 on Build GRCh38
rs777141885 Chr.17: 18245378-18245378 on Build GRCh38
rs748093461 Chr.17: 18245053-18245053 on Build GRCh38
rs201843497 Chr.17: 18245315-18245315 on Build GRCh38
rs758264349 Chr.17: 18245280-18245280 on Build GRCh38
rs552089359 Chr.17: 18244966-18244966 on Build GRCh38
rs764627492 Chr.17: 18245147-18245148 on Build GRCh38
rs1022663198 Chr.17: 18244934-18244934 on Build GRCh38
rs759199212 Chr.17: 18245101-18245101 on Build GRCh38
rs990251962 Chr.17: 18244969-18244969 on Build GRCh38
rs771551666 Chr.17: 18245111-18245111 on Build GRCh38
rs146409718 Chr.17: 18245186-18245186 on Build GRCh38
rs749826930 Chr.17: 18245242-18245242 on Build GRCh38
rs967399911 Chr.17: 18245050-18245050 on Build GRCh38
rs368156305 Chr.17: 18245357-18245357 on Build GRCh38
rs759448466 Chr.17: 18245432-18245432 on Build GRCh38
rs755052400 Chr.17: 18245044-18245044 on Build GRCh38
rs766906354 Chr.17: 18245212-18245212 on Build GRCh38
rs761013563 Chr.17: 18245133-18245136 on Build GRCh38
rs146460796 Chr.17: 18245406-18245406 on Build GRCh38
rs747108812 Chr.17: 18245043-18245043 on Build GRCh38
rs775707771 Chr.17: 18245330-18245330 on Build GRCh38
rs765209806 Chr.17: 18245345-18245345 on Build GRCh38
rs751422060 Chr.17: 18245431-18245431 on Build GRCh38
rs747338163 Chr.17: 18245402-18245402 on Build GRCh38
rs751793927 Chr.17: 18245159-18245159 on Build GRCh38
rs138406094 Chr.17: 18245236-18245236 on Build GRCh38
rs758978391 Chr.17: 18245310-18245310 on Build GRCh38
rs11382 Chr.17: 18244960-18244960 on Build GRCh38
rs376127631 Chr.17: 18245133-18245133 on Build GRCh38
rs931016077 Chr.17: 18245142-18245142 on Build GRCh38
rs374095780 Chr.17: 18245254-18245254 on Build GRCh38
rs143389844 Chr.17: 18245201-18245201 on Build GRCh38
rs986706801 Chr.17: 18245090-18245090 on Build GRCh38
rs767871060 Chr.17: 18245110-18245111 on Build GRCh38
rs576669385 Chr.17: 18245144-18245144 on Build GRCh38
rs752372422 Chr.17: 18245369-18245369 on Build GRCh38
rs753861761 Chr.17: 18245267-18245267 on Build GRCh38
rs143462111 Chr.17: 18245424-18245424 on Build GRCh38
rs11540885 Chr.17: 18245004-18245004 on Build GRCh38
rs200799100 Chr.17: 18245162-18245162 on Build GRCh38
rs754321107 Chr.17: 18245218-18245226 on Build GRCh38
rs534056531 Chr.17: 18244984-18244984 on Build GRCh38
rs763548959 Chr.17: 18245211-18245211 on Build GRCh38
rs535221206 Chr.17: 18245387-18245387 on Build GRCh38
rs763314144 Chr.17: 18245066-18245067 on Build GRCh38
rs763401145 Chr.17: 18245361-18245361 on Build GRCh38
rs369182133 Chr.17: 18245270-18245270 on Build GRCh38
rs529765136 Chr.17: 18245077-18245078 on Build GRCh38
rs780414188 Chr.17: 18245437-18245438 on Build GRCh38
rs928649369 Chr.17: 18245055-18245055 on Build GRCh38
rs749037386 Chr.17: 18245082-18245082 on Build GRCh38
rs139025109 Chr.17: 18245412-18245412 on Build GRCh38
rs761992745 Chr.17: 18245413-18245413 on Build GRCh38
rs1008479238 Chr.17: 18244904-18244904 on Build GRCh38
rs919756583 Chr.17: 18245264-18245264 on Build GRCh38
rs547527028 Chr.17: 18245256-18245256 on Build GRCh38
rs139780458 Chr.17: 18245100-18245100 on Build GRCh38
rs755706636 Chr.17: 18245441-18245441 on Build GRCh38
rs7498 Chr.17: 18245171-18245171 on Build GRCh38
rs1036368997 Chr.17: 18244948-18244949 on Build GRCh38
rs142904883 Chr.17: 18245441-18245442 on Build GRCh38
rs759682057 Chr.17: 18245221-18245221 on Build GRCh38
rs143892517 Chr.17: 18245253-18245253 on Build GRCh38

View Full Product Details