Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128915957-128916519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC8A
Entrez Gene ID
56262
Gene Name
leucine rich repeat containing 8 family member A
Gene Aliases
AGM5, LRRC8, SWELL1
UniGene
Hs.643600
Gene Chromosome Location
Chr.9: 128882112-128918042 on Build GRCh38
Transcripts
NM_001127244.1 NM_001127245.1 NM_019594.3 XM_005252095.2 XM_005252096.3 XM_006717186.2 XM_011518863.1 XM_011518864.2 XM_011518865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4699177 c.2278G>A p.E760K Chr.9: 128916216-128916216 on Build GRCh38
6435441 c.2383C>T p.P795S Chr.9: 128916321-128916321 on Build GRCh38
6115056 c.2160C>T p.I720I Chr.9: 128916098-128916098 on Build GRCh38
6425065 c.2363A>T p.E788V Chr.9: 128916301-128916301 on Build GRCh38
4941650 c.2293C>T p.R765W Chr.9: 128916231-128916231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760903402 Chr.9: 128916223-128916223 on Build GRCh38
rs763795899 Chr.9: 128916291-128916291 on Build GRCh38
rs541442315 Chr.9: 128916362-128916362 on Build GRCh38
rs994349524 Chr.9: 128916044-128916044 on Build GRCh38
rs566168299 Chr.9: 128916093-128916093 on Build GRCh38
rs754714008 Chr.9: 128916402-128916402 on Build GRCh38
rs986716086 Chr.9: 128916040-128916040 on Build GRCh38
rs952562964 Chr.9: 128916458-128916458 on Build GRCh38
rs764169809 Chr.9: 128916114-128916114 on Build GRCh38
rs377544052 Chr.9: 128916368-128916368 on Build GRCh38
rs780941988 Chr.9: 128916242-128916242 on Build GRCh38
rs768916187 Chr.9: 128916259-128916259 on Build GRCh38
rs973663730 Chr.9: 128916486-128916486 on Build GRCh38
rs1019714666 Chr.9: 128916426-128916426 on Build GRCh38
rs561749062 Chr.9: 128916406-128916406 on Build GRCh38
rs577481072 Chr.9: 128916260-128916260 on Build GRCh38
rs531989271 Chr.9: 128916255-128916255 on Build GRCh38
rs530321477 Chr.9: 128916379-128916379 on Build GRCh38
rs1037508831 Chr.9: 128916064-128916064 on Build GRCh38
rs986158748 Chr.9: 128916222-128916222 on Build GRCh38
rs572943400 Chr.9: 128916359-128916359 on Build GRCh38
rs747449292 Chr.9: 128916361-128916361 on Build GRCh38
rs1007863938 Chr.9: 128916425-128916425 on Build GRCh38
rs763672970 Chr.9: 128916235-128916235 on Build GRCh38
rs777304358 Chr.9: 128916366-128916366 on Build GRCh38
rs373487932 Chr.9: 128916209-128916209 on Build GRCh38
rs200873510 Chr.9: 128916188-128916188 on Build GRCh38
rs775809202 Chr.9: 128916112-128916112 on Build GRCh38
rs753526503 Chr.9: 128916060-128916060 on Build GRCh38
rs766782363 Chr.9: 128916226-128916226 on Build GRCh38
rs980163301 Chr.9: 128916477-128916477 on Build GRCh38
rs377425028 Chr.9: 128916459-128916459 on Build GRCh38
rs774080181 Chr.9: 128916373-128916373 on Build GRCh38
rs374629783 Chr.9: 128916340-128916340 on Build GRCh38
rs961278274 Chr.9: 128916436-128916436 on Build GRCh38
rs376840917 Chr.9: 128916399-128916399 on Build GRCh38
rs552348824 Chr.9: 128916073-128916073 on Build GRCh38
rs749787818 Chr.9: 128916152-128916152 on Build GRCh38
rs767057883 Chr.9: 128916311-128916311 on Build GRCh38
rs748386966 Chr.9: 128916344-128916344 on Build GRCh38
rs750320909 Chr.9: 128916323-128916323 on Build GRCh38
rs777181644 Chr.9: 128916284-128916284 on Build GRCh38
rs752227637 Chr.9: 128916231-128916231 on Build GRCh38
rs781005769 Chr.9: 128916178-128916178 on Build GRCh38
rs1022229958 Chr.9: 128916206-128916206 on Build GRCh38
rs767858656 Chr.9: 128916132-128916132 on Build GRCh38
rs554988954 Chr.9: 128916165-128916165 on Build GRCh38
rs761252884 Chr.9: 128916263-128916263 on Build GRCh38
rs977933538 Chr.9: 128916182-128916182 on Build GRCh38
rs150808591 Chr.9: 128916003-128916003 on Build GRCh38
rs760500008 Chr.9: 128916392-128916392 on Build GRCh38
rs1021450617 Chr.9: 128916391-128916391 on Build GRCh38
rs199588534 Chr.9: 128916338-128916338 on Build GRCh38
rs771624809 Chr.9: 128916238-128916238 on Build GRCh38
rs73672303 Chr.9: 128916479-128916479 on Build GRCh38
rs369716517 Chr.9: 128916254-128916254 on Build GRCh38
rs769948961 Chr.9: 128916105-128916105 on Build GRCh38
rs751995060 Chr.9: 128916130-128916130 on Build GRCh38
rs530504978 Chr.9: 128916174-128916174 on Build GRCh38
rs761728966 Chr.9: 128916445-128916445 on Build GRCh38
rs927334972 Chr.9: 128916478-128916478 on Build GRCh38
rs767950230 Chr.9: 128916215-128916215 on Build GRCh38
rs759926958 Chr.9: 128916367-128916367 on Build GRCh38
rs371629774 Chr.9: 128916339-128916339 on Build GRCh38
rs1043197355 Chr.9: 128916380-128916380 on Build GRCh38
rs746959164 Chr.9: 128916082-128916082 on Build GRCh38
rs868086143 Chr.9: 128916103-128916103 on Build GRCh38
rs919749885 Chr.9: 128916453-128916453 on Build GRCh38
rs140168683 Chr.9: 128916147-128916147 on Build GRCh38
rs773803974 Chr.9: 128916216-128916216 on Build GRCh38
rs561567188 Chr.9: 128916327-128916327 on Build GRCh38
rs778467791 Chr.9: 128916071-128916071 on Build GRCh38
rs910713857 Chr.9: 128915991-128915991 on Build GRCh38
rs557710052 Chr.9: 128916208-128916208 on Build GRCh38
rs200535573 Chr.9: 128916192-128916192 on Build GRCh38
rs751274880 Chr.9: 128916296-128916296 on Build GRCh38
rs898219618 Chr.9: 128916041-128916041 on Build GRCh38
rs768719966 Chr.9: 128916202-128916202 on Build GRCh38
rs1051157869 Chr.9: 128915988-128915988 on Build GRCh38
rs759696740 Chr.9: 128916162-128916162 on Build GRCh38
rs754648799 Chr.9: 128916070-128916070 on Build GRCh38
rs749515686 Chr.9: 128916199-128916199 on Build GRCh38
rs761348206 Chr.9: 128916309-128916309 on Build GRCh38
rs146661687 Chr.9: 128916261-128916261 on Build GRCh38
rs770843536 Chr.9: 128916087-128916087 on Build GRCh38
rs1057093265 Chr.9: 128916487-128916487 on Build GRCh38
rs747651882 Chr.9: 128916072-128916072 on Build GRCh38
rs756136520 Chr.9: 128916326-128916326 on Build GRCh38
rs759781334 Chr.9: 128916098-128916098 on Build GRCh38
rs377654430 Chr.9: 128916232-128916232 on Build GRCh38
rs766650861 Chr.9: 128916280-128916280 on Build GRCh38
rs773537614 Chr.9: 128916262-128916262 on Build GRCh38
rs766921702 Chr.9: 128916377-128916377 on Build GRCh38
rs139178076 Chr.9: 128916161-128916161 on Build GRCh38
rs376518155 Chr.9: 128916045-128916045 on Build GRCh38
rs954821870 Chr.9: 128916450-128916450 on Build GRCh38
rs973068968 Chr.9: 128916443-128916443 on Build GRCh38
rs762888887 Chr.9: 128916113-128916113 on Build GRCh38
rs754631786 Chr.9: 128916148-128916148 on Build GRCh38
rs373135213 Chr.9: 128916285-128916285 on Build GRCh38
rs1037838815 Chr.9: 128916012-128916012 on Build GRCh38
rs201931744 Chr.9: 128916047-128916047 on Build GRCh38
rs774017858 Chr.9: 128916374-128916374 on Build GRCh38
rs771981247 Chr.9: 128916350-128916350 on Build GRCh38
rs762377121 Chr.9: 128916131-128916131 on Build GRCh38
rs781543533 Chr.9: 128916053-128916053 on Build GRCh38
rs745610311 Chr.9: 128916179-128916179 on Build GRCh38
rs544964004 Chr.9: 128916183-128916183 on Build GRCh38
rs766265457 Chr.9: 128916396-128916396 on Build GRCh38
rs757926178 Chr.9: 128916154-128916154 on Build GRCh38
rs777805122 Chr.9: 128916078-128916078 on Build GRCh38
rs938686617 Chr.9: 128916341-128916341 on Build GRCh38
rs752079691 Chr.9: 128916046-128916046 on Build GRCh38

View Full Product Details