Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 61996245-61996739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT16
Entrez Gene ID
83851
Gene Name
synaptotagmin 16
Gene Aliases
CHR14SYT, SYT14L, Strep14, syt14r, yt14r
UniGene
Hs.404139
Gene Chromosome Location
Chr.14: 61812156-62112825 on Build GRCh38
Transcripts
NM_031914.2 XM_006720272.3 XM_011537219.1 XM_011537220.1 XM_011537221.2 XM_011537222.2 XM_011537223.2 XM_011537224.2 XM_011537225.2 XM_011537228.2 XM_017021693.1 XM_017021694.1 XM_017021695.1 XM_017021696.1 XM_017021697.1 XM_017021698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3496722 c.368G>A p.W123* Chr.14: 61996387-61996387 on Build GRCh38
3496718 c.289G>A p.D97N Chr.14: 61996308-61996308 on Build GRCh38
5726932 c.463C>A p.Q155K Chr.14: 61996482-61996482 on Build GRCh38
6795474 c.391C>T p.R131C Chr.14: 61996410-61996410 on Build GRCh38
1193687 c.497T>C p.L166S Chr.14: 61996516-61996516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745381339 Chr.14: 61996548-61996548 on Build GRCh38
rs766321009 Chr.14: 61996512-61996512 on Build GRCh38
rs932112028 Chr.14: 61996294-61996294 on Build GRCh38
rs1056470383 Chr.14: 61996705-61996705 on Build GRCh38
rs1057241395 Chr.14: 61996649-61996649 on Build GRCh38
rs754024359 Chr.14: 61996288-61996288 on Build GRCh38
rs199644885 Chr.14: 61996404-61996404 on Build GRCh38
rs770010275 Chr.14: 61996577-61996577 on Build GRCh38
rs903774672 Chr.14: 61996423-61996423 on Build GRCh38
rs751700527 Chr.14: 61996516-61996516 on Build GRCh38
rs747013440 Chr.14: 61996326-61996326 on Build GRCh38
rs764397012 Chr.14: 61996394-61996394 on Build GRCh38
rs572646326 Chr.14: 61996655-61996655 on Build GRCh38
rs747953042 Chr.14: 61996440-61996440 on Build GRCh38
rs758568904 Chr.14: 61996313-61996313 on Build GRCh38
rs761013827 Chr.14: 61996409-61996409 on Build GRCh38
rs555721038 Chr.14: 61996677-61996677 on Build GRCh38
rs947925930 Chr.14: 61996650-61996650 on Build GRCh38
rs1044214949 Chr.14: 61996479-61996479 on Build GRCh38
rs762344414 Chr.14: 61996573-61996573 on Build GRCh38
rs764486632 Chr.14: 61996280-61996280 on Build GRCh38
rs769771492 Chr.14: 61996441-61996441 on Build GRCh38
rs778221963 Chr.14: 61996547-61996547 on Build GRCh38
rs61743830 Chr.14: 61996571-61996571 on Build GRCh38
rs568329310 Chr.14: 61996531-61996531 on Build GRCh38
rs1023539130 Chr.14: 61996414-61996414 on Build GRCh38
rs751411372 Chr.14: 61996433-61996433 on Build GRCh38
rs1000801268 Chr.14: 61996695-61996695 on Build GRCh38
rs781263032 Chr.14: 61996524-61996524 on Build GRCh38
rs769159158 Chr.14: 61996478-61996478 on Build GRCh38
rs985810040 Chr.14: 61996656-61996656 on Build GRCh38
rs911112161 Chr.14: 61996663-61996663 on Build GRCh38
rs372814062 Chr.14: 61996337-61996337 on Build GRCh38
rs909503797 Chr.14: 61996563-61996563 on Build GRCh38
rs1036362664 Chr.14: 61996356-61996356 on Build GRCh38
rs767394508 Chr.14: 61996714-61996714 on Build GRCh38
rs564353410 Chr.14: 61996393-61996393 on Build GRCh38
rs777655325 Chr.14: 61996532-61996532 on Build GRCh38
rs201658725 Chr.14: 61996466-61996466 on Build GRCh38
rs894552664 Chr.14: 61996649-61996650 on Build GRCh38
rs759359796 Chr.14: 61996562-61996562 on Build GRCh38
rs368589747 Chr.14: 61996357-61996357 on Build GRCh38
rs759843963 Chr.14: 61996485-61996485 on Build GRCh38
rs897781206 Chr.14: 61996373-61996373 on Build GRCh38
rs773648307 Chr.14: 61996578-61996578 on Build GRCh38
rs192369127 Chr.14: 61996539-61996539 on Build GRCh38
rs777186420 Chr.14: 61996484-61996484 on Build GRCh38
rs575891770 Chr.14: 61996300-61996300 on Build GRCh38
rs200774034 Chr.14: 61996457-61996457 on Build GRCh38
rs575874193 Chr.14: 61996678-61996678 on Build GRCh38
rs376295336 Chr.14: 61996437-61996437 on Build GRCh38
rs17099370 Chr.14: 61996411-61996411 on Build GRCh38
rs1023868943 Chr.14: 61996541-61996541 on Build GRCh38
rs755942696 Chr.14: 61996528-61996528 on Build GRCh38
rs552601273 Chr.14: 61996628-61996628 on Build GRCh38
rs938659397 Chr.14: 61996598-61996598 on Build GRCh38
rs62639692 Chr.14: 61996593-61996593 on Build GRCh38
rs745937291 Chr.14: 61996303-61996303 on Build GRCh38
rs61743836 Chr.14: 61996585-61996585 on Build GRCh38
rs1029374869 Chr.14: 61996597-61996597 on Build GRCh38
rs924037133 Chr.14: 61996681-61996681 on Build GRCh38
rs1016678221 Chr.14: 61996385-61996385 on Build GRCh38
rs777268976 Chr.14: 61996442-61996442 on Build GRCh38
rs533386682 Chr.14: 61996410-61996410 on Build GRCh38
rs752732394 Chr.14: 61996525-61996525 on Build GRCh38
rs768539483 Chr.14: 61996329-61996329 on Build GRCh38
rs757495690 Chr.14: 61996297-61996297 on Build GRCh38
rs765678840 Chr.14: 61996495-61996495 on Build GRCh38
rs748847458 Chr.14: 61996538-61996538 on Build GRCh38
rs1012426255 Chr.14: 61996306-61996306 on Build GRCh38
rs371001125 Chr.14: 61996609-61996609 on Build GRCh38
rs200307441 Chr.14: 61996381-61996381 on Build GRCh38
rs887969321 Chr.14: 61996570-61996570 on Build GRCh38
rs756413004 Chr.14: 61996278-61996278 on Build GRCh38
rs183703376 Chr.14: 61996668-61996668 on Build GRCh38
rs762359257 Chr.14: 61996366-61996366 on Build GRCh38
rs763266467 Chr.14: 61996500-61996500 on Build GRCh38
rs551712196 Chr.14: 61996469-61996469 on Build GRCh38
rs142424272 Chr.14: 61996486-61996486 on Build GRCh38
rs896481412 Chr.14: 61996508-61996508 on Build GRCh38
rs189094610 Chr.14: 61996698-61996698 on Build GRCh38
rs544551861 Chr.14: 61996708-61996708 on Build GRCh38
rs374422747 Chr.14: 61996352-61996352 on Build GRCh38
rs771625061 Chr.14: 61996554-61996554 on Build GRCh38
rs761115481 Chr.14: 61996477-61996477 on Build GRCh38
rs765438180 Chr.14: 61996402-61996402 on Build GRCh38
rs1035256170 Chr.14: 61996287-61996287 on Build GRCh38
rs201321447 Chr.14: 61996336-61996336 on Build GRCh38
rs754894786 Chr.14: 61996519-61996519 on Build GRCh38
rs1889806 Chr.14: 61996496-61996496 on Build GRCh38
rs1024255866 Chr.14: 61996315-61996315 on Build GRCh38
rs756758496 Chr.14: 61996587-61996587 on Build GRCh38
rs868036207 Chr.14: 61996284-61996284 on Build GRCh38
rs779838284 Chr.14: 61996319-61996319 on Build GRCh38
rs367967436 Chr.14: 61996426-61996426 on Build GRCh38
rs757407684 Chr.14: 61996398-61996398 on Build GRCh38

View Full Product Details