Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28935005-28935602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD19
Entrez Gene ID
930
Gene Name
CD19 molecule
Gene Aliases
B4, CVID3
UniGene
Hs.652262
Gene Chromosome Location
Chr.16: 28931735-28939347 on Build GRCh38
Transcripts
NM_001178098.1 NM_001770.5 XM_006721103.3 XM_011545981.1 XM_017023893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5436644 c.917C>T p.S306F Chr.16: 28935525-28935525 on Build GRCh38
6304189 c.875T>C p.V292A Chr.16: 28935483-28935483 on Build GRCh38
3508529 c.870G>A p.W290* Chr.16: 28935478-28935478 on Build GRCh38
7082972 c.855G>A p.L285L Chr.16: 28935463-28935463 on Build GRCh38
6623503 c.866G>A p.G289D Chr.16: 28935474-28935474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766807192 Chr.16: 28935405-28935405 on Build GRCh38
rs571379580 Chr.16: 28935205-28935205 on Build GRCh38
rs1038359985 Chr.16: 28935192-28935192 on Build GRCh38
rs764120868 Chr.16: 28935575-28935575 on Build GRCh38
rs529388925 Chr.16: 28935115-28935117 on Build GRCh38
rs934487210 Chr.16: 28935087-28935087 on Build GRCh38
rs946042363 Chr.16: 28935211-28935211 on Build GRCh38
rs565796110 Chr.16: 28935127-28935127 on Build GRCh38
rs978463842 Chr.16: 28935271-28935271 on Build GRCh38
rs925822937 Chr.16: 28935301-28935301 on Build GRCh38
rs994062300 Chr.16: 28935203-28935203 on Build GRCh38
rs761593552 Chr.16: 28935397-28935397 on Build GRCh38
rs567435257 Chr.16: 28935325-28935325 on Build GRCh38
rs899417603 Chr.16: 28935427-28935427 on Build GRCh38
rs926584896 Chr.16: 28935342-28935342 on Build GRCh38
rs988695709 Chr.16: 28935102-28935102 on Build GRCh38
rs747751841 Chr.16: 28935539-28935539 on Build GRCh38
rs551427196 Chr.16: 28935160-28935160 on Build GRCh38
rs913962347 Chr.16: 28935119-28935119 on Build GRCh38
rs900288218 Chr.16: 28935557-28935557 on Build GRCh38
rs758204626 Chr.16: 28935484-28935484 on Build GRCh38
rs143951804 Chr.16: 28935508-28935508 on Build GRCh38
rs557406713 Chr.16: 28935465-28935465 on Build GRCh38
rs933715369 Chr.16: 28935335-28935335 on Build GRCh38
rs920828865 Chr.16: 28935390-28935390 on Build GRCh38
rs925770380 Chr.16: 28935061-28935061 on Build GRCh38
rs942477987 Chr.16: 28935042-28935042 on Build GRCh38
rs375531253 Chr.16: 28935448-28935448 on Build GRCh38
rs768176830 Chr.16: 28935497-28935497 on Build GRCh38
rs769587423 Chr.16: 28935569-28935569 on Build GRCh38
rs1017249152 Chr.16: 28935244-28935244 on Build GRCh38
rs769166684 Chr.16: 28935036-28935037 on Build GRCh38
rs547630306 Chr.16: 28935103-28935103 on Build GRCh38
rs997968195 Chr.16: 28935091-28935091 on Build GRCh38
rs938009075 Chr.16: 28935370-28935370 on Build GRCh38
rs960102219 Chr.16: 28935181-28935181 on Build GRCh38
rs776396802 Chr.16: 28935396-28935396 on Build GRCh38
rs764208673 Chr.16: 28935462-28935462 on Build GRCh38
rs776044371 Chr.16: 28935571-28935571 on Build GRCh38
rs538847317 Chr.16: 28935239-28935239 on Build GRCh38
rs1016488134 Chr.16: 28935153-28935153 on Build GRCh38
rs757503904 Chr.16: 28935467-28935467 on Build GRCh38
rs555411661 Chr.16: 28935324-28935324 on Build GRCh38
rs949441726 Chr.16: 28935331-28935331 on Build GRCh38
rs556348270 Chr.16: 28935167-28935167 on Build GRCh38
rs1005647727 Chr.16: 28935137-28935137 on Build GRCh38
rs978684599 Chr.16: 28935060-28935060 on Build GRCh38
rs745692453 Chr.16: 28935482-28935482 on Build GRCh38
rs967870735 Chr.16: 28935570-28935570 on Build GRCh38
rs761126860 Chr.16: 28935573-28935573 on Build GRCh38
rs537075106 Chr.16: 28935246-28935246 on Build GRCh38
rs905214455 Chr.16: 28935166-28935166 on Build GRCh38
rs774788019 Chr.16: 28935564-28935564 on Build GRCh38
rs533260074 Chr.16: 28935136-28935136 on Build GRCh38
rs1034514199 Chr.16: 28935171-28935171 on Build GRCh38
rs964377598 Chr.16: 28935266-28935266 on Build GRCh38
rs754194345 Chr.16: 28935326-28935326 on Build GRCh38
rs1038801404 Chr.16: 28935481-28935481 on Build GRCh38
rs746025937 Chr.16: 28935537-28935537 on Build GRCh38
rs1026509181 Chr.16: 28935229-28935229 on Build GRCh38
rs1046786733 Chr.16: 28935344-28935344 on Build GRCh38
rs756204094 Chr.16: 28935454-28935454 on Build GRCh38
rs553531692 Chr.16: 28935376-28935376 on Build GRCh38
rs942634737 Chr.16: 28935313-28935313 on Build GRCh38
rs1033335608 Chr.16: 28935316-28935316 on Build GRCh38
rs938230398 Chr.16: 28935180-28935180 on Build GRCh38
rs751992145 Chr.16: 28935420-28935420 on Build GRCh38
rs201149069 Chr.16: 28935421-28935421 on Build GRCh38
rs1039561622 Chr.16: 28935042-28935044 on Build GRCh38
rs534846444 Chr.16: 28935336-28935336 on Build GRCh38
rs890180764 Chr.16: 28935234-28935234 on Build GRCh38
rs780040144 Chr.16: 28935491-28935491 on Build GRCh38
rs778632114 Chr.16: 28935472-28935472 on Build GRCh38
rs571699553 Chr.16: 28935199-28935199 on Build GRCh38
rs371868433 Chr.16: 28935431-28935431 on Build GRCh38
rs900918422 Chr.16: 28935066-28935066 on Build GRCh38
rs769580431 Chr.16: 28935550-28935550 on Build GRCh38
rs746981179 Chr.16: 28935496-28935496 on Build GRCh38
rs759940827 Chr.16: 28935566-28935566 on Build GRCh38

View Full Product Details