Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56970947-56971461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CETP
Entrez Gene ID
1071
Gene Name
cholesteryl ester transfer protein
Gene Aliases
BPIFF, HDLCQ10
UniGene
Hs.89538
Gene Chromosome Location
Chr.16: 56961923-56983844 on Build GRCh38
Transcripts
NM_000078.2 NM_001286085.1 XM_006721124.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3999881 c.658+8C>T p.? Chr.16: 56971389-56971389 on Build GRCh38
971752 c.550T>C p.F184L Chr.16: 56971055-56971055 on Build GRCh38
1200873 c.635C>T p.A212V Chr.16: 56971358-56971358 on Build GRCh38
230321 c.566C>T p.S189F Chr.16: 56971071-56971071 on Build GRCh38
1302045 c.600C>G p.I200M Chr.16: 56971323-56971323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756059330 Chr.16: 56971390-56971390 on Build GRCh38
rs777256524 Chr.16: 56971383-56971383 on Build GRCh38
rs756541175 Chr.16: 56971083-56971083 on Build GRCh38
rs34523084 Chr.16: 56971251-56971251 on Build GRCh38
rs759428504 Chr.16: 56971002-56971002 on Build GRCh38
rs756599078 Chr.16: 56971303-56971303 on Build GRCh38
rs1532625 Chr.16: 56971389-56971389 on Build GRCh38
rs759224437 Chr.16: 56971123-56971123 on Build GRCh38
rs766881417 Chr.16: 56971367-56971367 on Build GRCh38
rs377464648 Chr.16: 56971001-56971001 on Build GRCh38
rs764223617 Chr.16: 56971037-56971037 on Build GRCh38
rs200704071 Chr.16: 56971020-56971020 on Build GRCh38
rs200426013 Chr.16: 56971299-56971299 on Build GRCh38
rs5885 Chr.16: 56971359-56971359 on Build GRCh38
rs774703022 Chr.16: 56971342-56971342 on Build GRCh38
rs550677685 Chr.16: 56971377-56971377 on Build GRCh38
rs780356088 Chr.16: 56971305-56971305 on Build GRCh38
rs747624840 Chr.16: 56970982-56970982 on Build GRCh38
rs757986542 Chr.16: 56971311-56971311 on Build GRCh38
rs138670739 Chr.16: 56971324-56971324 on Build GRCh38
rs376863212 Chr.16: 56970991-56970991 on Build GRCh38
rs754185945 Chr.16: 56971374-56971374 on Build GRCh38
rs767808237 Chr.16: 56971263-56971263 on Build GRCh38
rs373617305 Chr.16: 56971090-56971090 on Build GRCh38
rs34611098 Chr.16: 56971039-56971039 on Build GRCh38
rs372446155 Chr.16: 56971386-56971386 on Build GRCh38
rs780715695 Chr.16: 56971388-56971388 on Build GRCh38
rs148134355 Chr.16: 56970999-56970999 on Build GRCh38
rs751537902 Chr.16: 56971153-56971153 on Build GRCh38
rs373779560 Chr.16: 56971337-56971337 on Build GRCh38
rs774037270 Chr.16: 56971408-56971408 on Build GRCh38
rs772187360 Chr.16: 56971422-56971422 on Build GRCh38
rs953417499 Chr.16: 56971228-56971228 on Build GRCh38
rs12708973 Chr.16: 56970990-56970990 on Build GRCh38
rs761776694 Chr.16: 56971356-56971356 on Build GRCh38
rs751277957 Chr.16: 56971384-56971384 on Build GRCh38
rs190556780 Chr.16: 56971238-56971238 on Build GRCh38
rs12708972 Chr.16: 56970980-56970980 on Build GRCh38
rs775360565 Chr.16: 56971339-56971339 on Build GRCh38
rs773152860 Chr.16: 56971075-56971075 on Build GRCh38
rs142750310 Chr.16: 56971049-56971049 on Build GRCh38
rs768987111 Chr.16: 56971341-56971341 on Build GRCh38
rs1020430971 Chr.16: 56971148-56971148 on Build GRCh38
rs757640381 Chr.16: 56971043-56971043 on Build GRCh38
rs780200801 Chr.16: 56971394-56971394 on Build GRCh38
rs769887743 Chr.16: 56971338-56971338 on Build GRCh38
rs747748699 Chr.16: 56971087-56971087 on Build GRCh38
rs7205804 Chr.16: 56970977-56970977 on Build GRCh38
rs1016350654 Chr.16: 56971265-56971265 on Build GRCh38
rs775450632 Chr.16: 56971139-56971139 on Build GRCh38
rs368713153 Chr.16: 56970998-56970998 on Build GRCh38
rs770518166 Chr.16: 56971318-56971318 on Build GRCh38
rs146069143 Chr.16: 56971084-56971084 on Build GRCh38
rs368805891 Chr.16: 56971358-56971358 on Build GRCh38
rs1030375504 Chr.16: 56970976-56970976 on Build GRCh38
rs763230937 Chr.16: 56971151-56971151 on Build GRCh38
rs750986788 Chr.16: 56971293-56971293 on Build GRCh38
rs201267603 Chr.16: 56971308-56971308 on Build GRCh38
rs777194816 Chr.16: 56971103-56971103 on Build GRCh38
rs575830378 Chr.16: 56971152-56971152 on Build GRCh38
rs748361965 Chr.16: 56971415-56971415 on Build GRCh38
rs1053554564 Chr.16: 56971069-56971069 on Build GRCh38
rs771487390 Chr.16: 56971080-56971080 on Build GRCh38
rs771618514 Chr.16: 56970985-56970985 on Build GRCh38
rs370088702 Chr.16: 56971402-56971402 on Build GRCh38
rs201490558 Chr.16: 56971014-56971014 on Build GRCh38
rs767907136 Chr.16: 56971280-56971280 on Build GRCh38
rs758533363 Chr.16: 56971033-56971033 on Build GRCh38
rs557660317 Chr.16: 56971094-56971094 on Build GRCh38
rs771277935 Chr.16: 56971428-56971429 on Build GRCh38
rs147351998 Chr.16: 56971362-56971362 on Build GRCh38
rs370318351 Chr.16: 56971316-56971316 on Build GRCh38
rs769662260 Chr.16: 56971266-56971266 on Build GRCh38
rs149004351 Chr.16: 56970969-56970969 on Build GRCh38
rs773282847 Chr.16: 56971430-56971430 on Build GRCh38
rs371233223 Chr.16: 56971360-56971360 on Build GRCh38
rs546026764 Chr.16: 56971208-56971208 on Build GRCh38
rs113846146 Chr.16: 56971160-56971160 on Build GRCh38
rs763531115 Chr.16: 56971292-56971293 on Build GRCh38
rs762011312 Chr.16: 56971272-56971272 on Build GRCh38
rs770756667 Chr.16: 56971108-56971108 on Build GRCh38
rs769765149 Chr.16: 56971136-56971136 on Build GRCh38
rs746206629 Chr.16: 56971067-56971067 on Build GRCh38
rs570417790 Chr.16: 56971273-56971273 on Build GRCh38
rs376326645 Chr.16: 56971120-56971120 on Build GRCh38
rs746539100 Chr.16: 56971107-56971107 on Build GRCh38
rs777455962 Chr.16: 56971313-56971313 on Build GRCh38

View Full Product Details