Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32431933-32432492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFC
Entrez Gene ID
254240
Gene Name
BPI fold containing family C
Gene Aliases
BPIL2
UniGene
Hs.372939
Gene Chromosome Location
Chr.22: 32413845-32464465 on Build GRCh38
Transcripts
NM_174932.2 XM_011530088.2 XM_011530089.1 XM_011530090.1 XM_011530091.2 XM_017028740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5461028 c.1149+1G>A p.? Chr.22: 32432372-32432372 on Build GRCh38
6161892 c.1072C>A p.L358M Chr.22: 32432450-32432450 on Build GRCh38
6612389 c.1036C>A p.P346T Chr.22: 32432486-32432486 on Build GRCh38
3553759 c.1041C>T p.P347P Chr.22: 32432481-32432481 on Build GRCh38
3553758 c.1071C>T p.T357T Chr.22: 32432451-32432451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975289912 Chr.22: 32432398-32432398 on Build GRCh38
rs767097861 Chr.22: 32432324-32432324 on Build GRCh38
rs995368642 Chr.22: 32432335-32432335 on Build GRCh38
rs749052757 Chr.22: 32432463-32432463 on Build GRCh38
rs765809583 Chr.22: 32432347-32432347 on Build GRCh38
rs924392807 Chr.22: 32432444-32432444 on Build GRCh38
rs748814281 Chr.22: 32432176-32432176 on Build GRCh38
rs67451475 Chr.22: 32432014-32432015 on Build GRCh38
rs1023115484 Chr.22: 32432026-32432026 on Build GRCh38
rs187867571 Chr.22: 32432036-32432036 on Build GRCh38
rs193247551 Chr.22: 32432066-32432066 on Build GRCh38
rs758893216 Chr.22: 32432350-32432350 on Build GRCh38
rs768513502 Chr.22: 32432381-32432381 on Build GRCh38
rs567925163 Chr.22: 32432372-32432372 on Build GRCh38
rs373163358 Chr.22: 32432346-32432346 on Build GRCh38
rs926306 Chr.22: 32432085-32432085 on Build GRCh38
rs1001166216 Chr.22: 32432126-32432126 on Build GRCh38
rs200495148 Chr.22: 32432383-32432383 on Build GRCh38
rs548731557 Chr.22: 32432173-32432173 on Build GRCh38
rs185320000 Chr.22: 32432243-32432243 on Build GRCh38
rs545577736 Chr.22: 32432048-32432048 on Build GRCh38
rs754255627 Chr.22: 32432325-32432325 on Build GRCh38
rs1018789287 Chr.22: 32432470-32432470 on Build GRCh38
rs534844514 Chr.22: 32431967-32431967 on Build GRCh38
rs960257210 Chr.22: 32432162-32432162 on Build GRCh38
rs140684884 Chr.22: 32432447-32432447 on Build GRCh38
rs554549497 Chr.22: 32431972-32431972 on Build GRCh38
rs144222119 Chr.22: 32432425-32432425 on Build GRCh38
rs552712677 Chr.22: 32432469-32432469 on Build GRCh38
rs905527091 Chr.22: 32432115-32432115 on Build GRCh38
rs1010434807 Chr.22: 32432163-32432163 on Build GRCh38
rs1042805456 Chr.22: 32432224-32432224 on Build GRCh38
rs778814237 Chr.22: 32432465-32432465 on Build GRCh38
rs1043140698 Chr.22: 32431956-32431956 on Build GRCh38
rs150852332 Chr.22: 32432379-32432379 on Build GRCh38
rs397952508 Chr.22: 32432015-32432016 on Build GRCh38
rs934347311 Chr.22: 32432238-32432238 on Build GRCh38
rs963478607 Chr.22: 32432060-32432060 on Build GRCh38
rs899323076 Chr.22: 32432314-32432314 on Build GRCh38
rs74566641 Chr.22: 32431958-32431958 on Build GRCh38
rs562906643 Chr.22: 32432015-32432015 on Build GRCh38
rs62241169 Chr.22: 32432112-32432112 on Build GRCh38
rs749213600 Chr.22: 32432380-32432380 on Build GRCh38
rs574353918 Chr.22: 32431979-32431979 on Build GRCh38
rs5998490 Chr.22: 32432221-32432221 on Build GRCh38
rs9609551 Chr.22: 32432356-32432356 on Build GRCh38
rs1020861524 Chr.22: 32432168-32432168 on Build GRCh38
rs761458287 Chr.22: 32432323-32432323 on Build GRCh38
rs1047587850 Chr.22: 32432319-32432319 on Build GRCh38
rs934382507 Chr.22: 32432453-32432453 on Build GRCh38
rs968579277 Chr.22: 32432421-32432421 on Build GRCh38
rs1047535369 Chr.22: 32432069-32432069 on Build GRCh38
rs769444574 Chr.22: 32432377-32432377 on Build GRCh38
rs933224475 Chr.22: 32432292-32432292 on Build GRCh38
rs757410307 Chr.22: 32432428-32432428 on Build GRCh38
rs1013155742 Chr.22: 32432276-32432276 on Build GRCh38
rs753070287 Chr.22: 32432417-32432417 on Build GRCh38
rs139576658 Chr.22: 32432442-32432442 on Build GRCh38
rs535592103 Chr.22: 32432357-32432357 on Build GRCh38
rs781607711 Chr.22: 32432435-32432435 on Build GRCh38
rs554298521 Chr.22: 32432363-32432363 on Build GRCh38
rs867672436 Chr.22: 32432228-32432228 on Build GRCh38
rs546755313 Chr.22: 32432349-32432349 on Build GRCh38
rs973498927 Chr.22: 32432063-32432063 on Build GRCh38
rs759990056 Chr.22: 32432393-32432393 on Build GRCh38
rs1035944616 Chr.22: 32432010-32432010 on Build GRCh38
rs779637608 Chr.22: 32432128-32432128 on Build GRCh38
rs539865781 Chr.22: 32432291-32432291 on Build GRCh38
rs754964446 Chr.22: 32432464-32432464 on Build GRCh38
rs756312151 Chr.22: 32432446-32432446 on Build GRCh38
rs1037409565 Chr.22: 32432098-32432098 on Build GRCh38
rs896088099 Chr.22: 32432255-32432255 on Build GRCh38
rs765539718 Chr.22: 32432409-32432409 on Build GRCh38
rs912484476 Chr.22: 32432114-32432114 on Build GRCh38
rs976693305 Chr.22: 32432245-32432245 on Build GRCh38
rs370577187 Chr.22: 32432387-32432387 on Build GRCh38
rs200868839 Chr.22: 32432373-32432373 on Build GRCh38
rs764640606 Chr.22: 32432418-32432418 on Build GRCh38
rs755826899 Chr.22: 32432099-32432099 on Build GRCh38
rs58094336 Chr.22: 32432013-32432014 on Build GRCh38
rs759812639 Chr.22: 32432327-32432327 on Build GRCh38
rs999548193 Chr.22: 32431994-32431994 on Build GRCh38
rs562291935 Chr.22: 32432179-32432179 on Build GRCh38
rs543317637 Chr.22: 32432005-32432005 on Build GRCh38
rs923159064 Chr.22: 32431989-32431989 on Build GRCh38
rs879126459 Chr.22: 32432275-32432275 on Build GRCh38
rs568188138 Chr.22: 32432133-32432133 on Build GRCh38
rs748867998 Chr.22: 32432370-32432371 on Build GRCh38
rs1006375528 Chr.22: 32432096-32432096 on Build GRCh38
rs754420954 Chr.22: 32432204-32432204 on Build GRCh38
rs117216213 Chr.22: 32432388-32432388 on Build GRCh38
rs9609550 Chr.22: 32432237-32432237 on Build GRCh38
rs528665581 Chr.22: 32432145-32432145 on Build GRCh38

View Full Product Details