Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 51336277-51336775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT10
Entrez Gene ID
170685
Gene Name
nudix hydrolase 10
Gene Aliases
APS2, DIPP3-alpha, DIPP3a
UniGene
Hs.375178
Gene Chromosome Location
Chr.X: 51332231-51337525 on Build GRCh38
Transcripts
NM_001304963.1 NM_153183.3

SNP Detail

SNP ID Chromosome Location
rs960751357 Chr.X: 51336666-51336666 on Build GRCh38
rs185054598 Chr.X: 51336524-51336524 on Build GRCh38
rs944867507 Chr.X: 51336459-51336459 on Build GRCh38
rs782272319 Chr.X: 51336720-51336720 on Build GRCh38
rs1040530318 Chr.X: 51336490-51336490 on Build GRCh38
rs996427688 Chr.X: 51336502-51336502 on Build GRCh38
rs138821237 Chr.X: 51336420-51336420 on Build GRCh38
rs1005398968 Chr.X: 51336592-51336592 on Build GRCh38
rs782581225 Chr.X: 51336388-51336388 on Build GRCh38
rs782486419 Chr.X: 51336303-51336303 on Build GRCh38
rs782419829 Chr.X: 51336442-51336442 on Build GRCh38
rs35094808 Chr.X: 51336487-51336488 on Build GRCh38
rs782228640 Chr.X: 51336353-51336353 on Build GRCh38
rs900757819 Chr.X: 51336501-51336501 on Build GRCh38
rs979072320 Chr.X: 51336324-51336324 on Build GRCh38
rs1049560429 Chr.X: 51336503-51336503 on Build GRCh38
rs782607817 Chr.X: 51336583-51336583 on Build GRCh38
rs150189068 Chr.X: 51336383-51336383 on Build GRCh38
rs1013383330 Chr.X: 51336712-51336712 on Build GRCh38
rs1015026087 Chr.X: 51336658-51336658 on Build GRCh38

View Full Product Details