Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101884671-101885171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMAT1
Entrez Gene ID
84460
Gene Name
zinc finger matrin-type 1
Gene Aliases
-
UniGene
Hs.496512
Gene Chromosome Location
Chr.X: 101882288-101932135 on Build GRCh38
Transcripts
NM_001011657.3 NM_001282400.1 NM_001282401.1 XM_005262212.4 XM_005262213.4 XM_005262214.3 XM_005262216.4 XM_006724711.3 XM_017029903.1 XM_017029904.1

SNP Detail

SNP ID Chromosome Location
rs775359769 Chr.X: 101884965-101884965 on Build GRCh38
rs994196586 Chr.X: 101884741-101884741 on Build GRCh38
rs201830609 Chr.X: 101884750-101884750 on Build GRCh38
rs41298508 Chr.X: 101884959-101884959 on Build GRCh38
rs955913020 Chr.X: 101885084-101885084 on Build GRCh38
rs369778966 Chr.X: 101885065-101885065 on Build GRCh38
rs376911023 Chr.X: 101884788-101884788 on Build GRCh38
rs966484743 Chr.X: 101885053-101885053 on Build GRCh38
rs774470980 Chr.X: 101884836-101884836 on Build GRCh38
rs751004141 Chr.X: 101884856-101884856 on Build GRCh38
rs768367195 Chr.X: 101884731-101884731 on Build GRCh38
rs772266798 Chr.X: 101884781-101884781 on Build GRCh38
rs776011100 Chr.X: 101884811-101884811 on Build GRCh38
rs1026378658 Chr.X: 101884902-101884902 on Build GRCh38
rs754374119 Chr.X: 101884790-101884790 on Build GRCh38
rs753095147 Chr.X: 101884832-101884832 on Build GRCh38
rs761952893 Chr.X: 101884780-101884780 on Build GRCh38
rs960913728 Chr.X: 101885126-101885126 on Build GRCh38
rs1021957890 Chr.X: 101885048-101885048 on Build GRCh38
rs758010281 Chr.X: 101884843-101884843 on Build GRCh38
rs988846333 Chr.X: 101885135-101885135 on Build GRCh38
rs188291609 Chr.X: 101884929-101884929 on Build GRCh38
rs964601623 Chr.X: 101885014-101885014 on Build GRCh38
rs112942988 Chr.X: 101884844-101884844 on Build GRCh38
rs760026531 Chr.X: 101884814-101884814 on Build GRCh38
rs765537721 Chr.X: 101884825-101884825 on Build GRCh38
rs375564650 Chr.X: 101885039-101885039 on Build GRCh38
rs151148157 Chr.X: 101884783-101884783 on Build GRCh38
rs180736110 Chr.X: 101884998-101884998 on Build GRCh38
rs1049816808 Chr.X: 101884734-101884734 on Build GRCh38
rs889987288 Chr.X: 101884782-101884782 on Build GRCh38
rs773218374 Chr.X: 101884827-101884829 on Build GRCh38
rs928542260 Chr.X: 101885076-101885076 on Build GRCh38
rs763863439 Chr.X: 101885073-101885073 on Build GRCh38
rs749998930 Chr.X: 101884848-101884848 on Build GRCh38
rs372330189 Chr.X: 101884866-101884866 on Build GRCh38
rs781098079 Chr.X: 101884911-101884911 on Build GRCh38
rs890291236 Chr.X: 101884807-101884807 on Build GRCh38
rs541988151 Chr.X: 101884869-101884869 on Build GRCh38
rs977866010 Chr.X: 101885057-101885057 on Build GRCh38
rs991366188 Chr.X: 101885145-101885145 on Build GRCh38
rs17282855 Chr.X: 101884786-101884786 on Build GRCh38
rs749196440 Chr.X: 101884706-101884706 on Build GRCh38
rs779426937 Chr.X: 101884930-101884933 on Build GRCh38
rs375316195 Chr.X: 101884913-101884913 on Build GRCh38
rs774127795 Chr.X: 101884754-101884754 on Build GRCh38

View Full Product Details