Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 16194278-16194790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR99AHG
Entrez Gene ID
388815
Gene Name
mir-99a-let-7c cluster host gene
Gene Aliases
C21orf34, C21orf35, LINC00478, MONC
UniGene
Hs.473394
Gene Chromosome Location
Chr.21: 16070488-16631727 on Build GRCh38
Transcripts
NR_027790.3 NR_027791.3 NR_111004.2 NR_111005.2 NR_136541.1 NR_136542.1 NR_136543.1 NR_136551.1 NR_136552.1

SNP Detail

SNP ID Chromosome Location
rs773601359 Chr.21: 16194421-16194421 on Build GRCh38
rs769692123 Chr.21: 16194600-16194600 on Build GRCh38
rs750208905 Chr.21: 16194659-16194659 on Build GRCh38
rs1034401863 Chr.21: 16194683-16194683 on Build GRCh38
rs139060431 Chr.21: 16194562-16194562 on Build GRCh38
rs62219500 Chr.21: 16194487-16194487 on Build GRCh38
rs944800477 Chr.21: 16194645-16194645 on Build GRCh38
rs766045965 Chr.21: 16194446-16194446 on Build GRCh38
rs776665828 Chr.21: 16194636-16194636 on Build GRCh38
rs1020804175 Chr.21: 16194386-16194386 on Build GRCh38
rs759309440 Chr.21: 16194613-16194613 on Build GRCh38
rs777488920 Chr.21: 16194471-16194471 on Build GRCh38
rs747117854 Chr.21: 16194402-16194402 on Build GRCh38
rs971971850 Chr.21: 16194624-16194624 on Build GRCh38
rs375268411 Chr.21: 16194641-16194641 on Build GRCh38
rs1009027037 Chr.21: 16194433-16194433 on Build GRCh38
rs947981268 Chr.21: 16194307-16194307 on Build GRCh38
rs755256703 Chr.21: 16194670-16194670 on Build GRCh38
rs144814897 Chr.21: 16194583-16194583 on Build GRCh38
rs756321550 Chr.21: 16194315-16194315 on Build GRCh38
rs6517692 Chr.21: 16194754-16194754 on Build GRCh38
rs768758742 Chr.21: 16194482-16194482 on Build GRCh38
rs749095415 Chr.21: 16194765-16194765 on Build GRCh38
rs774824521 Chr.21: 16194612-16194612 on Build GRCh38
rs749708888 Chr.21: 16194607-16194607 on Build GRCh38
rs771358102 Chr.21: 16194608-16194608 on Build GRCh38
rs773513158 Chr.21: 16194494-16194494 on Build GRCh38
rs899148610 Chr.21: 16194314-16194314 on Build GRCh38
rs764098185 Chr.21: 16194537-16194537 on Build GRCh38
rs1010831645 Chr.21: 16194610-16194610 on Build GRCh38
rs576987101 Chr.21: 16194587-16194587 on Build GRCh38
rs775499501 Chr.21: 16194398-16194398 on Build GRCh38
rs756935674 Chr.21: 16194477-16194477 on Build GRCh38
rs143151691 Chr.21: 16194470-16194470 on Build GRCh38
rs780727886 Chr.21: 16194468-16194468 on Build GRCh38
rs958689370 Chr.21: 16194424-16194424 on Build GRCh38
rs765633036 Chr.21: 16194420-16194420 on Build GRCh38
rs759339023 Chr.21: 16194583-16194583 on Build GRCh38
rs772327049 Chr.21: 16194378-16194378 on Build GRCh38
rs763375914 Chr.21: 16194495-16194495 on Build GRCh38
rs776737590 Chr.21: 16194529-16194529 on Build GRCh38
rs187727206 Chr.21: 16194367-16194367 on Build GRCh38
rs372893223 Chr.21: 16194539-16194539 on Build GRCh38
rs765130821 Chr.21: 16194653-16194653 on Build GRCh38
rs747187857 Chr.21: 16194478-16194478 on Build GRCh38
rs150750465 Chr.21: 16194561-16194561 on Build GRCh38
rs545893065 Chr.21: 16194602-16194602 on Build GRCh38
rs368620969 Chr.21: 16194453-16194453 on Build GRCh38
rs772760246 Chr.21: 16194460-16194460 on Build GRCh38
rs990744221 Chr.21: 16194443-16194443 on Build GRCh38
rs763464292 Chr.21: 16194635-16194635 on Build GRCh38
rs958699278 Chr.21: 16194716-16194716 on Build GRCh38
rs930482320 Chr.21: 16194462-16194462 on Build GRCh38
rs767729347 Chr.21: 16194676-16194676 on Build GRCh38
rs775219038 Chr.21: 16194620-16194620 on Build GRCh38
rs148215361 Chr.21: 16194486-16194486 on Build GRCh38
rs957541264 Chr.21: 16194354-16194354 on Build GRCh38
rs746127465 Chr.21: 16194377-16194377 on Build GRCh38
rs771847325 Chr.21: 16194614-16194614 on Build GRCh38
rs777583529 Chr.21: 16194372-16194372 on Build GRCh38
rs771139060 Chr.21: 16194503-16194503 on Build GRCh38
rs752854462 Chr.21: 16194677-16194677 on Build GRCh38
rs752243573 Chr.21: 16194526-16194526 on Build GRCh38
rs753665132 Chr.21: 16194476-16194476 on Build GRCh38
rs543404056 Chr.21: 16194509-16194509 on Build GRCh38
rs762569709 Chr.21: 16194429-16194429 on Build GRCh38
rs759130255 Chr.21: 16194455-16194455 on Build GRCh38
rs768668698 Chr.21: 16194410-16194410 on Build GRCh38
rs767174255 Chr.21: 16194520-16194520 on Build GRCh38
rs974047289 Chr.21: 16194744-16194744 on Build GRCh38
rs146380740 Chr.21: 16194628-16194628 on Build GRCh38
rs377556417 Chr.21: 16194411-16194411 on Build GRCh38
rs559434401 Chr.21: 16194719-16194719 on Build GRCh38
rs920472457 Chr.21: 16194451-16194451 on Build GRCh38
rs16994440 Chr.21: 16194416-16194416 on Build GRCh38
rs1027586535 Chr.21: 16194753-16194753 on Build GRCh38
rs1044380867 Chr.21: 16194313-16194313 on Build GRCh38
rs900209595 Chr.21: 16194514-16194514 on Build GRCh38

View Full Product Details