Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139282991-139283534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MATR3
Entrez Gene ID
9782
Gene Name
matrin 3
Gene Aliases
ALS21, MPD2, VCPDM
UniGene
Hs.268939
Gene Chromosome Location
Chr.5: 139273752-139331677 on Build GRCh38
Transcripts
NM_001194954.1 NM_001282278.1 NM_199189.2 NR_036536.1

Gene Symbol
SNHG4
Entrez Gene ID
724102
Gene Name
small nucleolar RNA host gene 4
Gene Aliases
NCRNA00059, U19H
UniGene
Hs.268939
Gene Chromosome Location
Chr.5: 139273752-139283184 on Build GRCh38
Transcripts
NM_001194954.1 NM_001282278.1 NM_199189.2 NR_036536.1

SNP Detail

SNP ID Chromosome Location
rs1055788227 Chr.5: 139283111-139283111 on Build GRCh38
rs937926736 Chr.5: 139283386-139283386 on Build GRCh38
rs185810220 Chr.5: 139283511-139283511 on Build GRCh38
rs909071114 Chr.5: 139283379-139283379 on Build GRCh38
rs138918626 Chr.5: 139283219-139283219 on Build GRCh38
rs562606716 Chr.5: 139283172-139283172 on Build GRCh38
rs550758794 Chr.5: 139283445-139283445 on Build GRCh38
rs1010784712 Chr.5: 139283295-139283295 on Build GRCh38
rs960624533 Chr.5: 139283066-139283066 on Build GRCh38
rs903274594 Chr.5: 139283506-139283506 on Build GRCh38
rs551212112 Chr.5: 139283017-139283017 on Build GRCh38
rs547333742 Chr.5: 139283315-139283315 on Build GRCh38
rs967959266 Chr.5: 139283383-139283383 on Build GRCh38
rs1045808686 Chr.5: 139283138-139283138 on Build GRCh38
rs535855139 Chr.5: 139283412-139283412 on Build GRCh38
rs988490903 Chr.5: 139283106-139283106 on Build GRCh38
rs896176129 Chr.5: 139283353-139283353 on Build GRCh38
rs773357051 Chr.5: 139283143-139283143 on Build GRCh38
rs1009230365 Chr.5: 139283360-139283360 on Build GRCh38
rs1001112484 Chr.5: 139283430-139283430 on Build GRCh38
rs991165023 Chr.5: 139283402-139283402 on Build GRCh38
rs368316344 Chr.5: 139283415-139283415 on Build GRCh38
rs766534885 Chr.5: 139283222-139283222 on Build GRCh38
rs919015853 Chr.5: 139283328-139283328 on Build GRCh38
rs777714314 Chr.5: 139283161-139283163 on Build GRCh38
rs756760755 Chr.5: 139283344-139283344 on Build GRCh38
rs984636514 Chr.5: 139283346-139283346 on Build GRCh38
rs944712295 Chr.5: 139283249-139283250 on Build GRCh38
rs1056212172 Chr.5: 139283310-139283310 on Build GRCh38
rs111987011 Chr.5: 139283497-139283497 on Build GRCh38
rs568437759 Chr.5: 139283411-139283411 on Build GRCh38
rs552777550 Chr.5: 139283247-139283247 on Build GRCh38
rs950064777 Chr.5: 139283442-139283442 on Build GRCh38
rs877826 Chr.5: 139283108-139283108 on Build GRCh38
rs148986318 Chr.5: 139283287-139283287 on Build GRCh38
rs1033636545 Chr.5: 139283209-139283209 on Build GRCh38
rs1021322446 Chr.5: 139283376-139283376 on Build GRCh38
rs904961039 Chr.5: 139283296-139283298 on Build GRCh38
rs756116199 Chr.5: 139283366-139283366 on Build GRCh38
rs953478862 Chr.5: 139283455-139283455 on Build GRCh38
rs79351473 Chr.5: 139283203-139283203 on Build GRCh38
rs979283133 Chr.5: 139283148-139283148 on Build GRCh38
rs1028091089 Chr.5: 139283451-139283451 on Build GRCh38
rs1014271102 Chr.5: 139283137-139283137 on Build GRCh38
rs919515704 Chr.5: 139283265-139283265 on Build GRCh38
rs878871683 Chr.5: 139283441-139283441 on Build GRCh38
rs909131914 Chr.5: 139283039-139283039 on Build GRCh38
rs1037045230 Chr.5: 139283259-139283259 on Build GRCh38
rs866601021 Chr.5: 139283129-139283129 on Build GRCh38
rs1001924838 Chr.5: 139283299-139283301 on Build GRCh38
rs74564730 Chr.5: 139283280-139283280 on Build GRCh38
rs1045755283 Chr.5: 139283485-139283485 on Build GRCh38
rs1001872371 Chr.5: 139283033-139283033 on Build GRCh38
rs903328509 Chr.5: 139283154-139283154 on Build GRCh38

View Full Product Details