Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 115888319-115888823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00474
Entrez Gene ID
58483
Gene Name
long intergenic non-protein coding RNA 474
Gene Aliases
C9orf27, EST-YD1
UniGene
Hs.23367
Gene Chromosome Location
Chr.9: 115888265-115925098 on Build GRCh38
Transcripts
NR_024032.1

SNP Detail

SNP ID Chromosome Location
rs576592278 Chr.9: 115888557-115888557 on Build GRCh38
rs927403371 Chr.9: 115888477-115888477 on Build GRCh38
rs540471302 Chr.9: 115888513-115888513 on Build GRCh38
rs755685735 Chr.9: 115888574-115888574 on Build GRCh38
rs145845710 Chr.9: 115888447-115888447 on Build GRCh38
rs997292671 Chr.9: 115888667-115888667 on Build GRCh38
rs965274566 Chr.9: 115888527-115888527 on Build GRCh38
rs888664054 Chr.9: 115888526-115888526 on Build GRCh38
rs567044342 Chr.9: 115888420-115888420 on Build GRCh38
rs62577151 Chr.9: 115888556-115888556 on Build GRCh38
rs956478641 Chr.9: 115888617-115888617 on Build GRCh38
rs938799809 Chr.9: 115888481-115888481 on Build GRCh38
rs930122815 Chr.9: 115888495-115888495 on Build GRCh38
rs1989775 Chr.9: 115888410-115888410 on Build GRCh38
rs900156235 Chr.9: 115888652-115888652 on Build GRCh38
rs909567826 Chr.9: 115888646-115888646 on Build GRCh38
rs955873676 Chr.9: 115888715-115888715 on Build GRCh38
rs969000724 Chr.9: 115888789-115888789 on Build GRCh38
rs13294725 Chr.9: 115888448-115888448 on Build GRCh38
rs373949079 Chr.9: 115888391-115888391 on Build GRCh38
rs2040485 Chr.9: 115888445-115888445 on Build GRCh38
rs114551699 Chr.9: 115888459-115888459 on Build GRCh38
rs1243 Chr.9: 115888374-115888374 on Build GRCh38
rs918388275 Chr.9: 115888619-115888619 on Build GRCh38
rs547219896 Chr.9: 115888411-115888411 on Build GRCh38
rs761893331 Chr.9: 115888399-115888399 on Build GRCh38
rs370281165 Chr.9: 115888422-115888422 on Build GRCh38
rs891875613 Chr.9: 115888444-115888444 on Build GRCh38
rs748991593 Chr.9: 115888678-115888678 on Build GRCh38
rs530981390 Chr.9: 115888723-115888723 on Build GRCh38
rs138769301 Chr.9: 115888745-115888745 on Build GRCh38
rs947685631 Chr.9: 115888656-115888656 on Build GRCh38
rs1048094423 Chr.9: 115888352-115888352 on Build GRCh38
rs1030066203 Chr.9: 115888669-115888669 on Build GRCh38
rs897353878 Chr.9: 115888492-115888492 on Build GRCh38
rs765668769 Chr.9: 115888400-115888400 on Build GRCh38
rs867441643 Chr.9: 115888600-115888600 on Build GRCh38
rs9408881 Chr.9: 115888502-115888502 on Build GRCh38
rs142691530 Chr.9: 115888794-115888794 on Build GRCh38
rs545131948 Chr.9: 115888623-115888623 on Build GRCh38
rs546041349 Chr.9: 115888719-115888719 on Build GRCh38
rs554891818 Chr.9: 115888487-115888487 on Build GRCh38
rs1003442019 Chr.9: 115888417-115888417 on Build GRCh38
rs551284714 Chr.9: 115888367-115888367 on Build GRCh38
rs1057131300 Chr.9: 115888488-115888488 on Build GRCh38
rs929697907 Chr.9: 115888344-115888344 on Build GRCh38
rs375825579 Chr.9: 115888624-115888624 on Build GRCh38
rs887471262 Chr.9: 115888365-115888365 on Build GRCh38

View Full Product Details