Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 38481549-38481723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIFR
Entrez Gene ID
3977
Gene Name
leukemia inhibitory factor receptor alpha
Gene Aliases
CD118, LIF-R, SJS2, STWS, SWS
UniGene
Hs.133421
Gene Chromosome Location
Chr.5: 38474963-38606290 on Build GRCh38
Transcripts
NM_001127671.1 NM_002310.5 XM_011514040.2 XM_011514042.2 XM_017009462.1 XM_017009463.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3776696 c.3185C>T p.S1062F Chr.5: 38481704-38481704 on Build GRCh38
1213344 c.3197G>A p.R1066Q Chr.5: 38481692-38481692 on Build GRCh38
6700986 c.3273_3274insT p.Q1092fs*>7 Chr.5: 38481615-38481616 on Build GRCh38
3920020 c.3194C>T p.S1065F Chr.5: 38481695-38481695 on Build GRCh38
33086 c.3204T>G p.F1068L Chr.5: 38481685-38481685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566182850 Chr.5: 38481599-38481599 on Build GRCh38
rs761993625 Chr.5: 38481594-38481594 on Build GRCh38
rs754371834 Chr.5: 38481610-38481610 on Build GRCh38
rs867316235 Chr.5: 38481666-38481666 on Build GRCh38
rs747346598 Chr.5: 38481584-38481584 on Build GRCh38
rs751758745 Chr.5: 38481591-38481591 on Build GRCh38
rs147058538 Chr.5: 38481668-38481668 on Build GRCh38
rs776513402 Chr.5: 38481657-38481658 on Build GRCh38
rs943207929 Chr.5: 38481650-38481650 on Build GRCh38
rs756927234 Chr.5: 38481625-38481625 on Build GRCh38
rs764112814 Chr.5: 38481585-38481585 on Build GRCh38
rs768761408 Chr.5: 38481672-38481672 on Build GRCh38
rs745601310 Chr.5: 38481638-38481638 on Build GRCh38
rs193180465 Chr.5: 38481637-38481637 on Build GRCh38
rs368698516 Chr.5: 38481692-38481692 on Build GRCh38
rs373142822 Chr.5: 38481600-38481600 on Build GRCh38
rs532441112 Chr.5: 38481622-38481622 on Build GRCh38
rs972795829 Chr.5: 38481601-38481605 on Build GRCh38
rs746641016 Chr.5: 38481624-38481624 on Build GRCh38
rs768637423 Chr.5: 38481606-38481609 on Build GRCh38
rs775530423 Chr.5: 38481644-38481644 on Build GRCh38
rs757874049 Chr.5: 38481620-38481620 on Build GRCh38
rs769716512 Chr.5: 38481639-38481639 on Build GRCh38
rs3729751 Chr.5: 38481601-38481601 on Build GRCh38
rs1017110423 Chr.5: 38481582-38481582 on Build GRCh38
rs780468954 Chr.5: 38481604-38481604 on Build GRCh38

View Full Product Details