Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 56445607-56445858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929415
Entrez Gene ID
101929415
Gene Name
uncharacterized LOC101929415
Gene Aliases
-
UniGene
Hs.701520
Gene Chromosome Location
Chr.8: 56445807-56552067 on Build GRCh38
Transcripts
NR_125813.1 NM_001135690.2

Gene Symbol
PENK
Entrez Gene ID
5179
Gene Name
proenkephalin
Gene Aliases
PE, PENK-A
UniGene
Hs.339831
Gene Chromosome Location
Chr.8: 56440954-56446734 on Build GRCh38
Transcripts
NR_125813.1 NM_001135690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3900782 c.115C>T p.R39C Chr.8: 56445839-56445839 on Build GRCh38
3663811 c.138G>A p.L46L Chr.8: 56445816-56445816 on Build GRCh38
5710986 c.107G>A p.R36H Chr.8: 56445847-56445847 on Build GRCh38
3650005 c.135C>T p.F45F Chr.8: 56445819-56445819 on Build GRCh38
7264018 c.108C>T p.R36R Chr.8: 56445846-56445846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755541493 Chr.8: 56445696-56445696 on Build GRCh38
rs764634140 Chr.8: 56445805-56445805 on Build GRCh38
rs1051342590 Chr.8: 56445757-56445757 on Build GRCh38
rs776782733 Chr.8: 56445642-56445642 on Build GRCh38
rs766521586 Chr.8: 56445729-56445729 on Build GRCh38
rs779874497 Chr.8: 56445766-56445766 on Build GRCh38
rs200535254 Chr.8: 56445820-56445820 on Build GRCh38
rs554539595 Chr.8: 56445718-56445718 on Build GRCh38
rs745588035 Chr.8: 56445703-56445703 on Build GRCh38
rs1051000225 Chr.8: 56445685-56445685 on Build GRCh38
rs768574909 Chr.8: 56445774-56445774 on Build GRCh38
rs548139394 Chr.8: 56445788-56445788 on Build GRCh38
rs139296396 Chr.8: 56445817-56445817 on Build GRCh38
rs745703015 Chr.8: 56445763-56445763 on Build GRCh38
rs750682014 Chr.8: 56445682-56445682 on Build GRCh38
rs751969004 Chr.8: 56445730-56445730 on Build GRCh38
rs76880651 Chr.8: 56445697-56445697 on Build GRCh38
rs547553118 Chr.8: 56445634-56445634 on Build GRCh38
rs942043198 Chr.8: 56445635-56445635 on Build GRCh38
rs767823159 Chr.8: 56445784-56445784 on Build GRCh38
rs758263909 Chr.8: 56445771-56445772 on Build GRCh38
rs553820920 Chr.8: 56445765-56445765 on Build GRCh38
rs774270619 Chr.8: 56445632-56445632 on Build GRCh38
rs770698214 Chr.8: 56445698-56445698 on Build GRCh38
rs756244707 Chr.8: 56445805-56445806 on Build GRCh38
rs777487876 Chr.8: 56445742-56445742 on Build GRCh38
rs755250875 Chr.8: 56445731-56445731 on Build GRCh38
rs182635123 Chr.8: 56445741-56445741 on Build GRCh38
rs3808634 Chr.8: 56445692-56445692 on Build GRCh38
rs796813214 Chr.8: 56445818-56445818 on Build GRCh38
rs746975349 Chr.8: 56445773-56445773 on Build GRCh38
rs754906486 Chr.8: 56445756-56445756 on Build GRCh38
rs759579740 Chr.8: 56445633-56445633 on Build GRCh38
rs1010560355 Chr.8: 56445667-56445667 on Build GRCh38
rs751865887 Chr.8: 56445690-56445690 on Build GRCh38
rs762963271 Chr.8: 56445779-56445779 on Build GRCh38
rs934199391 Chr.8: 56445753-56445753 on Build GRCh38
rs753398150 Chr.8: 56445752-56445752 on Build GRCh38
rs983043928 Chr.8: 56445704-56445704 on Build GRCh38
rs765471636 Chr.8: 56445679-56445679 on Build GRCh38
rs950887957 Chr.8: 56445762-56445762 on Build GRCh38
rs747629089 Chr.8: 56445800-56445800 on Build GRCh38
rs778453235 Chr.8: 56445810-56445810 on Build GRCh38
rs535781218 Chr.8: 56445702-56445702 on Build GRCh38
rs747717615 Chr.8: 56445695-56445695 on Build GRCh38
rs1034934396 Chr.8: 56445699-56445699 on Build GRCh38
rs750089927 Chr.8: 56445819-56445819 on Build GRCh38
rs773284471 Chr.8: 56445777-56445777 on Build GRCh38
rs759775174 Chr.8: 56445783-56445783 on Build GRCh38
rs111909825 Chr.8: 56445683-56445683 on Build GRCh38
rs766787131 Chr.8: 56445687-56445687 on Build GRCh38
rs772065558 Chr.8: 56445708-56445708 on Build GRCh38
rs900460701 Chr.8: 56445666-56445666 on Build GRCh38
rs750451496 Chr.8: 56445786-56445787 on Build GRCh38
rs768102955 Chr.8: 56445732-56445732 on Build GRCh38
rs1010630064 Chr.8: 56445836-56445836 on Build GRCh38
rs577920703 Chr.8: 56445627-56445627 on Build GRCh38
rs1023218194 Chr.8: 56445837-56445837 on Build GRCh38
rs201829916 Chr.8: 56445797-56445797 on Build GRCh38
rs903474744 Chr.8: 56445794-56445794 on Build GRCh38
rs777806641 Chr.8: 56445773-56445774 on Build GRCh38
rs756784111 Chr.8: 56445806-56445806 on Build GRCh38
rs568142774 Chr.8: 56445722-56445722 on Build GRCh38
rs753186700 Chr.8: 56445739-56445739 on Build GRCh38
rs775249263 Chr.8: 56445713-56445713 on Build GRCh38
rs771165037 Chr.8: 56445781-56445781 on Build GRCh38
rs570539667 Chr.8: 56445647-56445647 on Build GRCh38
rs763228442 Chr.8: 56445725-56445725 on Build GRCh38

View Full Product Details