Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 66161936-66162181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SREK1
Entrez Gene ID
140890
Gene Name
splicing regulatory glutamic acid and lysine rich protein 1
Gene Aliases
SFRS12, SRrp508, SRrp86
UniGene
Hs.49367
Gene Chromosome Location
Chr.5: 66144218-66183616 on Build GRCh38
Transcripts
NM_001077199.2 NM_001270492.1 NM_001323527.1 NM_001323529.1 NM_001323533.1 NM_001323534.1 NM_001323535.1 NM_139168.3 XM_011543171.2 XM_017009055.1 XM_017009056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4006039 c.462C>T p.D154D Chr.5: 66162159-66162159 on Build GRCh38
5868703 c.435delG p.G146fs*8 Chr.5: 66162132-66162132 on Build GRCh38
5779056 c.450A>G p.A150A Chr.5: 66162147-66162147 on Build GRCh38
5868704 c.87delG p.G30fs*8 Chr.5: 66162132-66162132 on Build GRCh38
6875016 c.431G>A p.S144N Chr.5: 66162128-66162128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775668903 Chr.5: 66162139-66162139 on Build GRCh38
rs539191353 Chr.5: 66161990-66161990 on Build GRCh38
rs992149601 Chr.5: 66161986-66161986 on Build GRCh38
rs749500806 Chr.5: 66162088-66162088 on Build GRCh38
rs756407812 Chr.5: 66162081-66162081 on Build GRCh38
rs750614156 Chr.5: 66162077-66162077 on Build GRCh38
rs778809596 Chr.5: 66162083-66162083 on Build GRCh38
rs917927158 Chr.5: 66161987-66161987 on Build GRCh38
rs763355323 Chr.5: 66162147-66162147 on Build GRCh38
rs756425057 Chr.5: 66162067-66162067 on Build GRCh38
rs767140510 Chr.5: 66162089-66162089 on Build GRCh38
rs879045873 Chr.5: 66162044-66162044 on Build GRCh38
rs569530351 Chr.5: 66162103-66162103 on Build GRCh38
rs1038892602 Chr.5: 66162098-66162098 on Build GRCh38
rs778109713 Chr.5: 66162087-66162088 on Build GRCh38
rs745459857 Chr.5: 66162118-66162118 on Build GRCh38
rs757331534 Chr.5: 66162064-66162064 on Build GRCh38
rs555774034 Chr.5: 66162058-66162058 on Build GRCh38
rs752056060 Chr.5: 66162060-66162060 on Build GRCh38
rs374216246 Chr.5: 66162104-66162104 on Build GRCh38
rs945427454 Chr.5: 66161994-66161994 on Build GRCh38
rs536190085 Chr.5: 66161975-66161975 on Build GRCh38
rs777732706 Chr.5: 66162097-66162097 on Build GRCh38
rs767774799 Chr.5: 66162066-66162066 on Build GRCh38
rs565281010 Chr.5: 66162160-66162160 on Build GRCh38
rs747994056 Chr.5: 66162095-66162095 on Build GRCh38
rs776135795 Chr.5: 66162105-66162105 on Build GRCh38
rs762066920 Chr.5: 66162050-66162050 on Build GRCh38
rs758774322 Chr.5: 66162001-66162001 on Build GRCh38
rs769471962 Chr.5: 66162127-66162127 on Build GRCh38
rs936480643 Chr.5: 66162088-66162088 on Build GRCh38
rs758171915 Chr.5: 66162096-66162096 on Build GRCh38
rs1042479373 Chr.5: 66162075-66162075 on Build GRCh38

View Full Product Details