Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26093012-26093259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HFE
Entrez Gene ID
3077
Gene Name
hemochromatosis
Gene Aliases
HFE1, HH, HLA-H, MVCD7, TFQTL2
UniGene
Hs.233325
Gene Chromosome Location
Chr.6: 26087281-26096216 on Build GRCh38
Transcripts
NM_000410.3 NM_001300749.1 NM_139003.2 NM_139004.2 NM_139006.2 NM_139007.2 NM_139008.2 NM_139009.2 NM_139010.2 NM_139011.2 XM_011514543.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4403861 c.913G>A p.G305R Chr.6: 26093148-26093148 on Build GRCh38
5404954 c.991G>A p.G331S Chr.6: 26093226-26093226 on Build GRCh38
7317175 c.930C>T p.I310I Chr.6: 26093156-26093156 on Build GRCh38
7371057 c.934G>A p.G312R Chr.6: 26093160-26093160 on Build GRCh38
5500615 c.948C>G p.I316M Chr.6: 26093183-26093183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571150642 Chr.6: 26093139-26093139 on Build GRCh38
rs879360468 Chr.6: 26093078-26093078 on Build GRCh38
rs773972161 Chr.6: 26093166-26093166 on Build GRCh38
rs2794717 Chr.6: 26093069-26093069 on Build GRCh38
rs573745685 Chr.6: 26093233-26093233 on Build GRCh38
rs1005321343 Chr.6: 26093066-26093066 on Build GRCh38
rs537002912 Chr.6: 26093145-26093145 on Build GRCh38
rs897206619 Chr.6: 26093128-26093128 on Build GRCh38
rs781632455 Chr.6: 26093149-26093149 on Build GRCh38
rs138993448 Chr.6: 26093206-26093206 on Build GRCh38
rs761841001 Chr.6: 26093071-26093071 on Build GRCh38
rs1003455301 Chr.6: 26093076-26093076 on Build GRCh38
rs915098169 Chr.6: 26093138-26093138 on Build GRCh38
rs755751149 Chr.6: 26093112-26093112 on Build GRCh38
rs748882267 Chr.6: 26093156-26093156 on Build GRCh38
rs141229562 Chr.6: 26093178-26093178 on Build GRCh38
rs768426111 Chr.6: 26093160-26093160 on Build GRCh38
rs371192232 Chr.6: 26093177-26093177 on Build GRCh38
rs771135070 Chr.6: 26093146-26093146 on Build GRCh38
rs978229077 Chr.6: 26093169-26093169 on Build GRCh38
rs766108842 Chr.6: 26093193-26093193 on Build GRCh38
rs755284374 Chr.6: 26093122-26093122 on Build GRCh38
rs111033558 Chr.6: 26093215-26093215 on Build GRCh38
rs561001400 Chr.6: 26093056-26093056 on Build GRCh38
rs766197621 Chr.6: 26093110-26093110 on Build GRCh38
rs750403696 Chr.6: 26093080-26093080 on Build GRCh38
rs753732462 Chr.6: 26093194-26093194 on Build GRCh38
rs748266596 Chr.6: 26093125-26093125 on Build GRCh38
rs150716212 Chr.6: 26093191-26093191 on Build GRCh38
rs114038675 Chr.6: 26093120-26093120 on Build GRCh38
rs147519426 Chr.6: 26093170-26093170 on Build GRCh38
rs372856303 Chr.6: 26093129-26093129 on Build GRCh38
rs1800708 Chr.6: 26093075-26093075 on Build GRCh38
rs777836090 Chr.6: 26093235-26093235 on Build GRCh38
rs765545512 Chr.6: 26093226-26093226 on Build GRCh38
rs148632352 Chr.6: 26093171-26093171 on Build GRCh38
rs62625347 Chr.6: 26093091-26093091 on Build GRCh38

View Full Product Details