Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46040801-46041074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFL4
Entrez Gene ID
444882
Gene Name
IGF like family member 4
Gene Aliases
-
UniGene
Hs.531310
Gene Chromosome Location
Chr.19: 46039748-46041016 on Build GRCh38
Transcripts
NM_001002923.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998377 c.17C>A p.S6Y Chr.19: 46040946-46040946 on Build GRCh38
1712514 c.11G>A p.R4K Chr.19: 46040952-46040952 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs138465035 Chr.19: 46041049-46041050 on Build GRCh38
rs756804948 Chr.19: 46040982-46040982 on Build GRCh38
rs569811182 Chr.19: 46041003-46041003 on Build GRCh38
rs10518266 Chr.19: 46041053-46041053 on Build GRCh38
rs891231805 Chr.19: 46041039-46041039 on Build GRCh38
rs879134168 Chr.19: 46040987-46040987 on Build GRCh38
rs769177050 Chr.19: 46040994-46040994 on Build GRCh38
rs990456933 Chr.19: 46040866-46040866 on Build GRCh38
rs1031869431 Chr.19: 46040859-46040859 on Build GRCh38
rs932705898 Chr.19: 46041023-46041023 on Build GRCh38
rs915678842 Chr.19: 46040906-46040906 on Build GRCh38
rs991220948 Chr.19: 46040893-46040893 on Build GRCh38
rs769058483 Chr.19: 46041051-46041051 on Build GRCh38
rs201920109 Chr.19: 46040963-46040963 on Build GRCh38
rs956471282 Chr.19: 46040863-46040863 on Build GRCh38
rs56734695 Chr.19: 46040990-46040990 on Build GRCh38
rs763838348 Chr.19: 46040967-46040967 on Build GRCh38
rs371024083 Chr.19: 46041024-46041024 on Build GRCh38
rs749660572 Chr.19: 46040999-46040999 on Build GRCh38
rs200186851 Chr.19: 46040959-46040959 on Build GRCh38
rs977653886 Chr.19: 46040843-46040843 on Build GRCh38
rs752537696 Chr.19: 46040962-46040962 on Build GRCh38
rs773107657 Chr.19: 46040933-46040933 on Build GRCh38
rs925840559 Chr.19: 46040852-46040852 on Build GRCh38
rs753513190 Chr.19: 46040975-46040975 on Build GRCh38
rs186806261 Chr.19: 46041052-46041052 on Build GRCh38
rs745369787 Chr.19: 46040983-46040983 on Build GRCh38
rs543226561 Chr.19: 46040828-46040828 on Build GRCh38
rs957334761 Chr.19: 46040858-46040859 on Build GRCh38
rs868023408 Chr.19: 46041048-46041049 on Build GRCh38
rs145399842 Chr.19: 46040986-46040986 on Build GRCh38

View Full Product Details