Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109229263-109229492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARS
Entrez Gene ID
6301
Gene Name
seryl-tRNA synthetase
Gene Aliases
SERRS, SERS
UniGene
Hs.531176
Gene Chromosome Location
Chr.1: 109213893-109238182 on Build GRCh38
Transcripts
NM_006513.3 XM_006710813.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1600490 c.320G>T p.R107L Chr.1: 109229445-109229445 on Build GRCh38
1600491 c.356C>T p.A119V Chr.1: 109229481-109229481 on Build GRCh38
4895804 c.318C>T p.V106V Chr.1: 109229443-109229443 on Build GRCh38
6771935 c.320G>A p.R107Q Chr.1: 109229445-109229445 on Build GRCh38
4397155 c.330T>C p.I110I Chr.1: 109229455-109229455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933424376 Chr.1: 109229380-109229380 on Build GRCh38
rs780615205 Chr.1: 109229467-109229467 on Build GRCh38
rs955363429 Chr.1: 109229347-109229347 on Build GRCh38
rs139772934 Chr.1: 109229420-109229420 on Build GRCh38
rs775949136 Chr.1: 109229419-109229419 on Build GRCh38
rs139902476 Chr.1: 109229431-109229431 on Build GRCh38
rs986210226 Chr.1: 109229411-109229411 on Build GRCh38
rs375548086 Chr.1: 109229444-109229444 on Build GRCh38
rs368687246 Chr.1: 109229399-109229399 on Build GRCh38
rs752386701 Chr.1: 109229438-109229438 on Build GRCh38
rs1006708221 Chr.1: 109229405-109229405 on Build GRCh38
rs891584392 Chr.1: 109229300-109229300 on Build GRCh38
rs773102211 Chr.1: 109229369-109229369 on Build GRCh38
rs760491818 Chr.1: 109229390-109229390 on Build GRCh38
rs767953116 Chr.1: 109229445-109229445 on Build GRCh38
rs1010359074 Chr.1: 109229322-109229322 on Build GRCh38
rs200952306 Chr.1: 109229407-109229407 on Build GRCh38
rs756521030 Chr.1: 109229466-109229466 on Build GRCh38
rs750819700 Chr.1: 109229446-109229446 on Build GRCh38
rs555428768 Chr.1: 109229341-109229341 on Build GRCh38

View Full Product Details