Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15109868-15110099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYDE1
Entrez Gene ID
85360
Gene Name
synapse defective Rho GTPase homolog 1
Gene Aliases
7h3, SYD1
UniGene
Hs.528701
Gene Chromosome Location
Chr.19: 15107364-15114988 on Build GRCh38
Transcripts
NM_001300910.1 NM_033025.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6202114 c.597C>A p.S199S Chr.19: 15109870-15109870 on Build GRCh38
4386900 c.639C>A p.P213P Chr.19: 15109912-15109912 on Build GRCh38
5504495 c.678G>A p.R226R Chr.19: 15109951-15109951 on Build GRCh38
296526 c.708G>A p.P236P Chr.19: 15109981-15109981 on Build GRCh38
6404461 c.662G>T p.G221V Chr.19: 15109935-15109935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908770755 Chr.19: 15110008-15110008 on Build GRCh38
rs940270518 Chr.19: 15110026-15110026 on Build GRCh38
rs776618393 Chr.19: 15109980-15109980 on Build GRCh38
rs749505844 Chr.19: 15110045-15110045 on Build GRCh38
rs907187642 Chr.19: 15110053-15110053 on Build GRCh38
rs1050731012 Chr.19: 15109996-15109996 on Build GRCh38
rs2240297 Chr.19: 15110039-15110039 on Build GRCh38
rs1002843131 Chr.19: 15110056-15110056 on Build GRCh38
rs1051026364 Chr.19: 15109941-15109941 on Build GRCh38
rs1054584284 Chr.19: 15109971-15109971 on Build GRCh38
rs933969537 Chr.19: 15109910-15109915 on Build GRCh38
rs920926461 Chr.19: 15109991-15109991 on Build GRCh38
rs1013002734 Chr.19: 15110046-15110046 on Build GRCh38
rs971920408 Chr.19: 15109901-15109901 on Build GRCh38
rs946452543 Chr.19: 15109970-15109970 on Build GRCh38
rs975166842 Chr.19: 15109978-15109978 on Build GRCh38
rs780948091 Chr.19: 15109981-15109981 on Build GRCh38
rs933687965 Chr.19: 15109995-15109995 on Build GRCh38
rs757237808 Chr.19: 15109917-15109917 on Build GRCh38
rs745996614 Chr.19: 15110034-15110034 on Build GRCh38
rs940575637 Chr.19: 15109889-15109889 on Build GRCh38
rs554738306 Chr.19: 15110018-15110018 on Build GRCh38
rs889821150 Chr.19: 15109942-15109942 on Build GRCh38
rs536102093 Chr.19: 15109913-15109913 on Build GRCh38
rs915061115 Chr.19: 15109953-15109953 on Build GRCh38
rs778610532 Chr.19: 15110051-15110051 on Build GRCh38
rs956846628 Chr.19: 15109909-15109909 on Build GRCh38
rs923203532 Chr.19: 15109906-15109906 on Build GRCh38
rs751253610 Chr.19: 15109911-15109911 on Build GRCh38
rs780311492 Chr.19: 15110043-15110043 on Build GRCh38
rs1044525425 Chr.19: 15110052-15110052 on Build GRCh38
rs1003150564 Chr.19: 15110071-15110071 on Build GRCh38

View Full Product Details