Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15727420-15727645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHM2
Entrez Gene ID
23207
Gene Name
pleckstrin homology and RUN domain containing M2
Gene Aliases
SKIP
UniGene
Hs.646775
Gene Chromosome Location
Chr.1: 15681506-15734769 on Build GRCh38
Transcripts
NM_015164.2 XM_005245790.3 XM_005245791.4 XM_017000757.1 XM_017000758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1295347 c.1685C>T p.S562L Chr.1: 15727448-15727448 on Build GRCh38
6749997 c.1686A>G p.S562S Chr.1: 15727449-15727449 on Build GRCh38
3976388 c.1722T>C p.S574S Chr.1: 15727485-15727485 on Build GRCh38
4962517 c.1840G>A p.G614R Chr.1: 15727603-15727603 on Build GRCh38
5819217 c.1539G>A p.T513T Chr.1: 15727611-15727611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768469775 Chr.1: 15727611-15727611 on Build GRCh38
rs202081024 Chr.1: 15727490-15727490 on Build GRCh38
rs775326636 Chr.1: 15727468-15727468 on Build GRCh38
rs748904369 Chr.1: 15727477-15727477 on Build GRCh38
rs774072603 Chr.1: 15727489-15727489 on Build GRCh38
rs868129375 Chr.1: 15727528-15727528 on Build GRCh38
rs368020288 Chr.1: 15727541-15727541 on Build GRCh38
rs983530335 Chr.1: 15727509-15727509 on Build GRCh38
rs745835546 Chr.1: 15727453-15727453 on Build GRCh38
rs371661618 Chr.1: 15727542-15727542 on Build GRCh38
rs200088107 Chr.1: 15727491-15727491 on Build GRCh38
rs745792439 Chr.1: 15727572-15727572 on Build GRCh38
rs867087978 Chr.1: 15727591-15727591 on Build GRCh38
rs760098746 Chr.1: 15727626-15727626 on Build GRCh38
rs370067445 Chr.1: 15727586-15727586 on Build GRCh38
rs772819095 Chr.1: 15727621-15727621 on Build GRCh38
rs780927725 Chr.1: 15727441-15727441 on Build GRCh38
rs375612582 Chr.1: 15727456-15727456 on Build GRCh38
rs997954845 Chr.1: 15727481-15727481 on Build GRCh38
rs761868279 Chr.1: 15727588-15727590 on Build GRCh38
rs770377384 Chr.1: 15727627-15727627 on Build GRCh38
rs768343269 Chr.1: 15727479-15727479 on Build GRCh38
rs373517149 Chr.1: 15727610-15727610 on Build GRCh38
rs747848774 Chr.1: 15727615-15727615 on Build GRCh38
rs753161597 Chr.1: 15727529-15727529 on Build GRCh38
rs760178350 Chr.1: 15727518-15727518 on Build GRCh38
rs202014397 Chr.1: 15727567-15727567 on Build GRCh38
rs771469934 Chr.1: 15727619-15727619 on Build GRCh38
rs757229933 Chr.1: 15727557-15727557 on Build GRCh38
rs765666083 Chr.1: 15727519-15727519 on Build GRCh38
rs376758113 Chr.1: 15727510-15727510 on Build GRCh38
rs779799765 Chr.1: 15727587-15727587 on Build GRCh38
rs914807369 Chr.1: 15727449-15727449 on Build GRCh38
rs964951697 Chr.1: 15727470-15727470 on Build GRCh38

View Full Product Details