Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93209843-93210068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBR7
Entrez Gene ID
55148
Gene Name
ubiquitin protein ligase E3 component n-recognin 7 (putative)
Gene Aliases
C14orf130
UniGene
Hs.648806
Gene Chromosome Location
Chr.14: 93207056-93229215 on Build GRCh38
Transcripts
NM_175748.3 NR_038150.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1707741 c.230C>T p.A77V Chr.14: 93209903-93209903 on Build GRCh38
1231637 c.268G>T p.E90* Chr.14: 93209941-93209941 on Build GRCh38
3420105 c.244T>G p.C82G Chr.14: 93209917-93209917 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769204867 Chr.14: 93209978-93209978 on Build GRCh38
rs748527767 Chr.14: 93209983-93209983 on Build GRCh38
rs779550201 Chr.14: 93209968-93209968 on Build GRCh38
rs773758981 Chr.14: 93210021-93210021 on Build GRCh38
rs976268003 Chr.14: 93209906-93209906 on Build GRCh38
rs138426899 Chr.14: 93209922-93209922 on Build GRCh38
rs367684277 Chr.14: 93210025-93210025 on Build GRCh38
rs773528929 Chr.14: 93209996-93209996 on Build GRCh38
rs745548290 Chr.14: 93209949-93209949 on Build GRCh38
rs769222820 Chr.14: 93209952-93209952 on Build GRCh38
rs537043207 Chr.14: 93210039-93210039 on Build GRCh38
rs748804600 Chr.14: 93209976-93209976 on Build GRCh38
rs766229154 Chr.14: 93210018-93210018 on Build GRCh38
rs780773371 Chr.14: 93209938-93209938 on Build GRCh38
rs774763059 Chr.14: 93209981-93209981 on Build GRCh38
rs776490824 Chr.14: 93210040-93210040 on Build GRCh38
rs183524346 Chr.14: 93209895-93209895 on Build GRCh38
rs772591613 Chr.14: 93209986-93209986 on Build GRCh38
rs1041938279 Chr.14: 93209939-93209939 on Build GRCh38
rs903215144 Chr.14: 93209963-93209963 on Build GRCh38
rs371594824 Chr.14: 93209937-93209937 on Build GRCh38
rs777636482 Chr.14: 93209909-93209909 on Build GRCh38
rs760982436 Chr.14: 93209997-93209997 on Build GRCh38
rs143806979 Chr.14: 93209869-93209869 on Build GRCh38

View Full Product Details