Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 79087444-79087717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REG1B
Entrez Gene ID
5968
Gene Name
regenerating family member 1 beta
Gene Aliases
PSPS2, REGH, REGI-BETA, REGL
UniGene
Hs.4158
Gene Chromosome Location
Chr.2: 79085023-79088024 on Build GRCh38
Transcripts
NM_006507.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
125813 c.4G>T p.A2S Chr.2: 79087609-79087609 on Build GRCh38
3939184 c.12C>A p.T4T Chr.2: 79087601-79087601 on Build GRCh38
4720982 c.53C>A p.S18Y Chr.2: 79087560-79087560 on Build GRCh38
6092221 c.48C>G p.F16L Chr.2: 79087565-79087565 on Build GRCh38
3000577 c.24C>T p.F8F Chr.2: 79087589-79087589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748914368 Chr.2: 79087595-79087595 on Build GRCh38
rs772255795 Chr.2: 79087600-79087600 on Build GRCh38
rs775842584 Chr.2: 79087644-79087644 on Build GRCh38
rs749277780 Chr.2: 79087596-79087596 on Build GRCh38
rs753317922 Chr.2: 79087560-79087560 on Build GRCh38
rs757876537 Chr.2: 79087562-79087562 on Build GRCh38
rs1000609081 Chr.2: 79087495-79087495 on Build GRCh38
rs775823333 Chr.2: 79087599-79087600 on Build GRCh38
rs1032118276 Chr.2: 79087532-79087532 on Build GRCh38
rs200253381 Chr.2: 79087652-79087652 on Build GRCh38
rs773375427 Chr.2: 79087607-79087607 on Build GRCh38
rs780903529 Chr.2: 79087584-79087584 on Build GRCh38
rs756864866 Chr.2: 79087575-79087575 on Build GRCh38
rs957420926 Chr.2: 79087656-79087656 on Build GRCh38
rs75903719 Chr.2: 79087595-79087595 on Build GRCh38
rs1012338962 Chr.2: 79087536-79087536 on Build GRCh38
rs139308868 Chr.2: 79087586-79087586 on Build GRCh38
rs751095254 Chr.2: 79087572-79087572 on Build GRCh38
rs868243041 Chr.2: 79087589-79087589 on Build GRCh38
rs773535153 Chr.2: 79087508-79087508 on Build GRCh38
rs771207622 Chr.2: 79087612-79087612 on Build GRCh38
rs777317300 Chr.2: 79087565-79087565 on Build GRCh38
rs777453851 Chr.2: 79087499-79087499 on Build GRCh38
rs531427797 Chr.2: 79087676-79087676 on Build GRCh38
rs747114745 Chr.2: 79087609-79087609 on Build GRCh38
rs780648821 Chr.2: 79087653-79087653 on Build GRCh38
rs772480089 Chr.2: 79087598-79087598 on Build GRCh38
rs772343946 Chr.2: 79087501-79087501 on Build GRCh38
rs766727593 Chr.2: 79087520-79087520 on Build GRCh38
rs11548891 Chr.2: 79087635-79087635 on Build GRCh38
rs201376629 Chr.2: 79087620-79087620 on Build GRCh38
rs373401987 Chr.2: 79087519-79087519 on Build GRCh38
rs894790702 Chr.2: 79087535-79087535 on Build GRCh38
rs557526871 Chr.2: 79087481-79087481 on Build GRCh38
rs765601234 Chr.2: 79087541-79087541 on Build GRCh38
rs1013518297 Chr.2: 79087546-79087546 on Build GRCh38
rs202197145 Chr.2: 79087619-79087619 on Build GRCh38
rs765678743 Chr.2: 79087642-79087642 on Build GRCh38
rs1025271533 Chr.2: 79087564-79087564 on Build GRCh38
rs79463000 Chr.2: 79087538-79087538 on Build GRCh38
rs931970073 Chr.2: 79087655-79087655 on Build GRCh38
rs367561478 Chr.2: 79087527-79087527 on Build GRCh38
rs562351053 Chr.2: 79087674-79087674 on Build GRCh38

View Full Product Details