Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58349-58584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHBDF1
Entrez Gene ID
64285
Gene Name
rhomboid 5 homolog 1
Gene Aliases
C16orf8, Dist1, EGFR-RS, gene-89, gene-90, hDist1
UniGene
Hs.744992
Gene Chromosome Location
Chr.16: 58059-76355 on Build GRCh38
Transcripts
NM_022450.3 XM_005255494.1 XM_005255498.2 XM_006720921.1 XM_017023556.1 XM_017023557.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4721517 c.2338C>A p.L780I Chr.16: 58570-58570 on Build GRCh38
4058142 c.2469C>T p.Y823Y Chr.16: 58439-58439 on Build GRCh38
7191393 c.2408_2419del12 p.Q803_F807>L Chr.16: 58489-58500 on Build GRCh38
6366760 c.2559G>T p.Q853H Chr.16: 58349-58349 on Build GRCh38
4058141 c.2470G>A p.V824I Chr.16: 58438-58438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371297598 Chr.16: 58394-58394 on Build GRCh38
rs1028309211 Chr.16: 58426-58426 on Build GRCh38
rs188819873 Chr.16: 58506-58506 on Build GRCh38
rs769508190 Chr.16: 58514-58514 on Build GRCh38
rs201721796 Chr.16: 58555-58555 on Build GRCh38
rs751122410 Chr.16: 58554-58554 on Build GRCh38
rs762560464 Chr.16: 58434-58434 on Build GRCh38
rs750283540 Chr.16: 58446-58446 on Build GRCh38
rs748168212 Chr.16: 58478-58478 on Build GRCh38
rs761300254 Chr.16: 58387-58387 on Build GRCh38
rs773722303 Chr.16: 58385-58385 on Build GRCh38
rs759973395 Chr.16: 58513-58513 on Build GRCh38
rs772500595 Chr.16: 58382-58382 on Build GRCh38
rs765697682 Chr.16: 58490-58492 on Build GRCh38
rs763695736 Chr.16: 58538-58538 on Build GRCh38
rs199806322 Chr.16: 58439-58439 on Build GRCh38
rs141174547 Chr.16: 58523-58523 on Build GRCh38
rs771194144 Chr.16: 58390-58390 on Build GRCh38
rs752211014 Chr.16: 58425-58425 on Build GRCh38
rs57236135 Chr.16: 58409-58409 on Build GRCh38
rs972917611 Chr.16: 58483-58483 on Build GRCh38
rs369772748 Chr.16: 58438-58438 on Build GRCh38
rs891554641 Chr.16: 58384-58384 on Build GRCh38
rs776808259 Chr.16: 58512-58512 on Build GRCh38
rs767513643 Chr.16: 58556-58556 on Build GRCh38
rs772125685 Chr.16: 58490-58490 on Build GRCh38
rs368957950 Chr.16: 58463-58463 on Build GRCh38
rs778216721 Chr.16: 58391-58391 on Build GRCh38
rs199643977 Chr.16: 58448-58448 on Build GRCh38
rs762190927 Chr.16: 58425-58426 on Build GRCh38
rs1021913112 Chr.16: 58401-58401 on Build GRCh38
rs555521446 Chr.16: 58517-58517 on Build GRCh38
rs771515428 Chr.16: 58507-58507 on Build GRCh38
rs146954411 Chr.16: 58526-58526 on Build GRCh38

View Full Product Details