Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58547755-58548002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM28
Entrez Gene ID
10155
Gene Name
tripartite motif containing 28
Gene Aliases
KAP1, PPP1R157, RNF96, TF1B, TIF1B
UniGene
Hs.467408
Gene Chromosome Location
Chr.19: 58544469-58550715 on Build GRCh38
Transcripts
NM_005762.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1600242 c.865A>C p.K289Q Chr.19: 58547817-58547817 on Build GRCh38
6086213 c.859G>A p.V287I Chr.19: 58547811-58547811 on Build GRCh38
4082475 c.863A>G p.Q288R Chr.19: 58547815-58547815 on Build GRCh38
1002779 c.899T>A p.L300Q Chr.19: 58547851-58547851 on Build GRCh38
2974767 c.926G>A p.R309Q Chr.19: 58547878-58547878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773846415 Chr.19: 58547852-58547852 on Build GRCh38
rs779984035 Chr.19: 58547840-58547840 on Build GRCh38
rs758388758 Chr.19: 58547811-58547811 on Build GRCh38
rs755382796 Chr.19: 58547804-58547804 on Build GRCh38
rs930598978 Chr.19: 58547966-58547966 on Build GRCh38
rs777244956 Chr.19: 58547949-58547949 on Build GRCh38
rs759777699 Chr.19: 58547896-58547896 on Build GRCh38
rs201759225 Chr.19: 58547810-58547810 on Build GRCh38
rs781129647 Chr.19: 58547953-58547953 on Build GRCh38
rs200549746 Chr.19: 58547789-58547789 on Build GRCh38
rs772921165 Chr.19: 58547888-58547888 on Build GRCh38
rs757352851 Chr.19: 58547921-58547921 on Build GRCh38
rs749037250 Chr.19: 58547849-58547849 on Build GRCh38
rs769564940 Chr.19: 58547775-58547775 on Build GRCh38
rs765684894 Chr.19: 58547900-58547900 on Build GRCh38
rs754704248 Chr.19: 58547929-58547929 on Build GRCh38
rs542460622 Chr.19: 58547935-58547935 on Build GRCh38
rs770594213 Chr.19: 58547947-58547947 on Build GRCh38
rs753280654 Chr.19: 58547958-58547959 on Build GRCh38
rs750940640 Chr.19: 58547788-58547788 on Build GRCh38
rs768138544 Chr.19: 58547781-58547781 on Build GRCh38
rs773011304 Chr.19: 58547941-58547941 on Build GRCh38
rs745802032 Chr.19: 58547826-58547826 on Build GRCh38
rs771466139 Chr.19: 58547873-58547873 on Build GRCh38
rs758714859 Chr.19: 58547911-58547911 on Build GRCh38
rs775858258 Chr.19: 58547958-58547958 on Build GRCh38
rs747747399 Chr.19: 58547934-58547934 on Build GRCh38
rs370690334 Chr.19: 58547821-58547821 on Build GRCh38
rs922097204 Chr.19: 58547915-58547915 on Build GRCh38
rs769427388 Chr.19: 58547956-58547956 on Build GRCh38
rs766841079 Chr.19: 58547798-58547798 on Build GRCh38
rs149921098 Chr.19: 58547964-58547964 on Build GRCh38
rs113868504 Chr.19: 58547809-58547809 on Build GRCh38
rs764304860 Chr.19: 58547916-58547916 on Build GRCh38
rs75453307 Chr.19: 58547786-58547786 on Build GRCh38
rs778823281 Chr.19: 58547933-58547933 on Build GRCh38
rs766934679 Chr.19: 58547894-58547894 on Build GRCh38
rs761319341 Chr.19: 58547795-58547795 on Build GRCh38
rs774078560 Chr.19: 58547783-58547783 on Build GRCh38
rs968993665 Chr.19: 58547884-58547884 on Build GRCh38
rs749290543 Chr.19: 58547925-58547925 on Build GRCh38
rs752851138 Chr.19: 58547909-58547909 on Build GRCh38
rs747759690 Chr.19: 58547954-58547954 on Build GRCh38
rs1000718216 Chr.19: 58547819-58547819 on Build GRCh38
rs543150564 Chr.19: 58547801-58547801 on Build GRCh38
rs755979922 Chr.19: 58547837-58547837 on Build GRCh38
rs760262357 Chr.19: 58547891-58547891 on Build GRCh38
rs576523191 Chr.19: 58547850-58547850 on Build GRCh38
rs747881278 Chr.19: 58547870-58547870 on Build GRCh38
rs771685104 Chr.19: 58547940-58547940 on Build GRCh38
rs751584592 Chr.19: 58547920-58547920 on Build GRCh38
rs761190458 Chr.19: 58547785-58547785 on Build GRCh38
rs200132486 Chr.19: 58547944-58547944 on Build GRCh38

View Full Product Details