Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14787384-14787655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf60
Entrez Gene ID
144608
Gene Name
chromosome 12 open reading frame 60
Gene Aliases
-
UniGene
Hs.659951
Gene Chromosome Location
Chr.12: 14567991-14823858 on Build GRCh38
Transcripts
XM_017018871.1 XM_017018872.1 NM_016312.2

Gene Symbol
WBP11
Entrez Gene ID
51729
Gene Name
WW domain binding protein 11
Gene Aliases
NPWBP, PPP1R165, SIPP1, WBP-11
UniGene
Hs.655138
Gene Chromosome Location
Chr.12: 14786478-14803467 on Build GRCh38
Transcripts
XM_017018871.1 XM_017018872.1 NM_016312.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3769802 c.1596G>A p.G532G Chr.12: 14787395-14787395 on Build GRCh38
7327614 c.1591C>T p.P531S Chr.12: 14787400-14787400 on Build GRCh38
6519997 c.1501C>T p.P501S Chr.12: 14787490-14787490 on Build GRCh38
300131 c.1559_1560insC p.G521fs*28 Chr.12: 14787431-14787432 on Build GRCh38
3458615 c.1540C>T p.P514S Chr.12: 14787451-14787451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021570654 Chr.12: 14787490-14787490 on Build GRCh38
rs907730920 Chr.12: 14787622-14787622 on Build GRCh38
rs766690158 Chr.12: 14787412-14787412 on Build GRCh38
rs755159256 Chr.12: 14787433-14787433 on Build GRCh38
rs980569933 Chr.12: 14787597-14787597 on Build GRCh38
rs755889357 Chr.12: 14787530-14787531 on Build GRCh38
rs753797960 Chr.12: 14787550-14787552 on Build GRCh38
rs1019814994 Chr.12: 14787437-14787437 on Build GRCh38
rs892045000 Chr.12: 14787439-14787439 on Build GRCh38
rs914689620 Chr.12: 14787554-14787554 on Build GRCh38
rs3748291 Chr.12: 14787611-14787611 on Build GRCh38
rs775832194 Chr.12: 14787535-14787538 on Build GRCh38
rs768163101 Chr.12: 14787525-14787525 on Build GRCh38
rs760584916 Chr.12: 14787543-14787543 on Build GRCh38
rs1034812486 Chr.12: 14787583-14787583 on Build GRCh38
rs772047953 Chr.12: 14787536-14787536 on Build GRCh38
rs779515528 Chr.12: 14787443-14787443 on Build GRCh38
rs772852707 Chr.12: 14787541-14787541 on Build GRCh38
rs759174029 Chr.12: 14787431-14787431 on Build GRCh38
rs776924420 Chr.12: 14787548-14787548 on Build GRCh38
rs753290080 Chr.12: 14787466-14787466 on Build GRCh38
rs113227238 Chr.12: 14787556-14787556 on Build GRCh38
rs767139774 Chr.12: 14787431-14787432 on Build GRCh38
rs968924712 Chr.12: 14787570-14787570 on Build GRCh38
rs766497246 Chr.12: 14787544-14787544 on Build GRCh38
rs373106696 Chr.12: 14787571-14787572 on Build GRCh38
rs747468762 Chr.12: 14787497-14787497 on Build GRCh38
rs367636048 Chr.12: 14787529-14787529 on Build GRCh38
rs778298649 Chr.12: 14787475-14787475 on Build GRCh38
rs748692229 Chr.12: 14787516-14787516 on Build GRCh38
rs764353363 Chr.12: 14787611-14787611 on Build GRCh38
rs753830511 Chr.12: 14787425-14787425 on Build GRCh38
rs752438572 Chr.12: 14787518-14787520 on Build GRCh38
rs748107339 Chr.12: 14787530-14787530 on Build GRCh38
rs758771708 Chr.12: 14787469-14787469 on Build GRCh38
rs769331874 Chr.12: 14787513-14787513 on Build GRCh38
rs779800983 Chr.12: 14787515-14787515 on Build GRCh38
rs745834529 Chr.12: 14787451-14787451 on Build GRCh38

View Full Product Details