Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 149933339-149933612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR11
Entrez Gene ID
10903
Gene Name
myotubularin related protein 11
Gene Aliases
CRA
UniGene
Hs.425144
Gene Chromosome Location
Chr.1: 149928651-149936898 on Build GRCh38
Transcripts
NM_001145862.1 NM_181873.3 XM_006711137.1 XM_011509099.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3862624 c.946C>T p.Q316* Chr.1: 149933445-149933445 on Build GRCh38
4021908 c.754G>A p.A252T Chr.1: 149933421-149933421 on Build GRCh38
5598505 c.934C>A p.L312I Chr.1: 149933457-149933457 on Build GRCh38
6120984 c.672G>C p.G224G Chr.1: 149933503-149933503 on Build GRCh38
1626394 c.644+3G>A p.? Chr.1: 149933607-149933607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373015653 Chr.1: 149933550-149933550 on Build GRCh38
rs782381014 Chr.1: 149933407-149933407 on Build GRCh38
rs587694216 Chr.1: 149933536-149933536 on Build GRCh38
rs782272892 Chr.1: 149933554-149933555 on Build GRCh38
rs782298101 Chr.1: 149933573-149933573 on Build GRCh38
rs781926666 Chr.1: 149933433-149933433 on Build GRCh38
rs782193290 Chr.1: 149933543-149933543 on Build GRCh38
rs782687312 Chr.1: 149933373-149933373 on Build GRCh38
rs1035530047 Chr.1: 149933418-149933418 on Build GRCh38
rs955932830 Chr.1: 149933431-149933431 on Build GRCh38
rs782381450 Chr.1: 149933565-149933565 on Build GRCh38
rs587627375 Chr.1: 149933367-149933367 on Build GRCh38
rs782293441 Chr.1: 149933419-149933419 on Build GRCh38
rs781983295 Chr.1: 149933448-149933448 on Build GRCh38
rs781875491 Chr.1: 149933530-149933530 on Build GRCh38
rs370521994 Chr.1: 149933512-149933512 on Build GRCh38
rs185127068 Chr.1: 149933534-149933534 on Build GRCh38
rs782655568 Chr.1: 149933412-149933412 on Build GRCh38
rs782307985 Chr.1: 149933428-149933428 on Build GRCh38
rs782715704 Chr.1: 149933507-149933507 on Build GRCh38
rs782356629 Chr.1: 149933443-149933443 on Build GRCh38
rs782794136 Chr.1: 149933476-149933476 on Build GRCh38
rs782381124 Chr.1: 149933581-149933582 on Build GRCh38
rs369082035 Chr.1: 149933569-149933569 on Build GRCh38
rs782073735 Chr.1: 149933434-149933434 on Build GRCh38
rs782468024 Chr.1: 149933548-149933548 on Build GRCh38
rs368351924 Chr.1: 149933387-149933387 on Build GRCh38
rs782488466 Chr.1: 149933509-149933509 on Build GRCh38
rs781872294 Chr.1: 149933371-149933371 on Build GRCh38
rs371686231 Chr.1: 149933369-149933369 on Build GRCh38
rs1002669144 Chr.1: 149933394-149933394 on Build GRCh38
rs747342141 Chr.1: 149933490-149933490 on Build GRCh38
rs782540652 Chr.1: 149933372-149933372 on Build GRCh38
rs782465865 Chr.1: 149933376-149933376 on Build GRCh38
rs72692821 Chr.1: 149933386-149933386 on Build GRCh38
rs371986395 Chr.1: 149933368-149933368 on Build GRCh38
rs376638222 Chr.1: 149933564-149933564 on Build GRCh38
rs200975715 Chr.1: 149933464-149933464 on Build GRCh38

View Full Product Details