Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24268075-24268334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABGGTA
Entrez Gene ID
5875
Gene Name
Rab geranylgeranyltransferase alpha subunit
Gene Aliases
PTAR3
UniGene
Hs.377992
Gene Chromosome Location
Chr.14: 24265538-24271627 on Build GRCh38
Transcripts
NM_004581.5 NM_182836.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6425155 c.1067T>A p.L356Q Chr.14: 24268190-24268190 on Build GRCh38
5562305 c.1135C>T p.P379S Chr.14: 24268122-24268122 on Build GRCh38
6390410 c.1060T>G p.C354G Chr.14: 24268197-24268197 on Build GRCh38
3983209 c.1059-5C>A p.? Chr.14: 24268203-24268203 on Build GRCh38
1287514 c.1065G>T p.E355D Chr.14: 24268192-24268192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762482261 Chr.14: 24268167-24268167 on Build GRCh38
rs750056428 Chr.14: 24268166-24268166 on Build GRCh38
rs528953339 Chr.14: 24268095-24268095 on Build GRCh38
rs761644738 Chr.14: 24268150-24268150 on Build GRCh38
rs761736359 Chr.14: 24268248-24268248 on Build GRCh38
rs749349356 Chr.14: 24268108-24268108 on Build GRCh38
rs773362886 Chr.14: 24268104-24268104 on Build GRCh38
rs374908021 Chr.14: 24268208-24268208 on Build GRCh38
rs377178792 Chr.14: 24268109-24268109 on Build GRCh38
rs984593427 Chr.14: 24268160-24268160 on Build GRCh38
rs925486342 Chr.14: 24268288-24268288 on Build GRCh38
rs781610962 Chr.14: 24268207-24268207 on Build GRCh38
rs565706719 Chr.14: 24268173-24268173 on Build GRCh38
rs374107829 Chr.14: 24268094-24268094 on Build GRCh38
rs549076050 Chr.14: 24268125-24268125 on Build GRCh38
rs201877221 Chr.14: 24268216-24268216 on Build GRCh38
rs776574597 Chr.14: 24268145-24268145 on Build GRCh38
rs745936154 Chr.14: 24268235-24268235 on Build GRCh38
rs969206244 Chr.14: 24268260-24268260 on Build GRCh38
rs1027214823 Chr.14: 24268143-24268143 on Build GRCh38
rs556099115 Chr.14: 24268292-24268292 on Build GRCh38
rs755548071 Chr.14: 24268204-24268204 on Build GRCh38
rs770007363 Chr.14: 24268099-24268099 on Build GRCh38
rs192853311 Chr.14: 24268272-24268272 on Build GRCh38
rs368455746 Chr.14: 24268315-24268315 on Build GRCh38
rs760713919 Chr.14: 24268140-24268140 on Build GRCh38
rs776664277 Chr.14: 24268239-24268239 on Build GRCh38
rs917565940 Chr.14: 24268226-24268226 on Build GRCh38
rs771032580 Chr.14: 24268225-24268225 on Build GRCh38
rs779105303 Chr.14: 24268231-24268231 on Build GRCh38
rs768731521 Chr.14: 24268237-24268237 on Build GRCh38
rs749561397 Chr.14: 24268219-24268219 on Build GRCh38
rs370077003 Chr.14: 24268165-24268165 on Build GRCh38
rs751022237 Chr.14: 24268191-24268191 on Build GRCh38
rs536953259 Chr.14: 24268290-24268290 on Build GRCh38
rs756421546 Chr.14: 24268215-24268215 on Build GRCh38

View Full Product Details