Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 99221398-99221623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG2
Entrez Gene ID
85365
Gene Name
ALG2, alpha-1,3/1,6-mannosyltransferase
Gene Aliases
CDGIi, CMS14, CMSTA3, NET38, hALPG2
UniGene
Hs.40919
Gene Chromosome Location
Chr.9: 99216425-99221964 on Build GRCh38
Transcripts
NM_033087.3 NR_024532.1

SNP Detail

SNP ID Chromosome Location
rs368075764 Chr.9: 99221541-99221541 on Build GRCh38
rs981370536 Chr.9: 99221459-99221459 on Build GRCh38
rs370928507 Chr.9: 99221498-99221498 on Build GRCh38
rs188133373 Chr.9: 99221438-99221438 on Build GRCh38
rs959015731 Chr.9: 99221477-99221477 on Build GRCh38
rs77921350 Chr.9: 99221446-99221446 on Build GRCh38
rs1035196726 Chr.9: 99221567-99221567 on Build GRCh38
rs540482823 Chr.9: 99221483-99221483 on Build GRCh38
rs561843193 Chr.9: 99221457-99221457 on Build GRCh38
rs866814486 Chr.9: 99221530-99221530 on Build GRCh38
rs962017280 Chr.9: 99221447-99221447 on Build GRCh38
rs754476664 Chr.9: 99221549-99221549 on Build GRCh38
rs573845538 Chr.9: 99221514-99221514 on Build GRCh38
rs1032970344 Chr.9: 99221534-99221534 on Build GRCh38
rs199945081 Chr.9: 99221582-99221582 on Build GRCh38
rs969449256 Chr.9: 99221416-99221416 on Build GRCh38
rs747781559 Chr.9: 99221581-99221581 on Build GRCh38
rs757754283 Chr.9: 99221504-99221504 on Build GRCh38
rs371910648 Chr.9: 99221592-99221592 on Build GRCh38
rs932367551 Chr.9: 99221585-99221585 on Build GRCh38
rs375159554 Chr.9: 99221528-99221528 on Build GRCh38
rs769640531 Chr.9: 99221586-99221586 on Build GRCh38
rs768131229 Chr.9: 99221527-99221527 on Build GRCh38
rs557507140 Chr.9: 99221448-99221448 on Build GRCh38
rs1040699617 Chr.9: 99221537-99221537 on Build GRCh38
rs969481675 Chr.9: 99221542-99221542 on Build GRCh38
rs1015156576 Chr.9: 99221444-99221444 on Build GRCh38
rs758777017 Chr.9: 99221522-99221522 on Build GRCh38
rs982166395 Chr.9: 99221565-99221565 on Build GRCh38
rs1029544399 Chr.9: 99221419-99221419 on Build GRCh38
rs999181650 Chr.9: 99221510-99221510 on Build GRCh38
rs141064229 Chr.9: 99221418-99221418 on Build GRCh38
rs909538929 Chr.9: 99221456-99221456 on Build GRCh38
rs887967386 Chr.9: 99221435-99221435 on Build GRCh38
rs943937934 Chr.9: 99221523-99221523 on Build GRCh38
rs928095664 Chr.9: 99221437-99221437 on Build GRCh38
rs977329611 Chr.9: 99221507-99221507 on Build GRCh38
rs780869371 Chr.9: 99221558-99221558 on Build GRCh38
rs899620378 Chr.9: 99221476-99221476 on Build GRCh38
rs577994883 Chr.9: 99221462-99221462 on Build GRCh38
rs770722154 Chr.9: 99221599-99221599 on Build GRCh38
rs775003900 Chr.9: 99221557-99221557 on Build GRCh38
rs941073474 Chr.9: 99221460-99221460 on Build GRCh38
rs750973384 Chr.9: 99221520-99221520 on Build GRCh38
rs991607695 Chr.9: 99221479-99221479 on Build GRCh38
rs867509084 Chr.9: 99221574-99221574 on Build GRCh38
rs749236548 Chr.9: 99221591-99221591 on Build GRCh38

View Full Product Details