Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32931462-32931733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM98
Entrez Gene ID
26022
Gene Name
transmembrane protein 98
Gene Aliases
TADA1
UniGene
Hs.3447
Gene Chromosome Location
Chr.17: 32927910-32941649 on Build GRCh38
Transcripts
NM_001033504.1 NM_001301746.1 NM_015544.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146216 c.28_29GG>TT p.G10F Chr.17: 32931556-32931557 on Build GRCh38
6146217 c.19G>T p.V7F Chr.17: 32931547-32931547 on Build GRCh38
1710126 c.92A>G p.Y31C Chr.17: 32931620-32931620 on Build GRCh38
3516010 c.124G>A p.D42N Chr.17: 32931652-32931652 on Build GRCh38
4546478 c.3G>T p.M1I Chr.17: 32931531-32931531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765445676 Chr.17: 32931512-32931512 on Build GRCh38
rs937219646 Chr.17: 32931574-32931574 on Build GRCh38
rs927122365 Chr.17: 32931557-32931557 on Build GRCh38
rs919291494 Chr.17: 32931547-32931547 on Build GRCh38
rs72817027 Chr.17: 32931668-32931668 on Build GRCh38
rs747692196 Chr.17: 32931661-32931661 on Build GRCh38
rs548624041 Chr.17: 32931631-32931631 on Build GRCh38
rs768203786 Chr.17: 32931495-32931495 on Build GRCh38
rs183347748 Chr.17: 32931667-32931667 on Build GRCh38
rs763266387 Chr.17: 32931633-32931633 on Build GRCh38
rs750254295 Chr.17: 32931643-32931643 on Build GRCh38
rs762146943 Chr.17: 32931500-32931500 on Build GRCh38
rs756105053 Chr.17: 32931644-32931644 on Build GRCh38
rs757050347 Chr.17: 32931538-32931538 on Build GRCh38
rs753352735 Chr.17: 32931647-32931647 on Build GRCh38
rs148593533 Chr.17: 32931639-32931639 on Build GRCh38
rs562979762 Chr.17: 32931509-32931509 on Build GRCh38
rs11558978 Chr.17: 32931571-32931571 on Build GRCh38
rs754650194 Chr.17: 32931572-32931572 on Build GRCh38
rs949407898 Chr.17: 32931589-32931589 on Build GRCh38
rs754401477 Chr.17: 32931648-32931648 on Build GRCh38
rs11558976 Chr.17: 32931502-32931502 on Build GRCh38
rs750344344 Chr.17: 32931555-32931555 on Build GRCh38
rs752551213 Chr.17: 32931699-32931702 on Build GRCh38
rs751338942 Chr.17: 32931522-32931522 on Build GRCh38
rs150823123 Chr.17: 32931588-32931588 on Build GRCh38
rs775642061 Chr.17: 32931705-32931705 on Build GRCh38
rs909264097 Chr.17: 32931601-32931601 on Build GRCh38
rs139991977 Chr.17: 32931603-32931603 on Build GRCh38
rs142287076 Chr.17: 32931553-32931553 on Build GRCh38
rs758062509 Chr.17: 32931665-32931665 on Build GRCh38
rs775525737 Chr.17: 32931632-32931632 on Build GRCh38
rs747459135 Chr.17: 32931625-32931625 on Build GRCh38
rs1057055273 Chr.17: 32931581-32931581 on Build GRCh38
rs929263562 Chr.17: 32931560-32931560 on Build GRCh38
rs563295542 Chr.17: 32931692-32931692 on Build GRCh38
rs766488068 Chr.17: 32931646-32931646 on Build GRCh38
rs766381970 Chr.17: 32931503-32931503 on Build GRCh38
rs755800607 Chr.17: 32931699-32931700 on Build GRCh38
rs753929530 Chr.17: 32931506-32931506 on Build GRCh38
rs752631568 Chr.17: 32931498-32931498 on Build GRCh38
rs746408280 Chr.17: 32931691-32931691 on Build GRCh38
rs201623318 Chr.17: 32931650-32931650 on Build GRCh38
rs771487741 Chr.17: 32931629-32931629 on Build GRCh38
rs558041387 Chr.17: 32931613-32931613 on Build GRCh38
rs375260492 Chr.17: 32931630-32931630 on Build GRCh38
rs59203084 Chr.17: 32931585-32931585 on Build GRCh38
rs534917314 Chr.17: 32931584-32931584 on Build GRCh38
rs777054187 Chr.17: 32931677-32931677 on Build GRCh38
rs779651037 Chr.17: 32931566-32931566 on Build GRCh38
rs531690352 Chr.17: 32931558-32931558 on Build GRCh38
rs778466915 Chr.17: 32931579-32931579 on Build GRCh38
rs771397935 Chr.17: 32931672-32931672 on Build GRCh38

View Full Product Details