Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 157221694-157221967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIA2
Entrez Gene ID
2891
Gene Name
glutamate ionotropic receptor AMPA type subunit 2
Gene Aliases
GLUR2, GLURB, GluA2, GluR-K2, HBGR2
UniGene
Hs.32763
Gene Chromosome Location
Chr.4: 157205099-157370583 on Build GRCh38
Transcripts
NM_000826.3 NM_001083619.1 NM_001083620.1 XM_017008113.1 XM_017008114.1 XM_017008115.1 XM_017008116.1 XM_017008117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6368752 c.224A>G p.N75S Chr.4: 157221802-157221802 on Build GRCh38
5654498 c.214G>C p.A72P Chr.4: 157221792-157221792 on Build GRCh38
3373421 c.198G>C p.E66D Chr.4: 157221776-157221776 on Build GRCh38
3373420 c.198G>C p.E66D Chr.4: 157221776-157221776 on Build GRCh38
6456750 c.161C>T p.S54L Chr.4: 157221739-157221739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763718575 Chr.4: 157221857-157221857 on Build GRCh38
rs140732434 Chr.4: 157221758-157221758 on Build GRCh38
rs759505646 Chr.4: 157221845-157221845 on Build GRCh38
rs781509007 Chr.4: 157221891-157221891 on Build GRCh38
rs200344530 Chr.4: 157221850-157221850 on Build GRCh38
rs746340381 Chr.4: 157221791-157221791 on Build GRCh38
rs576771736 Chr.4: 157221887-157221887 on Build GRCh38
rs933803219 Chr.4: 157221910-157221910 on Build GRCh38
rs1002897105 Chr.4: 157221864-157221864 on Build GRCh38
rs922684665 Chr.4: 157221906-157221906 on Build GRCh38
rs780333791 Chr.4: 157221810-157221810 on Build GRCh38
rs1020233584 Chr.4: 157221860-157221860 on Build GRCh38
rs112964042 Chr.4: 157221938-157221938 on Build GRCh38
rs747246147 Chr.4: 157221816-157221816 on Build GRCh38
rs368492736 Chr.4: 157221935-157221936 on Build GRCh38
rs761840250 Chr.4: 157221828-157221828 on Build GRCh38
rs760528220 Chr.4: 157221853-157221853 on Build GRCh38
rs776909992 Chr.4: 157221822-157221822 on Build GRCh38
rs761153861 Chr.4: 157221857-157221858 on Build GRCh38
rs772460585 Chr.4: 157221803-157221803 on Build GRCh38
rs995943284 Chr.4: 157221794-157221794 on Build GRCh38
rs756783598 Chr.4: 157221743-157221743 on Build GRCh38
rs769894401 Chr.4: 157221829-157221829 on Build GRCh38
rs1035752489 Chr.4: 157221932-157221932 on Build GRCh38
rs779340546 Chr.4: 157221750-157221750 on Build GRCh38
rs866122046 Chr.4: 157221722-157221722 on Build GRCh38
rs371794977 Chr.4: 157221839-157221839 on Build GRCh38
rs374906047 Chr.4: 157221846-157221846 on Build GRCh38
rs558802212 Chr.4: 157221861-157221861 on Build GRCh38
rs13138842 Chr.4: 157221940-157221940 on Build GRCh38
rs113562722 Chr.4: 157221764-157221764 on Build GRCh38
rs555973053 Chr.4: 157221929-157221929 on Build GRCh38
rs768924263 Chr.4: 157221817-157221817 on Build GRCh38
rs748804696 Chr.4: 157221740-157221740 on Build GRCh38
rs777347678 Chr.4: 157221727-157221727 on Build GRCh38
rs1040087772 Chr.4: 157221739-157221739 on Build GRCh38
rs543978974 Chr.4: 157221920-157221920 on Build GRCh38
rs567395648 Chr.4: 157221834-157221834 on Build GRCh38
rs556868336 Chr.4: 157221779-157221779 on Build GRCh38
rs371830281 Chr.4: 157221858-157221858 on Build GRCh38
rs967767601 Chr.4: 157221862-157221865 on Build GRCh38

View Full Product Details