Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46871468-46871739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRP4
Entrez Gene ID
4038
Gene Name
LDL receptor related protein 4
Gene Aliases
CLSS, CMS17, LRP-4, LRP10, MEGF7, SOST2
UniGene
Hs.4930
Gene Chromosome Location
Chr.11: 46854715-46918622 on Build GRCh38
Transcripts
NM_002334.3 XM_011520103.2 XM_011520104.2 XM_017017734.1 NR_038909.1

Gene Symbol
LRP4-AS1
Entrez Gene ID
100507401
Gene Name
LRP4 antisense RNA 1
Gene Aliases
-
UniGene
Hs.675301
Gene Chromosome Location
Chr.11: 46846411-46874416 on Build GRCh38
Transcripts
NM_002334.3 XM_011520103.2 XM_011520104.2 XM_017017734.1 NR_038909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6501901 c.4620G>C p.E1540D Chr.11: 46871597-46871597 on Build GRCh38
3967531 c.4632C>T p.L1544L Chr.11: 46871585-46871585 on Build GRCh38
3869203 c.4617C>T p.I1539I Chr.11: 46871600-46871600 on Build GRCh38
688014 c.4691A>G p.Q1564R Chr.11: 46871526-46871526 on Build GRCh38
6209949 c.4645C>T p.R1549W Chr.11: 46871572-46871572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779760008 Chr.11: 46871491-46871493 on Build GRCh38
rs2306030 Chr.11: 46871669-46871669 on Build GRCh38
rs886048348 Chr.11: 46871571-46871571 on Build GRCh38
rs968178243 Chr.11: 46871519-46871519 on Build GRCh38
rs768340393 Chr.11: 46871648-46871649 on Build GRCh38
rs937615204 Chr.11: 46871513-46871513 on Build GRCh38
rs764949031 Chr.11: 46871572-46871572 on Build GRCh38
rs201083257 Chr.11: 46871565-46871565 on Build GRCh38
rs995601058 Chr.11: 46871686-46871686 on Build GRCh38
rs748959880 Chr.11: 46871643-46871647 on Build GRCh38
rs981590645 Chr.11: 46871695-46871695 on Build GRCh38
rs201453686 Chr.11: 46871668-46871668 on Build GRCh38
rs142352845 Chr.11: 46871583-46871583 on Build GRCh38
rs754937150 Chr.11: 46871613-46871613 on Build GRCh38
rs755863702 Chr.11: 46871616-46871616 on Build GRCh38
rs762437676 Chr.11: 46871652-46871652 on Build GRCh38
rs750052078 Chr.11: 46871582-46871582 on Build GRCh38
rs562126805 Chr.11: 46871615-46871615 on Build GRCh38
rs2306029 Chr.11: 46871557-46871557 on Build GRCh38
rs140495790 Chr.11: 46871604-46871604 on Build GRCh38
rs904671325 Chr.11: 46871688-46871688 on Build GRCh38
rs1006797545 Chr.11: 46871664-46871664 on Build GRCh38
rs145206396 Chr.11: 46871552-46871552 on Build GRCh38
rs757651850 Chr.11: 46871716-46871716 on Build GRCh38
rs765643777 Chr.11: 46871654-46871654 on Build GRCh38
rs748001051 Chr.11: 46871495-46871495 on Build GRCh38
rs183295145 Chr.11: 46871638-46871638 on Build GRCh38
rs367630949 Chr.11: 46871657-46871657 on Build GRCh38
rs61742905 Chr.11: 46871543-46871543 on Build GRCh38
rs370986346 Chr.11: 46871489-46871489 on Build GRCh38
rs147609642 Chr.11: 46871623-46871623 on Build GRCh38
rs759786789 Chr.11: 46871534-46871534 on Build GRCh38
rs534000413 Chr.11: 46871493-46871493 on Build GRCh38
rs928821259 Chr.11: 46871698-46871698 on Build GRCh38
rs762314855 Chr.11: 46871570-46871570 on Build GRCh38
rs777314555 Chr.11: 46871648-46871648 on Build GRCh38
rs1001062876 Chr.11: 46871708-46871708 on Build GRCh38
rs151261586 Chr.11: 46871600-46871600 on Build GRCh38
rs774598185 Chr.11: 46871505-46871505 on Build GRCh38
rs547976247 Chr.11: 46871693-46871693 on Build GRCh38
rs146993399 Chr.11: 46871605-46871605 on Build GRCh38
rs368518969 Chr.11: 46871637-46871637 on Build GRCh38
rs377698925 Chr.11: 46871687-46871687 on Build GRCh38
rs746049711 Chr.11: 46871633-46871633 on Build GRCh38
rs769261146 Chr.11: 46871647-46871647 on Build GRCh38

View Full Product Details