Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129670246-129670511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMCC1
Entrez Gene ID
23023
Gene Name
transmembrane and coiled-coil domain family 1
Gene Aliases
-
UniGene
Hs.477547
Gene Chromosome Location
Chr.3: 129647792-129893605 on Build GRCh38
Transcripts
NM_001017395.3 NM_001128224.2 NR_033361.1 XM_006713542.3 XM_006713543.3 XM_006713544.1 XM_006713550.3 XM_006713552.3 XM_006713553.3 XM_011512572.2 XM_011512573.2 XM_011512576.2 XM_011512579.2 XM_011512580.2 XM_011512581.2 XM_011512582.1 XM_011512583.2 XM_011512584.2 XM_017005931.1 XM_017005932.1 XM_017005933.1 XM_017005934.1 XM_017005935.1 XM_017005936.1 XM_017005937.1 XM_017005938.1 XM_017005939.1 XM_017005940.1 XM_017005941.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7178438 c.1378G>T p.D460Y Chr.3: 129670463-129670463 on Build GRCh38
1616874 c.1391A>G p.H464R Chr.3: 129670450-129670450 on Build GRCh38
7178213 c.1379_1380insA p.D460fs*6 Chr.3: 129670461-129670462 on Build GRCh38
7394026 c.1488G>T p.Q496H Chr.3: 129670353-129670353 on Build GRCh38
1670564 c.1504C>T p.R502* Chr.3: 129670337-129670337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764587813 Chr.3: 129670486-129670486 on Build GRCh38
rs1037198228 Chr.3: 129670458-129670458 on Build GRCh38
rs775919058 Chr.3: 129670444-129670444 on Build GRCh38
rs536071481 Chr.3: 129670430-129670430 on Build GRCh38
rs780103399 Chr.3: 129670324-129670324 on Build GRCh38
rs756164649 Chr.3: 129670323-129670323 on Build GRCh38
rs937104690 Chr.3: 129670317-129670317 on Build GRCh38
rs757417212 Chr.3: 129670293-129670293 on Build GRCh38
rs565949741 Chr.3: 129670371-129670371 on Build GRCh38
rs749114709 Chr.3: 129670333-129670333 on Build GRCh38
rs765853995 Chr.3: 129670439-129670439 on Build GRCh38
rs750511728 Chr.3: 129670308-129670308 on Build GRCh38
rs368370907 Chr.3: 129670432-129670432 on Build GRCh38
rs763234013 Chr.3: 129670450-129670450 on Build GRCh38
rs764236714 Chr.3: 129670281-129670281 on Build GRCh38
rs768436278 Chr.3: 129670368-129670368 on Build GRCh38
rs777393244 Chr.3: 129670441-129670449 on Build GRCh38
rs945689988 Chr.3: 129670274-129670274 on Build GRCh38
rs372355467 Chr.3: 129670295-129670295 on Build GRCh38
rs996316607 Chr.3: 129670397-129670397 on Build GRCh38
rs774568800 Chr.3: 129670487-129670487 on Build GRCh38
rs868732148 Chr.3: 129670365-129670365 on Build GRCh38
rs374324986 Chr.3: 129670375-129670375 on Build GRCh38
rs772756236 Chr.3: 129670426-129670426 on Build GRCh38
rs200100581 Chr.3: 129670443-129670443 on Build GRCh38
rs1008064816 Chr.3: 129670364-129670364 on Build GRCh38
rs1041647965 Chr.3: 129670287-129670287 on Build GRCh38
rs751740185 Chr.3: 129670289-129670289 on Build GRCh38
rs146919661 Chr.3: 129670374-129670374 on Build GRCh38

View Full Product Details