Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 71872834-71873103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D15
Entrez Gene ID
64786
Gene Name
TBC1 domain family member 15
Gene Aliases
RAB7-GAP
UniGene
Hs.284630
Gene Chromosome Location
Chr.12: 71839707-71926849 on Build GRCh38
Transcripts
NM_001146213.1 NM_001146214.1 NM_022771.4 NR_027449.1 XM_006719564.2 XM_006719565.3 XM_006719566.2 XM_006719567.2 XM_011538679.1 XM_011538680.1 XM_011538681.2 XM_011538682.1 XM_017019824.1 XM_017019825.1 XM_017019826.1 XM_017019827.1 XM_017019828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6797200 c.204+10delA p.? Chr.12: 71873013-71873013 on Build GRCh38
4044700 c.146T>C p.V49A Chr.12: 71872945-71872945 on Build GRCh38
4841868 c.182C>G p.S61C Chr.12: 71872981-71872981 on Build GRCh38
4044698 c.136G>A p.E46K Chr.12: 71872935-71872935 on Build GRCh38
6073638 c.185G>A p.S62N Chr.12: 71872984-71872984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866109080 Chr.12: 71872996-71872996 on Build GRCh38
rs1036084380 Chr.12: 71873055-71873055 on Build GRCh38
rs749842185 Chr.12: 71872886-71872887 on Build GRCh38
rs201854407 Chr.12: 71872890-71872890 on Build GRCh38
rs754934561 Chr.12: 71873020-71873020 on Build GRCh38
rs774570800 Chr.12: 71873051-71873051 on Build GRCh38
rs746957411 Chr.12: 71872896-71872896 on Build GRCh38
rs371305670 Chr.12: 71872958-71872958 on Build GRCh38
rs112382916 Chr.12: 71872922-71872922 on Build GRCh38
rs773071977 Chr.12: 71873024-71873024 on Build GRCh38
rs377339611 Chr.12: 71873040-71873040 on Build GRCh38
rs139515644 Chr.12: 71872872-71872875 on Build GRCh38
rs375369151 Chr.12: 71872970-71872970 on Build GRCh38
rs756211167 Chr.12: 71872954-71872954 on Build GRCh38
rs766654769 Chr.12: 71872933-71872933 on Build GRCh38
rs549129883 Chr.12: 71873042-71873042 on Build GRCh38
rs760019816 Chr.12: 71872935-71872935 on Build GRCh38
rs138621844 Chr.12: 71872934-71872934 on Build GRCh38
rs758155901 Chr.12: 71872981-71872981 on Build GRCh38
rs747700737 Chr.12: 71872912-71872912 on Build GRCh38
rs949032758 Chr.12: 71873039-71873039 on Build GRCh38
rs769217374 Chr.12: 71873037-71873037 on Build GRCh38
rs746867460 Chr.12: 71872993-71872993 on Build GRCh38
rs774809208 Chr.12: 71873018-71873019 on Build GRCh38
rs770671325 Chr.12: 71873049-71873049 on Build GRCh38
rs373268873 Chr.12: 71873033-71873033 on Build GRCh38
rs367780618 Chr.12: 71872875-71872878 on Build GRCh38
rs776276939 Chr.12: 71872908-71872908 on Build GRCh38
rs772760613 Chr.12: 71872921-71872921 on Build GRCh38
rs769546859 Chr.12: 71872913-71872913 on Build GRCh38
rs768057008 Chr.12: 71872943-71872943 on Build GRCh38
rs779701272 Chr.12: 71872989-71872989 on Build GRCh38
rs966508150 Chr.12: 71872892-71872892 on Build GRCh38
rs768772519 Chr.12: 71872905-71872905 on Build GRCh38
rs752755940 Chr.12: 71872944-71872944 on Build GRCh38
rs763334828 Chr.12: 71872928-71872928 on Build GRCh38
rs758161422 Chr.12: 71872886-71872886 on Build GRCh38
rs771377627 Chr.12: 71873012-71873013 on Build GRCh38
rs924575334 Chr.12: 71872942-71872942 on Build GRCh38
rs781327553 Chr.12: 71873022-71873022 on Build GRCh38

View Full Product Details