Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75050228-75050499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC2
Entrez Gene ID
3747
Gene Name
potassium voltage-gated channel subfamily C member 2
Gene Aliases
KV3.2
UniGene
Hs.27214
Gene Chromosome Location
Chr.12: 75040078-75209815 on Build GRCh38
Transcripts
NM_001260497.1 NM_001260498.1 NM_001260499.1 NM_139136.3 NM_139137.3 NM_153748.2 XM_005268855.3 XM_006719382.3 XM_006719383.3 XM_006719384.3 XM_006719385.3 XM_017019282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7447849 c.1613C>A p.S538* Chr.12: 75050392-75050392 on Build GRCh38
3786102 c.1594C>T p.L532F Chr.12: 75050411-75050411 on Build GRCh38
6073595 c.1513T>A p.S505T Chr.12: 75050492-75050492 on Build GRCh38
6073598 c.1563G>C p.Q521H Chr.12: 75050442-75050442 on Build GRCh38
943421 c.1594C>A p.L532I Chr.12: 75050411-75050411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201050565 Chr.12: 75050475-75050475 on Build GRCh38
rs200429167 Chr.12: 75050474-75050474 on Build GRCh38
rs781461580 Chr.12: 75050405-75050405 on Build GRCh38
rs111701449 Chr.12: 75050267-75050267 on Build GRCh38
rs770646073 Chr.12: 75050414-75050414 on Build GRCh38
rs201668041 Chr.12: 75050413-75050413 on Build GRCh38
rs898800620 Chr.12: 75050382-75050382 on Build GRCh38
rs202162135 Chr.12: 75050440-75050440 on Build GRCh38
rs79815499 Chr.12: 75050409-75050409 on Build GRCh38
rs775119334 Chr.12: 75050441-75050441 on Build GRCh38
rs767895720 Chr.12: 75050306-75050306 on Build GRCh38
rs201137392 Chr.12: 75050396-75050396 on Build GRCh38
rs201383604 Chr.12: 75050372-75050372 on Build GRCh38
rs762174667 Chr.12: 75050470-75050470 on Build GRCh38
rs763831110 Chr.12: 75050459-75050459 on Build GRCh38
rs148259589 Chr.12: 75050268-75050268 on Build GRCh38
rs200570594 Chr.12: 75050445-75050445 on Build GRCh38
rs967789486 Chr.12: 75050383-75050383 on Build GRCh38
rs774278663 Chr.12: 75050426-75050426 on Build GRCh38
rs755168734 Chr.12: 75050380-75050380 on Build GRCh38
rs751365248 Chr.12: 75050294-75050294 on Build GRCh38
rs575894642 Chr.12: 75050323-75050323 on Build GRCh38
rs977872075 Chr.12: 75050391-75050391 on Build GRCh38
rs750903919 Chr.12: 75050346-75050346 on Build GRCh38
rs747182899 Chr.12: 75050374-75050374 on Build GRCh38
rs776202734 Chr.12: 75050462-75050462 on Build GRCh38
rs758436408 Chr.12: 75050369-75050369 on Build GRCh38
rs745827163 Chr.12: 75050436-75050436 on Build GRCh38
rs753519354 Chr.12: 75050253-75050253 on Build GRCh38
rs571188150 Chr.12: 75050337-75050337 on Build GRCh38
rs201942907 Chr.12: 75050397-75050397 on Build GRCh38
rs200410459 Chr.12: 75050401-75050401 on Build GRCh38
rs923590869 Chr.12: 75050407-75050407 on Build GRCh38

View Full Product Details