Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116620883-116621116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNLIPRP2
Entrez Gene ID
5408
Gene Name
pancreatic lipase related protein 2 (gene/pseudogene)
Gene Aliases
PLRP2
UniGene
Hs.423598
Gene Chromosome Location
Chr.10: 116620953-116645143 on Build GRCh38
Transcripts
NM_005396.4 NR_103727.1

SNP Detail

SNP ID Chromosome Location
rs377437793 Chr.10: 116621059-116621059 on Build GRCh38
rs376047295 Chr.10: 116621026-116621026 on Build GRCh38
rs782730889 Chr.10: 116621027-116621027 on Build GRCh38
rs781916846 Chr.10: 116620995-116620995 on Build GRCh38
rs782428368 Chr.10: 116621015-116621015 on Build GRCh38
rs782189412 Chr.10: 116621008-116621008 on Build GRCh38
rs575850900 Chr.10: 116620953-116620953 on Build GRCh38
rs980905625 Chr.10: 116620904-116620904 on Build GRCh38
rs782644182 Chr.10: 116620993-116620993 on Build GRCh38
rs543019753 Chr.10: 116620964-116620964 on Build GRCh38
rs781977424 Chr.10: 116620936-116620936 on Build GRCh38
rs782050754 Chr.10: 116620957-116620957 on Build GRCh38
rs576982702 Chr.10: 116621007-116621007 on Build GRCh38
rs782646197 Chr.10: 116621018-116621018 on Build GRCh38
rs150533225 Chr.10: 116621066-116621066 on Build GRCh38
rs370011255 Chr.10: 116621046-116621046 on Build GRCh38
rs1033856423 Chr.10: 116620915-116620915 on Build GRCh38
rs781806020 Chr.10: 116620955-116620955 on Build GRCh38
rs147974184 Chr.10: 116620943-116620943 on Build GRCh38
rs781886949 Chr.10: 116620981-116620981 on Build GRCh38
rs782809478 Chr.10: 116620977-116620977 on Build GRCh38
rs963588116 Chr.10: 116620944-116620944 on Build GRCh38
rs141631824 Chr.10: 116620991-116620991 on Build GRCh38
rs3010502 Chr.10: 116620999-116620999 on Build GRCh38
rs559522197 Chr.10: 116621024-116621024 on Build GRCh38
rs782702305 Chr.10: 116620999-116620999 on Build GRCh38
rs572986730 Chr.10: 116621045-116621045 on Build GRCh38
rs782390014 Chr.10: 116620926-116620926 on Build GRCh38

View Full Product Details