Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55401177-55401406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LANCL2
Entrez Gene ID
55915
Gene Name
LanC like 2
Gene Aliases
GPR69B, TASP
UniGene
Hs.595384
Gene Chromosome Location
Chr.7: 55365448-55433742 on Build GRCh38
Transcripts
NM_018697.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1239311 c.704G>A p.G235D Chr.7: 55401199-55401199 on Build GRCh38
4755638 c.755A>G p.Y252C Chr.7: 55401250-55401250 on Build GRCh38
3698484 c.731A>C p.K244T Chr.7: 55401226-55401226 on Build GRCh38
1256315 c.747G>A p.P249P Chr.7: 55401242-55401242 on Build GRCh38
6178042 c.766C>T p.R256W Chr.7: 55401261-55401261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371227788 Chr.7: 55401371-55401371 on Build GRCh38
rs988185931 Chr.7: 55401206-55401207 on Build GRCh38
rs746631664 Chr.7: 55401340-55401340 on Build GRCh38
rs914503446 Chr.7: 55401336-55401336 on Build GRCh38
rs749653004 Chr.7: 55401223-55401223 on Build GRCh38
rs149433421 Chr.7: 55401229-55401229 on Build GRCh38
rs768031875 Chr.7: 55401230-55401230 on Build GRCh38
rs769859480 Chr.7: 55401236-55401236 on Build GRCh38
rs201326525 Chr.7: 55401274-55401274 on Build GRCh38
rs754151076 Chr.7: 55401325-55401325 on Build GRCh38
rs200888580 Chr.7: 55401362-55401362 on Build GRCh38
rs752840045 Chr.7: 55401307-55401307 on Build GRCh38
rs35961951 Chr.7: 55401265-55401266 on Build GRCh38
rs550637807 Chr.7: 55401275-55401275 on Build GRCh38
rs779386534 Chr.7: 55401338-55401338 on Build GRCh38
rs779592312 Chr.7: 55401228-55401228 on Build GRCh38
rs200782197 Chr.7: 55401218-55401218 on Build GRCh38
rs768426127 Chr.7: 55401351-55401351 on Build GRCh38
rs761840899 Chr.7: 55401235-55401235 on Build GRCh38
rs373843323 Chr.7: 55401342-55401342 on Build GRCh38
rs756671170 Chr.7: 55401212-55401212 on Build GRCh38
rs773084629 Chr.7: 55401241-55401241 on Build GRCh38
rs946401245 Chr.7: 55401222-55401222 on Build GRCh38
rs377444813 Chr.7: 55401272-55401272 on Build GRCh38
rs368760594 Chr.7: 55401335-55401335 on Build GRCh38
rs376348172 Chr.7: 55401354-55401354 on Build GRCh38
rs569380328 Chr.7: 55401359-55401359 on Build GRCh38
rs185788792 Chr.7: 55401378-55401378 on Build GRCh38
rs758828282 Chr.7: 55401316-55401316 on Build GRCh38
rs140738512 Chr.7: 55401262-55401262 on Build GRCh38
rs1019418856 Chr.7: 55401314-55401314 on Build GRCh38
rs901391612 Chr.7: 55401254-55401254 on Build GRCh38
rs374460024 Chr.7: 55401242-55401242 on Build GRCh38
rs771695305 Chr.7: 55401226-55401226 on Build GRCh38
rs776342559 Chr.7: 55401234-55401234 on Build GRCh38

View Full Product Details