Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207466691-207466942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CR2
Entrez Gene ID
1380
Gene Name
complement component 3d receptor 2
Gene Aliases
C3DR, CD21, CR, CVID7, SLEB9
UniGene
Hs.445757
Gene Chromosome Location
Chr.1: 207454300-207489895 on Build GRCh38
Transcripts
NM_001006658.2 NM_001877.4 XM_011509206.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5815501 c.339T>A p.S113S Chr.1: 207466806-207466806 on Build GRCh38
5774195 c.274C>A p.P92T Chr.1: 207466741-207466741 on Build GRCh38
4976964 c.434C>T p.T145I Chr.1: 207466901-207466901 on Build GRCh38
3482629 c.417G>A p.G139G Chr.1: 207466884-207466884 on Build GRCh38
3482628 c.346T>G p.F116V Chr.1: 207466813-207466813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563764676 Chr.1: 207466886-207466886 on Build GRCh38
rs764012037 Chr.1: 207466730-207466730 on Build GRCh38
rs543886209 Chr.1: 207466856-207466856 on Build GRCh38
rs963901983 Chr.1: 207466821-207466821 on Build GRCh38
rs374754086 Chr.1: 207466792-207466792 on Build GRCh38
rs763604504 Chr.1: 207466884-207466884 on Build GRCh38
rs769241648 Chr.1: 207466824-207466824 on Build GRCh38
rs1037853856 Chr.1: 207466776-207466776 on Build GRCh38
rs761774180 Chr.1: 207466887-207466887 on Build GRCh38
rs903091303 Chr.1: 207466910-207466910 on Build GRCh38
rs750210048 Chr.1: 207466892-207466892 on Build GRCh38
rs986427524 Chr.1: 207466915-207466915 on Build GRCh38
rs747481282 Chr.1: 207466819-207466819 on Build GRCh38
rs141434018 Chr.1: 207466763-207466763 on Build GRCh38
rs765552836 Chr.1: 207466912-207466912 on Build GRCh38
rs201962814 Chr.1: 207466830-207466830 on Build GRCh38
rs750220618 Chr.1: 207466750-207466750 on Build GRCh38
rs748428964 Chr.1: 207466853-207466853 on Build GRCh38
rs775187056 Chr.1: 207466862-207466862 on Build GRCh38
rs777726381 Chr.1: 207466816-207466816 on Build GRCh38
rs890412935 Chr.1: 207466879-207466879 on Build GRCh38
rs757158490 Chr.1: 207466735-207466735 on Build GRCh38
rs767784162 Chr.1: 207466879-207466879 on Build GRCh38
rs138096192 Chr.1: 207466842-207466842 on Build GRCh38
rs201017642 Chr.1: 207466891-207466891 on Build GRCh38
rs371546344 Chr.1: 207466743-207466743 on Build GRCh38
rs755202444 Chr.1: 207466773-207466773 on Build GRCh38
rs762329677 Chr.1: 207466874-207466874 on Build GRCh38
rs753028434 Chr.1: 207466918-207466918 on Build GRCh38
rs762434408 Chr.1: 207466878-207466878 on Build GRCh38
rs200595124 Chr.1: 207466843-207466843 on Build GRCh38
rs756014299 Chr.1: 207466895-207466895 on Build GRCh38
rs1016408105 Chr.1: 207466777-207466777 on Build GRCh38
rs561466012 Chr.1: 207466836-207466836 on Build GRCh38
rs751574877 Chr.1: 207466734-207466734 on Build GRCh38
rs748210120 Chr.1: 207466795-207466795 on Build GRCh38
rs543047560 Chr.1: 207466860-207466860 on Build GRCh38
rs367581933 Chr.1: 207466801-207466801 on Build GRCh38
rs201032699 Chr.1: 207466756-207466756 on Build GRCh38

View Full Product Details