Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43646338-43646611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLAUR
Entrez Gene ID
5329
Gene Name
plasminogen activator, urokinase receptor
Gene Aliases
CD87, U-PAR, UPAR, URKR
UniGene
Hs.466871
Gene Chromosome Location
Chr.19: 43646095-43670346 on Build GRCh38
Transcripts
NM_001005376.2 XM_011527029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
190375 c.757C>T p.R253C Chr.19: 43646546-43646546 on Build GRCh38
1394304 c.758G>A p.R253H Chr.19: 43646545-43646545 on Build GRCh38
6026218 c.802C>A p.L268M Chr.19: 43646501-43646501 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1011613108 Chr.19: 43646587-43646587 on Build GRCh38
rs548739224 Chr.19: 43646442-43646442 on Build GRCh38
rs117363374 Chr.19: 43646454-43646454 on Build GRCh38
rs991706836 Chr.19: 43646374-43646374 on Build GRCh38
rs377422818 Chr.19: 43646546-43646546 on Build GRCh38
rs1040791504 Chr.19: 43646485-43646485 on Build GRCh38
rs988607163 Chr.19: 43646559-43646559 on Build GRCh38
rs568562599 Chr.19: 43646445-43646445 on Build GRCh38
rs895593961 Chr.19: 43646438-43646438 on Build GRCh38
rs528271761 Chr.19: 43646372-43646372 on Build GRCh38
rs1047160819 Chr.19: 43646433-43646433 on Build GRCh38
rs138522808 Chr.19: 43646525-43646525 on Build GRCh38
rs186353857 Chr.19: 43646463-43646463 on Build GRCh38
rs377483791 Chr.19: 43646449-43646449 on Build GRCh38
rs768424274 Chr.19: 43646441-43646441 on Build GRCh38
rs1013749244 Chr.19: 43646440-43646440 on Build GRCh38
rs1002461359 Chr.19: 43646466-43646466 on Build GRCh38
rs993617911 Chr.19: 43646404-43646404 on Build GRCh38
rs943750018 Chr.19: 43646437-43646437 on Build GRCh38
rs374159845 Chr.19: 43646545-43646545 on Build GRCh38
rs570791364 Chr.19: 43646521-43646521 on Build GRCh38
rs886889745 Chr.19: 43646436-43646436 on Build GRCh38
rs767351281 Chr.19: 43646511-43646511 on Build GRCh38
rs751812356 Chr.19: 43646548-43646548 on Build GRCh38
rs750080469 Chr.19: 43646459-43646459 on Build GRCh38
rs373237015 Chr.19: 43646411-43646411 on Build GRCh38
rs761406058 Chr.19: 43646444-43646444 on Build GRCh38
rs920131254 Chr.19: 43646380-43646380 on Build GRCh38

View Full Product Details