Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 71884830-71885081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D15
Entrez Gene ID
64786
Gene Name
TBC1 domain family member 15
Gene Aliases
RAB7-GAP
UniGene
Hs.284630
Gene Chromosome Location
Chr.12: 71839707-71926849 on Build GRCh38
Transcripts
NM_001146213.1 NM_001146214.1 NM_022771.4 NR_027449.1 XM_006719564.2 XM_006719565.3 XM_006719566.2 XM_006719567.2 XM_011538679.1 XM_011538680.1 XM_011538681.2 XM_011538682.1 XM_017019824.1 XM_017019825.1 XM_017019826.1 XM_017019827.1 XM_017019828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7263068 c.475G>A p.V159I Chr.12: 71884942-71884942 on Build GRCh38
5970836 c.506A>G p.D169G Chr.12: 71884973-71884973 on Build GRCh38
273332 c.439T>C p.W147R Chr.12: 71884906-71884906 on Build GRCh38
4428959 c.547T>G p.L183V Chr.12: 71885014-71885014 on Build GRCh38
5677659 c.426A>T p.K142N Chr.12: 71884893-71884893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973658650 Chr.12: 71884878-71884878 on Build GRCh38
rs756666077 Chr.12: 71884918-71884918 on Build GRCh38
rs908896641 Chr.12: 71884873-71884873 on Build GRCh38
rs765199238 Chr.12: 71884989-71884989 on Build GRCh38
rs755238663 Chr.12: 71885032-71885032 on Build GRCh38
rs893750024 Chr.12: 71885011-71885011 on Build GRCh38
rs559036629 Chr.12: 71884971-71884971 on Build GRCh38
rs766770522 Chr.12: 71884854-71884854 on Build GRCh38
rs200459507 Chr.12: 71884969-71884969 on Build GRCh38
rs373705972 Chr.12: 71884999-71884999 on Build GRCh38
rs765976613 Chr.12: 71885026-71885026 on Build GRCh38
rs761737005 Chr.12: 71884987-71884987 on Build GRCh38
rs775389540 Chr.12: 71884879-71884879 on Build GRCh38
rs573583308 Chr.12: 71884958-71884958 on Build GRCh38
rs146267554 Chr.12: 71884902-71884902 on Build GRCh38
rs920879951 Chr.12: 71884894-71884894 on Build GRCh38
rs749343509 Chr.12: 71885044-71885044 on Build GRCh38
rs369961626 Chr.12: 71885034-71885034 on Build GRCh38
rs760324166 Chr.12: 71884977-71884977 on Build GRCh38
rs774668422 Chr.12: 71884973-71884973 on Build GRCh38
rs376375651 Chr.12: 71884914-71884914 on Build GRCh38
rs751226919 Chr.12: 71885031-71885031 on Build GRCh38
rs34185988 Chr.12: 71884925-71884925 on Build GRCh38
rs757925435 Chr.12: 71885007-71885007 on Build GRCh38
rs375567304 Chr.12: 71885035-71885035 on Build GRCh38
rs1012069698 Chr.12: 71885013-71885013 on Build GRCh38
rs755400995 Chr.12: 71884953-71884953 on Build GRCh38
rs1057252106 Chr.12: 71884941-71884941 on Build GRCh38
rs376625945 Chr.12: 71885033-71885033 on Build GRCh38
rs760545671 Chr.12: 71884897-71884897 on Build GRCh38
rs772446238 Chr.12: 71885043-71885043 on Build GRCh38
rs779730555 Chr.12: 71884942-71884942 on Build GRCh38
rs200977186 Chr.12: 71884938-71884938 on Build GRCh38
rs749914058 Chr.12: 71884996-71884996 on Build GRCh38
rs902198254 Chr.12: 71884947-71884947 on Build GRCh38
rs776349137 Chr.12: 71884983-71884983 on Build GRCh38
rs748701195 Chr.12: 71884956-71884956 on Build GRCh38
rs144254414 Chr.12: 71884980-71884980 on Build GRCh38
rs190286145 Chr.12: 71884950-71884950 on Build GRCh38
rs551976520 Chr.12: 71884928-71884928 on Build GRCh38
rs149981863 Chr.12: 71884939-71884939 on Build GRCh38

View Full Product Details