Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143437985-143438226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3H3
Entrez Gene ID
23144
Gene Name
zinc finger CCCH-type containing 3
Gene Aliases
ZC3HDC3
UniGene
Hs.521915
Gene Chromosome Location
Chr.8: 143437655-143549729 on Build GRCh38
Transcripts
NM_015117.2 XM_006716536.3 XM_017013248.1 XM_017013249.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5457379 c.2840G>A p.R947H Chr.8: 143438063-143438063 on Build GRCh38
4839902 c.2842C>A p.L948M Chr.8: 143438061-143438061 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753215909 Chr.8: 143438051-143438051 on Build GRCh38
rs200065716 Chr.8: 143438173-143438174 on Build GRCh38
rs1025278189 Chr.8: 143438182-143438182 on Build GRCh38
rs995118088 Chr.8: 143438173-143438173 on Build GRCh38
rs1039856459 Chr.8: 143438126-143438126 on Build GRCh38
rs767153451 Chr.8: 143438022-143438022 on Build GRCh38
rs935251312 Chr.8: 143438090-143438090 on Build GRCh38
rs184233972 Chr.8: 143438161-143438161 on Build GRCh38
rs766025299 Chr.8: 143438039-143438039 on Build GRCh38
rs745662796 Chr.8: 143438054-143438054 on Build GRCh38
rs187902174 Chr.8: 143438092-143438092 on Build GRCh38
rs978316704 Chr.8: 143438123-143438123 on Build GRCh38
rs749938120 Chr.8: 143438036-143438036 on Build GRCh38
rs768580293 Chr.8: 143438119-143438119 on Build GRCh38
rs958777576 Chr.8: 143438010-143438010 on Build GRCh38
rs761902233 Chr.8: 143438135-143438135 on Build GRCh38
rs201332231 Chr.8: 143438042-143438042 on Build GRCh38
rs991992037 Chr.8: 143438030-143438030 on Build GRCh38
rs191930318 Chr.8: 143438118-143438118 on Build GRCh38
rs762816332 Chr.8: 143438005-143438005 on Build GRCh38
rs754205521 Chr.8: 143438041-143438041 on Build GRCh38
rs761779578 Chr.8: 143438021-143438021 on Build GRCh38
rs76247763 Chr.8: 143438201-143438201 on Build GRCh38
rs771104198 Chr.8: 143438107-143438107 on Build GRCh38
rs530372498 Chr.8: 143438011-143438011 on Build GRCh38
rs760314103 Chr.8: 143438038-143438038 on Build GRCh38
rs200072279 Chr.8: 143438117-143438117 on Build GRCh38
rs768518961 Chr.8: 143438006-143438006 on Build GRCh38
rs747078975 Chr.8: 143438106-143438106 on Build GRCh38
rs779209233 Chr.8: 143438046-143438046 on Build GRCh38
rs149968226 Chr.8: 143438061-143438061 on Build GRCh38
rs534309623 Chr.8: 143438162-143438162 on Build GRCh38
rs936819361 Chr.8: 143438174-143438174 on Build GRCh38
rs887336560 Chr.8: 143438191-143438191 on Build GRCh38
rs774234200 Chr.8: 143438128-143438128 on Build GRCh38

View Full Product Details