Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186311471-186311700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRG4
Entrez Gene ID
10216
Gene Name
proteoglycan 4
Gene Aliases
CACP, HAPO, JCAP, MSF, SZP
UniGene
Hs.647723
Gene Chromosome Location
Chr.1: 186296273-186314562 on Build GRCh38
Transcripts
NM_001127708.2 NM_001127709.2 NM_001127710.2 NM_001303232.1 NM_005807.4 XM_017000002.1 XM_017000003.1 NM_003292.2

Gene Symbol
TPR
Entrez Gene ID
7175
Gene Name
translocated promoter region, nuclear basket protein
Gene Aliases
-
UniGene
Hs.279640
Gene Chromosome Location
Chr.1: 186311652-186375325 on Build GRCh38
Transcripts
NM_001127708.2 NM_001127709.2 NM_001127710.2 NM_001303232.1 NM_005807.4 XM_017000002.1 XM_017000003.1 NM_003292.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4995336 c.3712G>A p.G1238R Chr.1: 186311515-186311515 on Build GRCh38
1315796 c.3737C>T p.A1246V Chr.1: 186311540-186311540 on Build GRCh38
337630 c.3700A>G p.K1234E Chr.1: 186311503-186311503 on Build GRCh38
5616680 c.3678A>G p.A1226A Chr.1: 186311481-186311481 on Build GRCh38
302986 c.3777G>A p.V1259V Chr.1: 186311580-186311580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770837321 Chr.1: 186311558-186311558 on Build GRCh38
rs779247168 Chr.1: 186311541-186311541 on Build GRCh38
rs757584066 Chr.1: 186311523-186311523 on Build GRCh38
rs369724686 Chr.1: 186311540-186311540 on Build GRCh38
rs571703726 Chr.1: 186311549-186311549 on Build GRCh38
rs762407968 Chr.1: 186311599-186311599 on Build GRCh38
rs550560270 Chr.1: 186311533-186311533 on Build GRCh38
rs568855633 Chr.1: 186311596-186311596 on Build GRCh38
rs750699622 Chr.1: 186311629-186311629 on Build GRCh38
rs750923984 Chr.1: 186311531-186311531 on Build GRCh38
rs766759179 Chr.1: 186311643-186311643 on Build GRCh38
rs764799096 Chr.1: 186311496-186311496 on Build GRCh38
rs759379313 Chr.1: 186311575-186311575 on Build GRCh38
rs780627785 Chr.1: 186311535-186311535 on Build GRCh38
rs532167626 Chr.1: 186311517-186311517 on Build GRCh38
rs1048064484 Chr.1: 186311606-186311606 on Build GRCh38
rs751187594 Chr.1: 186311604-186311604 on Build GRCh38
rs1006844016 Chr.1: 186311664-186311664 on Build GRCh38
rs756527086 Chr.1: 186311632-186311632 on Build GRCh38
rs375980337 Chr.1: 186311501-186311501 on Build GRCh38
rs140871526 Chr.1: 186311505-186311505 on Build GRCh38
rs763631861 Chr.1: 186311600-186311600 on Build GRCh38
rs147581705 Chr.1: 186311590-186311590 on Build GRCh38
rs959104016 Chr.1: 186311653-186311653 on Build GRCh38
rs767915239 Chr.1: 186311622-186311622 on Build GRCh38
rs199887246 Chr.1: 186311612-186311612 on Build GRCh38
rs536374246 Chr.1: 186311658-186311658 on Build GRCh38
rs554363596 Chr.1: 186311662-186311662 on Build GRCh38
rs751240720 Chr.1: 186311521-186311521 on Build GRCh38
rs372007052 Chr.1: 186311562-186311562 on Build GRCh38

View Full Product Details