Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19681475-19681700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC47A2
Entrez Gene ID
146802
Gene Name
solute carrier family 47 member 2
Gene Aliases
MATE2, MATE2-B, MATE2-K, MATE2K
UniGene
Hs.126830
Gene Chromosome Location
Chr.17: 19678313-19718979 on Build GRCh38
Transcripts
NM_001099646.1 NM_001256663.1 NM_152908.3 NR_135624.1 NR_135625.1 XM_017024221.1 XM_017024222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
976440 c.1354G>T p.G452C Chr.17: 19681589-19681589 on Build GRCh38
1710006 c.1232G>A p.G411E Chr.17: 19681645-19681645 on Build GRCh38
3515063 c.1220G>A p.G407E Chr.17: 19681657-19681657 on Build GRCh38
976438 c.1393_1396delATGA p.M465fs*75 Chr.17: 19681547-19681550 on Build GRCh38
1659305 c.1282G>A p.G428S Chr.17: 19681661-19681661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368029589 Chr.17: 19681517-19681517 on Build GRCh38
rs534912913 Chr.17: 19681558-19681558 on Build GRCh38
rs781073358 Chr.17: 19681673-19681673 on Build GRCh38
rs748154238 Chr.17: 19681570-19681578 on Build GRCh38
rs767987489 Chr.17: 19681641-19681641 on Build GRCh38
rs759592392 Chr.17: 19681518-19681518 on Build GRCh38
rs139110700 Chr.17: 19681583-19681583 on Build GRCh38
rs1009609609 Chr.17: 19681537-19681537 on Build GRCh38
rs753371039 Chr.17: 19681565-19681565 on Build GRCh38
rs776482989 Chr.17: 19681589-19681589 on Build GRCh38
rs201704161 Chr.17: 19681594-19681594 on Build GRCh38
rs373893783 Chr.17: 19681508-19681508 on Build GRCh38
rs763270502 Chr.17: 19681539-19681539 on Build GRCh38
rs761275367 Chr.17: 19681646-19681646 on Build GRCh38
rs754493465 Chr.17: 19681573-19681573 on Build GRCh38
rs764600870 Chr.17: 19681654-19681654 on Build GRCh38
rs775664257 Chr.17: 19681599-19681599 on Build GRCh38
rs937997773 Chr.17: 19681501-19681501 on Build GRCh38
rs769786354 Chr.17: 19681675-19681675 on Build GRCh38
rs139862736 Chr.17: 19681509-19681509 on Build GRCh38
rs374248552 Chr.17: 19681564-19681564 on Build GRCh38
rs771004939 Chr.17: 19681513-19681513 on Build GRCh38
rs762219698 Chr.17: 19681639-19681639 on Build GRCh38
rs778596221 Chr.17: 19681576-19681576 on Build GRCh38
rs745747264 Chr.17: 19681674-19681674 on Build GRCh38
rs774690198 Chr.17: 19681620-19681620 on Build GRCh38
rs151173119 Chr.17: 19681499-19681499 on Build GRCh38
rs754441994 Chr.17: 19681530-19681530 on Build GRCh38
rs34399035 Chr.17: 19681658-19681658 on Build GRCh38
rs200937883 Chr.17: 19681577-19681577 on Build GRCh38
rs968860036 Chr.17: 19681534-19681534 on Build GRCh38
rs751938252 Chr.17: 19681556-19681556 on Build GRCh38
rs758146654 Chr.17: 19681578-19681578 on Build GRCh38
rs757161308 Chr.17: 19681672-19681672 on Build GRCh38
rs150768699 Chr.17: 19681540-19681540 on Build GRCh38
rs200117290 Chr.17: 19681590-19681590 on Build GRCh38
rs750930475 Chr.17: 19681642-19681642 on Build GRCh38
rs969962303 Chr.17: 19681588-19681588 on Build GRCh38
rs539685879 Chr.17: 19681659-19681659 on Build GRCh38
rs372698065 Chr.17: 19681606-19681606 on Build GRCh38
rs371452443 Chr.17: 19681498-19681498 on Build GRCh38
rs751378851 Chr.17: 19681664-19681664 on Build GRCh38
rs746867638 Chr.17: 19681584-19681584 on Build GRCh38
rs752332889 Chr.17: 19681587-19681587 on Build GRCh38
rs764308768 Chr.17: 19681619-19681619 on Build GRCh38

View Full Product Details