Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57741696-57741937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf31
Entrez Gene ID
280636
Gene Name
chromosome 11 open reading frame 31
Gene Aliases
C17orf10, SELH
UniGene
Hs.655373
Gene Chromosome Location
Chr.11: 57741250-57743554 on Build GRCh38
Transcripts
NM_001321335.1 NM_170746.3 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001321335.1 NM_170746.3 NR_037646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4626847 c.112A>G p.I38V Chr.11: 57741707-57741707 on Build GRCh38
1726639 c.242T>C p.V81A Chr.11: 57741928-57741928 on Build GRCh38
4645053 c.180G>A p.L60L Chr.11: 57741866-57741866 on Build GRCh38
3716806 c.122+6G>A p.? Chr.11: 57741723-57741723 on Build GRCh38
7357968 c.106G>T p.V36F Chr.11: 57741701-57741701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112893850 Chr.11: 57741718-57741718 on Build GRCh38
rs756524130 Chr.11: 57741803-57741803 on Build GRCh38
rs765173414 Chr.11: 57741839-57741839 on Build GRCh38
rs550979949 Chr.11: 57741770-57741770 on Build GRCh38
rs768867284 Chr.11: 57741900-57741900 on Build GRCh38
rs766222383 Chr.11: 57741762-57741762 on Build GRCh38
rs757723961 Chr.11: 57741886-57741886 on Build GRCh38
rs529454955 Chr.11: 57741735-57741735 on Build GRCh38
rs772219776 Chr.11: 57741776-57741776 on Build GRCh38
rs551304941 Chr.11: 57741756-57741756 on Build GRCh38
rs756702621 Chr.11: 57741764-57741764 on Build GRCh38
rs779812647 Chr.11: 57741796-57741796 on Build GRCh38
rs569454280 Chr.11: 57741762-57741762 on Build GRCh38
rs777390256 Chr.11: 57741866-57741866 on Build GRCh38
rs11552989 Chr.11: 57741820-57741820 on Build GRCh38
rs774698908 Chr.11: 57741774-57741774 on Build GRCh38
rs1001289016 Chr.11: 57741831-57741831 on Build GRCh38
rs768334379 Chr.11: 57741827-57741827 on Build GRCh38
rs752418887 Chr.11: 57741760-57741760 on Build GRCh38
rs774408150 Chr.11: 57741745-57741745 on Build GRCh38
rs1045583815 Chr.11: 57741798-57741799 on Build GRCh38
rs373321278 Chr.11: 57741717-57741717 on Build GRCh38
rs759950859 Chr.11: 57741765-57741765 on Build GRCh38
rs759396454 Chr.11: 57741749-57741749 on Build GRCh38
rs770358910 Chr.11: 57741726-57741726 on Build GRCh38
rs868133397 Chr.11: 57741828-57741828 on Build GRCh38
rs761455323 Chr.11: 57741788-57741788 on Build GRCh38
rs764367571 Chr.11: 57741874-57741874 on Build GRCh38
rs764203025 Chr.11: 57741790-57741790 on Build GRCh38
rs757041217 Chr.11: 57741909-57741909 on Build GRCh38
rs376289885 Chr.11: 57741725-57741725 on Build GRCh38
rs762608791 Chr.11: 57741919-57741919 on Build GRCh38
rs747935080 Chr.11: 57741906-57741906 on Build GRCh38
rs766674032 Chr.11: 57741738-57741738 on Build GRCh38
rs145767870 Chr.11: 57741761-57741762 on Build GRCh38
rs764144437 Chr.11: 57741781-57741781 on Build GRCh38
rs371576974 Chr.11: 57741899-57741899 on Build GRCh38
rs771399022 Chr.11: 57741811-57741811 on Build GRCh38
rs180731500 Chr.11: 57741826-57741826 on Build GRCh38
rs746001100 Chr.11: 57741890-57741890 on Build GRCh38
rs754260556 Chr.11: 57741761-57741761 on Build GRCh38
rs760435060 Chr.11: 57741780-57741780 on Build GRCh38
rs778867933 Chr.11: 57741815-57741815 on Build GRCh38
rs367704288 Chr.11: 57741865-57741865 on Build GRCh38
rs758034360 Chr.11: 57741849-57741850 on Build GRCh38
rs778109616 Chr.11: 57741769-57741769 on Build GRCh38
rs533666992 Chr.11: 57741807-57741807 on Build GRCh38
rs1011226756 Chr.11: 57741884-57741884 on Build GRCh38
rs745912453 Chr.11: 57741819-57741819 on Build GRCh38
rs758146744 Chr.11: 57741795-57741795 on Build GRCh38
rs753100093 Chr.11: 57741809-57741809 on Build GRCh38
rs201826539 Chr.11: 57741852-57741852 on Build GRCh38
rs200278935 Chr.11: 57741862-57741862 on Build GRCh38
rs776313439 Chr.11: 57741902-57741902 on Build GRCh38
rs537644420 Chr.11: 57741896-57741896 on Build GRCh38
rs369458858 Chr.11: 57741768-57741768 on Build GRCh38
rs778389647 Chr.11: 57741772-57741772 on Build GRCh38
rs751242092 Chr.11: 57741858-57741858 on Build GRCh38
rs756012733 Chr.11: 57741734-57741735 on Build GRCh38
rs377525426 Chr.11: 57741793-57741793 on Build GRCh38
rs770899758 Chr.11: 57741772-57741772 on Build GRCh38
rs753151249 Chr.11: 57741799-57741799 on Build GRCh38
rs753708997 Chr.11: 57741782-57741782 on Build GRCh38
rs965666608 Chr.11: 57741771-57741771 on Build GRCh38
rs376466544 Chr.11: 57741878-57741878 on Build GRCh38
rs892844080 Chr.11: 57741872-57741872 on Build GRCh38
rs749751894 Chr.11: 57741810-57741810 on Build GRCh38
rs759754533 Chr.11: 57741775-57741775 on Build GRCh38
rs775670097 Chr.11: 57741822-57741822 on Build GRCh38
rs776421824 Chr.11: 57741832-57741832 on Build GRCh38
rs539705727 Chr.11: 57741767-57741767 on Build GRCh38
rs747719783 Chr.11: 57741829-57741853 on Build GRCh38

View Full Product Details