Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196905221-196905446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR4
Entrez Gene ID
10877
Gene Name
complement factor H related 4
Gene Aliases
CFHL4, FHR-4, FHR4
UniGene
Hs.710100
Gene Chromosome Location
Chr.1: 196888014-196918911 on Build GRCh38
Transcripts
NM_001201550.2 NM_001201551.1 NM_006684.4 XM_006711129.3 XM_017000110.1 XM_017000111.1 XM_017000112.1 XM_017000113.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
146720 c.385G>T p.G129* Chr.1: 196905239-196905239 on Build GRCh38
3480874 c.380C>T p.S127F Chr.1: 196905234-196905234 on Build GRCh38
7145743 c.370G>A p.E124K Chr.1: 196905224-196905224 on Build GRCh38
3480875 c.385G>A p.G129R Chr.1: 196905239-196905239 on Build GRCh38
5126361 c.436+7T>G p.? Chr.1: 196905297-196905297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048177195 Chr.1: 196905371-196905371 on Build GRCh38
rs774966634 Chr.1: 196905272-196905272 on Build GRCh38
rs763765475 Chr.1: 196905311-196905311 on Build GRCh38
rs766719213 Chr.1: 196905297-196905297 on Build GRCh38
rs968228383 Chr.1: 196905413-196905413 on Build GRCh38
rs149436909 Chr.1: 196905364-196905364 on Build GRCh38
rs776016827 Chr.1: 196905253-196905253 on Build GRCh38
rs1045013628 Chr.1: 196905305-196905305 on Build GRCh38
rs573229037 Chr.1: 196905328-196905328 on Build GRCh38
rs762208280 Chr.1: 196905275-196905275 on Build GRCh38
rs779625743 Chr.1: 196905264-196905264 on Build GRCh38
rs892434544 Chr.1: 196905395-196905395 on Build GRCh38
rs751564137 Chr.1: 196905314-196905314 on Build GRCh38
rs757646955 Chr.1: 196905409-196905409 on Build GRCh38
rs760767089 Chr.1: 196905294-196905294 on Build GRCh38
rs563719986 Chr.1: 196905323-196905323 on Build GRCh38
rs772653217 Chr.1: 196905282-196905282 on Build GRCh38
rs749721946 Chr.1: 196905268-196905268 on Build GRCh38
rs558757581 Chr.1: 196905267-196905267 on Build GRCh38
rs540497237 Chr.1: 196905330-196905330 on Build GRCh38
rs762871813 Chr.1: 196905301-196905301 on Build GRCh38
rs1022372979 Chr.1: 196905411-196905411 on Build GRCh38
rs768900805 Chr.1: 196905270-196905270 on Build GRCh38
rs769441598 Chr.1: 196905249-196905249 on Build GRCh38
rs773453214 Chr.1: 196905291-196905291 on Build GRCh38
rs779752642 Chr.1: 196905331-196905331 on Build GRCh38

View Full Product Details