Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Apoe
Entrez Gene ID
100773810
Gene Name
apolipoprotein E
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614150: 265580 - 267440 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505453.2 XP_003505501.1 2 - 3 226 94

View Full Product Details