Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68041183-68041680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCIRG1
Entrez Gene ID
10312
Gene Name
T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3
Gene Aliases
ATP6N1C, ATP6V0A3, Atp6i, OC-116kDa, OC116, OPTB1, Stv1, TIRC7, Vph1, a3
UniGene
Hs.495985
Gene Chromosome Location
Chr.11: 68038995-68053839 on Build GRCh38
Transcripts
NM_006019.3 XM_005273709.3 XM_011544726.2 XM_017017089.1 XM_017017090.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5694588 c.88G>T p.G30C Chr.11: 68041359-68041359 on Build GRCh38
6070241 c.82C>T p.R28W Chr.11: 68041353-68041353 on Build GRCh38
2042421 c.28G>A p.V10M Chr.11: 68041299-68041299 on Build GRCh38
5586766 c.39C>T p.V13V Chr.11: 68041310-68041310 on Build GRCh38
6586930 c.45C>G p.L15L Chr.11: 68041316-68041316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758686699 Chr.11: 68041232-68041232 on Build GRCh38
rs1048500502 Chr.11: 68041554-68041554 on Build GRCh38
rs201972729 Chr.11: 68041279-68041279 on Build GRCh38
rs546761404 Chr.11: 68041621-68041621 on Build GRCh38
rs551897492 Chr.11: 68041208-68041208 on Build GRCh38
rs990180507 Chr.11: 68041469-68041469 on Build GRCh38
rs770787381 Chr.11: 68041363-68041363 on Build GRCh38
rs563996115 Chr.11: 68041287-68041287 on Build GRCh38
rs757721543 Chr.11: 68041332-68041332 on Build GRCh38
rs780455595 Chr.11: 68041408-68041408 on Build GRCh38
rs570336235 Chr.11: 68041472-68041472 on Build GRCh38
rs765426561 Chr.11: 68041305-68041305 on Build GRCh38
rs915427435 Chr.11: 68041281-68041281 on Build GRCh38
rs564296236 Chr.11: 68041575-68041575 on Build GRCh38
rs781759806 Chr.11: 68041336-68041336 on Build GRCh38
rs983456239 Chr.11: 68041601-68041601 on Build GRCh38
rs779555019 Chr.11: 68041347-68041347 on Build GRCh38
rs752849425 Chr.11: 68041320-68041320 on Build GRCh38
rs373244328 Chr.11: 68041292-68041292 on Build GRCh38
rs528516549 Chr.11: 68041614-68041614 on Build GRCh38
rs911791343 Chr.11: 68041604-68041604 on Build GRCh38
rs111424326 Chr.11: 68041331-68041331 on Build GRCh38
rs938436163 Chr.11: 68041510-68041510 on Build GRCh38
rs939907107 Chr.11: 68041585-68041585 on Build GRCh38
rs1038990877 Chr.11: 68041620-68041620 on Build GRCh38
rs775458381 Chr.11: 68041464-68041464 on Build GRCh38
rs748751488 Chr.11: 68041350-68041350 on Build GRCh38
rs768049500 Chr.11: 68041406-68041406 on Build GRCh38
rs758590062 Chr.11: 68041322-68041322 on Build GRCh38
rs999154250 Chr.11: 68041470-68041470 on Build GRCh38
rs753005260 Chr.11: 68041222-68041222 on Build GRCh38
rs775576535 Chr.11: 68041368-68041368 on Build GRCh38
rs1008771663 Chr.11: 68041236-68041236 on Build GRCh38
rs1031917384 Chr.11: 68041329-68041329 on Build GRCh38
rs769465490 Chr.11: 68041268-68041268 on Build GRCh38
rs772995359 Chr.11: 68041254-68041254 on Build GRCh38
rs34506126 Chr.11: 68041464-68041464 on Build GRCh38
rs541552559 Chr.11: 68041541-68041541 on Build GRCh38
rs549532182 Chr.11: 68041362-68041362 on Build GRCh38
rs746440466 Chr.11: 68041337-68041337 on Build GRCh38
rs902167110 Chr.11: 68041468-68041468 on Build GRCh38
rs370896484 Chr.11: 68041415-68041415 on Build GRCh38
rs776470661 Chr.11: 68041533-68041533 on Build GRCh38
rs948415491 Chr.11: 68041552-68041552 on Build GRCh38
rs920726854 Chr.11: 68041570-68041570 on Build GRCh38
rs759582690 Chr.11: 68041302-68041302 on Build GRCh38
rs762979070 Chr.11: 68041369-68041369 on Build GRCh38
rs778259261 Chr.11: 68041246-68041246 on Build GRCh38
rs751881962 Chr.11: 68041392-68041392 on Build GRCh38
rs774416952 Chr.11: 68041346-68041346 on Build GRCh38
rs181288551 Chr.11: 68041514-68041514 on Build GRCh38
rs375645026 Chr.11: 68041438-68041438 on Build GRCh38
rs781772977 Chr.11: 68041258-68041258 on Build GRCh38
rs138949422 Chr.11: 68041632-68041632 on Build GRCh38
rs7116924 Chr.11: 68041471-68041471 on Build GRCh38
rs902261584 Chr.11: 68041641-68041641 on Build GRCh38
rs539777397 Chr.11: 68041449-68041449 on Build GRCh38
rs768203698 Chr.11: 68041288-68041288 on Build GRCh38
rs747914463 Chr.11: 68041354-68041354 on Build GRCh38
rs796582267 Chr.11: 68041446-68041446 on Build GRCh38
rs200524399 Chr.11: 68041407-68041407 on Build GRCh38
rs771896469 Chr.11: 68041358-68041358 on Build GRCh38
rs767392136 Chr.11: 68041298-68041298 on Build GRCh38
rs199548738 Chr.11: 68041463-68041464 on Build GRCh38
rs377303800 Chr.11: 68041389-68041389 on Build GRCh38
rs370255672 Chr.11: 68041439-68041439 on Build GRCh38
rs572050518 Chr.11: 68041630-68041630 on Build GRCh38
rs921757628 Chr.11: 68041624-68041624 on Build GRCh38
rs773182208 Chr.11: 68041300-68041300 on Build GRCh38
rs929362651 Chr.11: 68041374-68041374 on Build GRCh38
rs139397145 Chr.11: 68041353-68041353 on Build GRCh38
rs200201422 Chr.11: 68041265-68041265 on Build GRCh38
rs377034463 Chr.11: 68041330-68041330 on Build GRCh38
rs141859450 Chr.11: 68041361-68041361 on Build GRCh38
rs764330464 Chr.11: 68041388-68041388 on Build GRCh38
rs1023541524 Chr.11: 68041528-68041528 on Build GRCh38
rs998261191 Chr.11: 68041638-68041638 on Build GRCh38
rs574223009 Chr.11: 68041524-68041524 on Build GRCh38
rs537756296 Chr.11: 68041473-68041473 on Build GRCh38
rs763969095 Chr.11: 68041312-68041312 on Build GRCh38
rs764230678 Chr.11: 68041328-68041328 on Build GRCh38
rs928094040 Chr.11: 68041506-68041506 on Build GRCh38
rs565923410 Chr.11: 68041445-68041445 on Build GRCh38

View Full Product Details