Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 71548341-71548834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP5-9
Entrez Gene ID
3846
Gene Name
keratin associated protein 5-9
Gene Aliases
KRN1, KRTAP5-1, KRTAP5.9
UniGene
Hs.709220
Gene Chromosome Location
Chr.11: 71548420-71549607 on Build GRCh38
Transcripts
NM_005553.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6552371 c.99C>T p.P33P Chr.11: 71548756-71548756 on Build GRCh38
4607285 c.17G>C p.C6S Chr.11: 71548674-71548674 on Build GRCh38
6445704 c.132C>T p.P44P Chr.11: 71548789-71548789 on Build GRCh38
2044610 c.114C>T p.P38P Chr.11: 71548771-71548771 on Build GRCh38
6698569 c.68G>T p.S23I Chr.11: 71548725-71548725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189729045 Chr.11: 71548795-71548795 on Build GRCh38
rs114947572 Chr.11: 71548742-71548742 on Build GRCh38
rs371557159 Chr.11: 71548536-71548536 on Build GRCh38
rs373099716 Chr.11: 71548790-71548790 on Build GRCh38
rs777278841 Chr.11: 71548722-71548722 on Build GRCh38
rs34213141 Chr.11: 71548679-71548679 on Build GRCh38
rs553550535 Chr.11: 71548438-71548438 on Build GRCh38
rs562742491 Chr.11: 71548789-71548789 on Build GRCh38
rs752348931 Chr.11: 71548621-71548621 on Build GRCh38
rs767204405 Chr.11: 71548804-71548804 on Build GRCh38
rs911912063 Chr.11: 71548710-71548710 on Build GRCh38
rs749903640 Chr.11: 71548808-71548808 on Build GRCh38
rs917793819 Chr.11: 71548368-71548368 on Build GRCh38
rs987912265 Chr.11: 71548445-71548445 on Build GRCh38
rs760420 Chr.11: 71548456-71548456 on Build GRCh38
rs760566213 Chr.11: 71548775-71548776 on Build GRCh38
rs775169431 Chr.11: 71548656-71548656 on Build GRCh38
rs972641226 Chr.11: 71548393-71548393 on Build GRCh38
rs10792769 Chr.11: 71548776-71548776 on Build GRCh38
rs143860148 Chr.11: 71548611-71548611 on Build GRCh38
rs148620622 Chr.11: 71548622-71548622 on Build GRCh38
rs770486465 Chr.11: 71548643-71548643 on Build GRCh38
rs372093334 Chr.11: 71548712-71548712 on Build GRCh38
rs932000838 Chr.11: 71548420-71548420 on Build GRCh38
rs764088711 Chr.11: 71548776-71548777 on Build GRCh38
rs190769804 Chr.11: 71548490-71548490 on Build GRCh38
rs773945163 Chr.11: 71548678-71548679 on Build GRCh38
rs146869772 Chr.11: 71548772-71548772 on Build GRCh38
rs768180039 Chr.11: 71548403-71548403 on Build GRCh38
rs775132722 Chr.11: 71548753-71548753 on Build GRCh38
rs373000485 Chr.11: 71548808-71548809 on Build GRCh38
rs373649192 Chr.11: 71548754-71548754 on Build GRCh38
rs544644191 Chr.11: 71548777-71548777 on Build GRCh38
rs755520396 Chr.11: 71548367-71548367 on Build GRCh38
rs12791610 Chr.11: 71548770-71548770 on Build GRCh38
rs540343025 Chr.11: 71548467-71548467 on Build GRCh38
rs375818152 Chr.11: 71548767-71548767 on Build GRCh38
rs180808688 Chr.11: 71548663-71548663 on Build GRCh38
rs527854627 Chr.11: 71548651-71548651 on Build GRCh38
rs751226463 Chr.11: 71548723-71548723 on Build GRCh38
rs746825830 Chr.11: 71548658-71548658 on Build GRCh38
rs117909539 Chr.11: 71548514-71548514 on Build GRCh38
rs149916781 Chr.11: 71548482-71548482 on Build GRCh38
rs770459725 Chr.11: 71548617-71548628 on Build GRCh38
rs749031695 Chr.11: 71548760-71548760 on Build GRCh38
rs563238622 Chr.11: 71548537-71548537 on Build GRCh38
rs376944910 Chr.11: 71548731-71548731 on Build GRCh38
rs1005148367 Chr.11: 71548605-71548605 on Build GRCh38
rs542556367 Chr.11: 71548812-71548812 on Build GRCh38
rs537886869 Chr.11: 71548669-71548669 on Build GRCh38
rs752976175 Chr.11: 71548695-71548695 on Build GRCh38
rs765825946 Chr.11: 71548690-71548690 on Build GRCh38
rs185257151 Chr.11: 71548766-71548766 on Build GRCh38
rs906757548 Chr.11: 71548598-71548598 on Build GRCh38
rs972996036 Chr.11: 71548725-71548725 on Build GRCh38
rs759113727 Chr.11: 71548690-71548710 on Build GRCh38
rs894890374 Chr.11: 71548638-71548638 on Build GRCh38
rs1046098874 Chr.11: 71548588-71548588 on Build GRCh38
rs780079719 Chr.11: 71548759-71548759 on Build GRCh38
rs781051251 Chr.11: 71548646-71548646 on Build GRCh38
rs372666324 Chr.11: 71548634-71548634 on Build GRCh38
rs746496967 Chr.11: 71548641-71548641 on Build GRCh38
rs776833376 Chr.11: 71548771-71548771 on Build GRCh38
rs530326729 Chr.11: 71548581-71548581 on Build GRCh38
rs771824071 Chr.11: 71548742-71548743 on Build GRCh38
rs549508385 Chr.11: 71548678-71548678 on Build GRCh38
rs756691342 Chr.11: 71548726-71548726 on Build GRCh38
rs911291710 Chr.11: 71548430-71548430 on Build GRCh38
rs369936376 Chr.11: 71548769-71548769 on Build GRCh38
rs535686093 Chr.11: 71548397-71548397 on Build GRCh38
rs961504915 Chr.11: 71548373-71548373 on Build GRCh38
rs552325299 Chr.11: 71548657-71548657 on Build GRCh38
rs1011880011 Chr.11: 71548580-71548580 on Build GRCh38
rs745812109 Chr.11: 71548650-71548650 on Build GRCh38
rs574635185 Chr.11: 71548394-71548394 on Build GRCh38
rs779179015 Chr.11: 71548612-71548612 on Build GRCh38
rs1043287502 Chr.11: 71548535-71548535 on Build GRCh38

View Full Product Details