Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72089332-72089843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRTOMT
Entrez Gene ID
220074
Gene Name
leucine rich transmembrane and O-methyltransferase domain containing
Gene Aliases
CFAP111, DFNB63, LRRC51
UniGene
Hs.317243
Gene Chromosome Location
Chr.11: 72079982-72110782 on Build GRCh38
Transcripts
NM_001145307.4 NM_001145308.4 NM_001145309.3 NM_001145310.3 NM_001205138.3 NM_001271471.2 NM_001318803.1 NM_145309.5 NR_026886.3 NR_134858.1 XM_006718473.3 XM_006718474.3 XM_011544847.2 XM_011544848.2 XM_017017359.1 XM_017017360.1

SNP Detail

SNP ID Chromosome Location
rs916168686 Chr.11: 72089626-72089626 on Build GRCh38
rs943462395 Chr.11: 72089440-72089445 on Build GRCh38
rs957780385 Chr.11: 72089598-72089598 on Build GRCh38
rs1029606899 Chr.11: 72089402-72089402 on Build GRCh38
rs114066713 Chr.11: 72089773-72089773 on Build GRCh38
rs780745302 Chr.11: 72089379-72089379 on Build GRCh38
rs535220386 Chr.11: 72089720-72089720 on Build GRCh38
rs924682113 Chr.11: 72089593-72089593 on Build GRCh38
rs769012399 Chr.11: 72089395-72089395 on Build GRCh38
rs1044780688 Chr.11: 72089676-72089676 on Build GRCh38
rs990305346 Chr.11: 72089621-72089621 on Build GRCh38
rs564453771 Chr.11: 72089396-72089397 on Build GRCh38
rs779195348 Chr.11: 72089592-72089592 on Build GRCh38
rs149059223 Chr.11: 72089721-72089721 on Build GRCh38
rs576468595 Chr.11: 72089434-72089435 on Build GRCh38
rs955491748 Chr.11: 72089486-72089486 on Build GRCh38
rs567544190 Chr.11: 72089361-72089361 on Build GRCh38
rs535172991 Chr.11: 72089387-72089387 on Build GRCh38
rs758969303 Chr.11: 72089379-72089379 on Build GRCh38
rs757604686 Chr.11: 72089457-72089457 on Build GRCh38
rs747706206 Chr.11: 72089402-72089403 on Build GRCh38
rs1053324502 Chr.11: 72089625-72089625 on Build GRCh38
rs757722975 Chr.11: 72089550-72089551 on Build GRCh38
rs1039053729 Chr.11: 72089441-72089441 on Build GRCh38
rs748292427 Chr.11: 72089602-72089603 on Build GRCh38
rs545241288 Chr.11: 72089649-72089649 on Build GRCh38
rs933461383 Chr.11: 72089612-72089612 on Build GRCh38
rs984806421 Chr.11: 72089418-72089418 on Build GRCh38
rs977475469 Chr.11: 72089561-72089561 on Build GRCh38
rs577947006 Chr.11: 72089416-72089416 on Build GRCh38
rs553481632 Chr.11: 72089393-72089393 on Build GRCh38
rs779156439 Chr.11: 72089407-72089407 on Build GRCh38

View Full Product Details