Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73235097-73235609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P2RY2
Entrez Gene ID
5029
Gene Name
purinergic receptor P2Y2
Gene Aliases
HP2U, P2RU1, P2U, P2U1, P2UR, P2Y2, P2Y2R
UniGene
Hs.339
Gene Chromosome Location
Chr.11: 73200416-73246743 on Build GRCh38
Transcripts
NM_002564.3 NM_176071.2 NM_176072.2 XM_005274019.4 XM_005274020.4 XM_005274021.4 XM_011545074.2 XM_017017839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6014563 c.1051G>C p.D351H Chr.11: 73235210-73235210 on Build GRCh38
6736504 c.1089G>A p.E363E Chr.11: 73235248-73235248 on Build GRCh38
3752720 c.1000C>T p.R334C Chr.11: 73235159-73235159 on Build GRCh38
6736489 c.1082G>A p.R361Q Chr.11: 73235241-73235241 on Build GRCh38
4896128 c.1092C>T p.S364S Chr.11: 73235251-73235251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902380731 Chr.11: 73235542-73235542 on Build GRCh38
rs759522848 Chr.11: 73235315-73235315 on Build GRCh38
rs764293969 Chr.11: 73235119-73235119 on Build GRCh38
rs765534001 Chr.11: 73235267-73235267 on Build GRCh38
rs769990257 Chr.11: 73235155-73235156 on Build GRCh38
rs756855478 Chr.11: 73235278-73235278 on Build GRCh38
rs780682826 Chr.11: 73235282-73235282 on Build GRCh38
rs148042673 Chr.11: 73235286-73235286 on Build GRCh38
rs751238150 Chr.11: 73235277-73235277 on Build GRCh38
rs773395396 Chr.11: 73235343-73235343 on Build GRCh38
rs1039650890 Chr.11: 73235477-73235477 on Build GRCh38
rs147574123 Chr.11: 73235361-73235361 on Build GRCh38
rs74472890 Chr.11: 73235234-73235234 on Build GRCh38
rs749119899 Chr.11: 73235293-73235293 on Build GRCh38
rs999309871 Chr.11: 73235290-73235290 on Build GRCh38
rs766911840 Chr.11: 73235178-73235178 on Build GRCh38
rs982707039 Chr.11: 73235408-73235408 on Build GRCh38
rs1032500962 Chr.11: 73235295-73235295 on Build GRCh38
rs113302517 Chr.11: 73235145-73235145 on Build GRCh38
rs777557502 Chr.11: 73235238-73235238 on Build GRCh38
rs768416986 Chr.11: 73235294-73235294 on Build GRCh38
rs145119561 Chr.11: 73235266-73235266 on Build GRCh38
rs767543372 Chr.11: 73235141-73235141 on Build GRCh38
rs767594635 Chr.11: 73235164-73235164 on Build GRCh38
rs750931409 Chr.11: 73235144-73235144 on Build GRCh38
rs367746501 Chr.11: 73235344-73235344 on Build GRCh38
rs907575836 Chr.11: 73235471-73235471 on Build GRCh38
rs201458131 Chr.11: 73235157-73235157 on Build GRCh38
rs141485889 Chr.11: 73235256-73235256 on Build GRCh38
rs900382387 Chr.11: 73235129-73235129 on Build GRCh38
rs751144768 Chr.11: 73235292-73235292 on Build GRCh38
rs781122178 Chr.11: 73235445-73235445 on Build GRCh38
rs752299195 Chr.11: 73235218-73235218 on Build GRCh38
rs375404183 Chr.11: 73235206-73235208 on Build GRCh38
rs747192977 Chr.11: 73235303-73235303 on Build GRCh38
rs769641333 Chr.11: 73235509-73235509 on Build GRCh38
rs909928719 Chr.11: 73235458-73235458 on Build GRCh38
rs758832801 Chr.11: 73235273-73235273 on Build GRCh38
rs773391543 Chr.11: 73235257-73235257 on Build GRCh38
rs974326392 Chr.11: 73235481-73235481 on Build GRCh38
rs17244555 Chr.11: 73235263-73235263 on Build GRCh38
rs188419665 Chr.11: 73235490-73235490 on Build GRCh38
rs771515142 Chr.11: 73235156-73235156 on Build GRCh38
rs773444523 Chr.11: 73235298-73235298 on Build GRCh38
rs777612853 Chr.11: 73235276-73235276 on Build GRCh38
rs150017416 Chr.11: 73235152-73235152 on Build GRCh38
rs192691668 Chr.11: 73235561-73235561 on Build GRCh38
rs544831543 Chr.11: 73235360-73235360 on Build GRCh38
rs138929283 Chr.11: 73235253-73235253 on Build GRCh38
rs900443102 Chr.11: 73235478-73235478 on Build GRCh38
rs942711835 Chr.11: 73235475-73235475 on Build GRCh38
rs775834297 Chr.11: 73235326-73235326 on Build GRCh38
rs538740504 Chr.11: 73235265-73235265 on Build GRCh38
rs754347715 Chr.11: 73235182-73235182 on Build GRCh38
rs753166635 Chr.11: 73235268-73235268 on Build GRCh38
rs1047362736 Chr.11: 73235250-73235250 on Build GRCh38
rs990449423 Chr.11: 73235354-73235354 on Build GRCh38
rs750558013 Chr.11: 73235166-73235166 on Build GRCh38
rs763463265 Chr.11: 73235334-73235334 on Build GRCh38
rs113741989 Chr.11: 73235160-73235160 on Build GRCh38
rs140659510 Chr.11: 73235563-73235563 on Build GRCh38
rs944173286 Chr.11: 73235577-73235577 on Build GRCh38
rs752715769 Chr.11: 73235453-73235453 on Build GRCh38
rs746589198 Chr.11: 73235241-73235241 on Build GRCh38
rs751779387 Chr.11: 73235130-73235130 on Build GRCh38
rs143795035 Chr.11: 73235135-73235135 on Build GRCh38
rs997264796 Chr.11: 73235131-73235131 on Build GRCh38
rs1025157472 Chr.11: 73235426-73235426 on Build GRCh38
rs769478974 Chr.11: 73235255-73235255 on Build GRCh38
rs771003497 Chr.11: 73235312-73235312 on Build GRCh38
rs754463063 Chr.11: 73235394-73235394 on Build GRCh38
rs200977338 Chr.11: 73235151-73235151 on Build GRCh38
rs745774369 Chr.11: 73235254-73235254 on Build GRCh38
rs949536788 Chr.11: 73235202-73235202 on Build GRCh38
rs780523873 Chr.11: 73235150-73235150 on Build GRCh38
rs77179176 Chr.11: 73235541-73235541 on Build GRCh38
rs917635166 Chr.11: 73235195-73235195 on Build GRCh38
rs745804553 Chr.11: 73235285-73235285 on Build GRCh38
rs776294740 Chr.11: 73235412-73235412 on Build GRCh38
rs367926847 Chr.11: 73235576-73235576 on Build GRCh38
rs776717413 Chr.11: 73235313-73235313 on Build GRCh38
rs551184201 Chr.11: 73235586-73235586 on Build GRCh38
rs536206854 Chr.11: 73235162-73235162 on Build GRCh38
rs146076516 Chr.11: 73235252-73235252 on Build GRCh38
rs151310960 Chr.11: 73235143-73235143 on Build GRCh38
rs933274682 Chr.11: 73235489-73235489 on Build GRCh38
rs765842225 Chr.11: 73235322-73235322 on Build GRCh38
rs1626154 Chr.11: 73235159-73235159 on Build GRCh38
rs779296583 Chr.11: 73235196-73235196 on Build GRCh38
rs755322202 Chr.11: 73235183-73235183 on Build GRCh38
rs200033279 Chr.11: 73235175-73235175 on Build GRCh38

View Full Product Details