Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75421979-75422502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL35
Entrez Gene ID
283212
Gene Name
kelch like family member 35
Gene Aliases
-
UniGene
Hs.292451
Gene Chromosome Location
Chr.11: 75422394-75434550 on Build GRCh38
Transcripts
NM_001039548.2 XM_011544954.2 NM_001256802.1

Gene Symbol
RPS3
Entrez Gene ID
6188
Gene Name
ribosomal protein S3
Gene Aliases
S3
UniGene
Hs.546286
Gene Chromosome Location
Chr.11: 75399491-75422302 on Build GRCh38
Transcripts
NM_001039548.2 XM_011544954.2 NM_001256802.1

SNP Detail

SNP ID Chromosome Location
rs1009207692 Chr.11: 75422069-75422069 on Build GRCh38
rs779171886 Chr.11: 75422403-75422403 on Build GRCh38
rs1030514998 Chr.11: 75422132-75422132 on Build GRCh38
rs867615071 Chr.11: 75422194-75422196 on Build GRCh38
rs147393088 Chr.11: 75422311-75422311 on Build GRCh38
rs940134978 Chr.11: 75422471-75422471 on Build GRCh38
rs1023514868 Chr.11: 75422415-75422415 on Build GRCh38
rs574321168 Chr.11: 75422195-75422198 on Build GRCh38
rs572953922 Chr.11: 75422208-75422208 on Build GRCh38
rs763093779 Chr.11: 75422278-75422278 on Build GRCh38
rs950794210 Chr.11: 75422258-75422258 on Build GRCh38
rs11236425 Chr.11: 75422047-75422047 on Build GRCh38
rs762131876 Chr.11: 75422130-75422131 on Build GRCh38
rs578055928 Chr.11: 75422381-75422381 on Build GRCh38
rs898649122 Chr.11: 75422186-75422186 on Build GRCh38
rs11236426 Chr.11: 75422215-75422215 on Build GRCh38
rs565444012 Chr.11: 75422276-75422276 on Build GRCh38
rs566656890 Chr.11: 75422395-75422395 on Build GRCh38
rs556630043 Chr.11: 75422361-75422361 on Build GRCh38
rs768622406 Chr.11: 75422015-75422015 on Build GRCh38
rs940082189 Chr.11: 75422063-75422063 on Build GRCh38
rs182453601 Chr.11: 75422446-75422446 on Build GRCh38
rs77761323 Chr.11: 75422451-75422451 on Build GRCh38
rs537502003 Chr.11: 75422425-75422425 on Build GRCh38
rs945042261 Chr.11: 75422432-75422432 on Build GRCh38
rs1026206759 Chr.11: 75422245-75422245 on Build GRCh38
rs890239620 Chr.11: 75422461-75422461 on Build GRCh38
rs991020799 Chr.11: 75422413-75422413 on Build GRCh38
rs890175375 Chr.11: 75422148-75422148 on Build GRCh38
rs776479617 Chr.11: 75422048-75422048 on Build GRCh38
rs149951016 Chr.11: 75422197-75422199 on Build GRCh38
rs1053314601 Chr.11: 75422349-75422349 on Build GRCh38
rs917329303 Chr.11: 75422300-75422300 on Build GRCh38
rs138126692 Chr.11: 75422338-75422338 on Build GRCh38
rs1050013938 Chr.11: 75422208-75422211 on Build GRCh38
rs1017443978 Chr.11: 75422151-75422151 on Build GRCh38
rs143670546 Chr.11: 75422341-75422341 on Build GRCh38
rs766527047 Chr.11: 75422340-75422340 on Build GRCh38
rs542635003 Chr.11: 75422044-75422044 on Build GRCh38
rs546565823 Chr.11: 75422394-75422394 on Build GRCh38
rs992803115 Chr.11: 75422269-75422269 on Build GRCh38
rs981426266 Chr.11: 75422449-75422449 on Build GRCh38
rs904818423 Chr.11: 75422036-75422036 on Build GRCh38
rs866950735 Chr.11: 75422225-75422225 on Build GRCh38
rs1007296270 Chr.11: 75422477-75422477 on Build GRCh38
rs530079188 Chr.11: 75422368-75422368 on Build GRCh38
rs1007430528 Chr.11: 75422149-75422149 on Build GRCh38
rs757337396 Chr.11: 75422169-75422169 on Build GRCh38

View Full Product Details